Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87348768-87349342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROT
Entrez Gene ID
54677
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
COT
UniGene
Hs.125039
Gene Chromosome Location
Chr.7: 87345635-87399796 on Build GRCh38
Transcripts
NM_001143935.1 NM_001243745.1 NM_021151.3 XM_011516337.2 XM_017012370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6178521 c.104A>C p.Y35S Chr.7: 87349172-87349172 on Build GRCh38
1643397 c.96delA p.K34fs*6 Chr.7: 87349164-87349164 on Build GRCh38
4674769 c.95_96insA p.Y35fs*3 Chr.7: 87349163-87349164 on Build GRCh38
1452551 c.62C>A p.P21Q Chr.7: 87349130-87349130 on Build GRCh38
1202322 c.88G>T p.E30* Chr.7: 87349156-87349156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760951827 Chr.7: 87349081-87349081 on Build GRCh38
rs113592238 Chr.7: 87349158-87349158 on Build GRCh38
rs1044850408 Chr.7: 87349244-87349244 on Build GRCh38
rs752674828 Chr.7: 87348982-87348982 on Build GRCh38
rs535344381 Chr.7: 87348996-87348996 on Build GRCh38
rs11971666 Chr.7: 87349238-87349238 on Build GRCh38
rs770594340 Chr.7: 87349131-87349131 on Build GRCh38
rs555931620 Chr.7: 87348966-87348966 on Build GRCh38
rs140842977 Chr.7: 87349102-87349102 on Build GRCh38
rs147240633 Chr.7: 87349084-87349084 on Build GRCh38
rs756051146 Chr.7: 87349003-87349003 on Build GRCh38
rs756999481 Chr.7: 87349164-87349164 on Build GRCh38
rs752943012 Chr.7: 87349069-87349069 on Build GRCh38
rs183177301 Chr.7: 87348824-87348824 on Build GRCh38
rs1004228468 Chr.7: 87348866-87348867 on Build GRCh38
rs777583546 Chr.7: 87349224-87349224 on Build GRCh38
rs551460924 Chr.7: 87348928-87348928 on Build GRCh38
rs779947503 Chr.7: 87349029-87349030 on Build GRCh38
rs1013293657 Chr.7: 87348951-87348951 on Build GRCh38
rs925595133 Chr.7: 87349194-87349194 on Build GRCh38
rs754075882 Chr.7: 87349089-87349089 on Build GRCh38
rs1031245744 Chr.7: 87348868-87348868 on Build GRCh38
rs775037264 Chr.7: 87349041-87349041 on Build GRCh38
rs757379228 Chr.7: 87349092-87349092 on Build GRCh38
rs997362808 Chr.7: 87349275-87349275 on Build GRCh38
rs201700761 Chr.7: 87349139-87349139 on Build GRCh38
rs77107729 Chr.7: 87348945-87348945 on Build GRCh38
rs920413890 Chr.7: 87349024-87349024 on Build GRCh38
rs142380435 Chr.7: 87348931-87348931 on Build GRCh38
rs778936531 Chr.7: 87349100-87349100 on Build GRCh38
rs772473391 Chr.7: 87349274-87349274 on Build GRCh38
rs1038014405 Chr.7: 87349305-87349305 on Build GRCh38
rs947789597 Chr.7: 87348836-87348836 on Build GRCh38
rs754865059 Chr.7: 87349103-87349103 on Build GRCh38
rs769627937 Chr.7: 87349113-87349113 on Build GRCh38
rs955383485 Chr.7: 87348992-87348992 on Build GRCh38
rs776045920 Chr.7: 87349310-87349310 on Build GRCh38
rs748762185 Chr.7: 87349051-87349051 on Build GRCh38
rs777350070 Chr.7: 87349116-87349116 on Build GRCh38
rs369691197 Chr.7: 87349105-87349105 on Build GRCh38
rs1045085841 Chr.7: 87348983-87348983 on Build GRCh38
rs761000290 Chr.7: 87349164-87349164 on Build GRCh38
rs935524956 Chr.7: 87348896-87348896 on Build GRCh38
rs765963910 Chr.7: 87349036-87349036 on Build GRCh38
rs975493496 Chr.7: 87348941-87348941 on Build GRCh38
rs955590002 Chr.7: 87349315-87349315 on Build GRCh38
rs940565611 Chr.7: 87349004-87349004 on Build GRCh38
rs575414855 Chr.7: 87349146-87349146 on Build GRCh38
rs778596623 Chr.7: 87349055-87349055 on Build GRCh38
rs776057015 Chr.7: 87349150-87349150 on Build GRCh38
rs201394030 Chr.7: 87349174-87349174 on Build GRCh38
rs374673999 Chr.7: 87349184-87349184 on Build GRCh38
rs906176794 Chr.7: 87348853-87348853 on Build GRCh38
rs1022432964 Chr.7: 87348900-87348900 on Build GRCh38
rs143103247 Chr.7: 87349142-87349142 on Build GRCh38
rs1006539260 Chr.7: 87349008-87349008 on Build GRCh38
rs368674447 Chr.7: 87349208-87349208 on Build GRCh38
rs940513182 Chr.7: 87348799-87348799 on Build GRCh38
rs547437928 Chr.7: 87348820-87348820 on Build GRCh38
rs765327773 Chr.7: 87349198-87349198 on Build GRCh38
rs540500046 Chr.7: 87349319-87349319 on Build GRCh38
rs375446390 Chr.7: 87349053-87349053 on Build GRCh38
rs748938256 Chr.7: 87349121-87349121 on Build GRCh38
rs1016206004 Chr.7: 87349021-87349021 on Build GRCh38
rs1043994735 Chr.7: 87348854-87348854 on Build GRCh38
rs1030180624 Chr.7: 87349119-87349119 on Build GRCh38
rs747831338 Chr.7: 87349107-87349107 on Build GRCh38
rs911228444 Chr.7: 87348818-87348818 on Build GRCh38
rs935493924 Chr.7: 87349196-87349196 on Build GRCh38
rs918086998 Chr.7: 87349226-87349226 on Build GRCh38
rs567836795 Chr.7: 87348995-87348995 on Build GRCh38
rs113489014 Chr.7: 87348793-87348793 on Build GRCh38
rs754050219 Chr.7: 87349176-87349176 on Build GRCh38
rs904840373 Chr.7: 87349264-87349264 on Build GRCh38
rs781060257 Chr.7: 87349206-87349206 on Build GRCh38
rs1037444636 Chr.7: 87348804-87348804 on Build GRCh38
rs753873696 Chr.7: 87349029-87349032 on Build GRCh38
rs993419266 Chr.7: 87349312-87349312 on Build GRCh38
rs1052726930 Chr.7: 87349172-87349172 on Build GRCh38
rs802037 Chr.7: 87349031-87349031 on Build GRCh38
rs558249553 Chr.7: 87348895-87348895 on Build GRCh38
rs762439177 Chr.7: 87349033-87349033 on Build GRCh38
rs755900759 Chr.7: 87349216-87349216 on Build GRCh38
rs892673445 Chr.7: 87348883-87348883 on Build GRCh38
rs955405514 Chr.7: 87349135-87349135 on Build GRCh38
rs772636771 Chr.7: 87349149-87349149 on Build GRCh38
rs1045144433 Chr.7: 87348842-87348842 on Build GRCh38
rs750445851 Chr.7: 87349203-87349203 on Build GRCh38
rs911185532 Chr.7: 87349148-87349148 on Build GRCh38
rs371392044 Chr.7: 87349144-87349144 on Build GRCh38

View Full Product Details