Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92458182-92458712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATAD1
Entrez Gene ID
57798
Gene Name
GATA zinc finger domain containing 1
Gene Aliases
CMD2B, ODAG, RG083M05.2
UniGene
Hs.21145
Gene Chromosome Location
Chr.7: 92447448-92494631 on Build GRCh38
Transcripts
NM_021167.4 NR_052016.1

SNP Detail

SNP ID Chromosome Location
rs867643251 Chr.7: 92458403-92458403 on Build GRCh38
rs183702741 Chr.7: 92458601-92458601 on Build GRCh38
rs563989578 Chr.7: 92458254-92458254 on Build GRCh38
rs919212143 Chr.7: 92458630-92458630 on Build GRCh38
rs1053760038 Chr.7: 92458302-92458302 on Build GRCh38
rs898936561 Chr.7: 92458229-92458229 on Build GRCh38
rs930415634 Chr.7: 92458538-92458538 on Build GRCh38
rs893984226 Chr.7: 92458339-92458339 on Build GRCh38
rs561085755 Chr.7: 92458436-92458436 on Build GRCh38
rs549731604 Chr.7: 92458674-92458674 on Build GRCh38
rs552759747 Chr.7: 92458508-92458508 on Build GRCh38
rs551892949 Chr.7: 92458528-92458528 on Build GRCh38
rs367677614 Chr.7: 92458206-92458206 on Build GRCh38
rs906760062 Chr.7: 92458389-92458389 on Build GRCh38
rs148717683 Chr.7: 92458307-92458307 on Build GRCh38
rs1050095487 Chr.7: 92458547-92458547 on Build GRCh38
rs1018782100 Chr.7: 92458355-92458355 on Build GRCh38
rs776577594 Chr.7: 92458529-92458529 on Build GRCh38
rs34343533 Chr.7: 92458244-92458244 on Build GRCh38
rs559783173 Chr.7: 92458318-92458318 on Build GRCh38
rs915917244 Chr.7: 92458411-92458411 on Build GRCh38
rs191480624 Chr.7: 92458507-92458507 on Build GRCh38
rs573181080 Chr.7: 92458652-92458652 on Build GRCh38
rs967310455 Chr.7: 92458363-92458363 on Build GRCh38
rs971355222 Chr.7: 92458417-92458417 on Build GRCh38
rs1038973862 Chr.7: 92458484-92458484 on Build GRCh38
rs747959207 Chr.7: 92458353-92458353 on Build GRCh38
rs981433547 Chr.7: 92458424-92458424 on Build GRCh38
rs371632143 Chr.7: 92458591-92458591 on Build GRCh38
rs773865764 Chr.7: 92458472-92458472 on Build GRCh38
rs188250836 Chr.7: 92458686-92458686 on Build GRCh38
rs994535367 Chr.7: 92458244-92458244 on Build GRCh38
rs1032826843 Chr.7: 92458394-92458394 on Build GRCh38
rs888756834 Chr.7: 92458260-92458260 on Build GRCh38
rs777739151 Chr.7: 92458409-92458409 on Build GRCh38
rs1021246357 Chr.7: 92458366-92458366 on Build GRCh38
rs1002570074 Chr.7: 92458401-92458401 on Build GRCh38
rs34606827 Chr.7: 92458550-92458551 on Build GRCh38
rs770429958 Chr.7: 92458451-92458451 on Build GRCh38
rs943475657 Chr.7: 92458276-92458276 on Build GRCh38
rs940269522 Chr.7: 92458454-92458454 on Build GRCh38
rs769530251 Chr.7: 92458364-92458364 on Build GRCh38
rs574276877 Chr.7: 92458303-92458303 on Build GRCh38
rs888787863 Chr.7: 92458641-92458641 on Build GRCh38
rs745459455 Chr.7: 92458551-92458551 on Build GRCh38

View Full Product Details