Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117367166-117367657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
UniGene
Hs.352412
Gene Chromosome Location
Chr.7: 117363222-117427523 on Build GRCh38
Transcripts
NM_001301821.1 NM_001301822.1 NM_130768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
744028 c.1236G>A p.L412L Chr.7: 117367391-117367391 on Build GRCh38
6175954 c.1252A>G p.K418E Chr.7: 117367375-117367375 on Build GRCh38
1084456 c.1231G>A p.D411N Chr.7: 117367396-117367396 on Build GRCh38
3632265 c.1270C>T p.Q424* Chr.7: 117367357-117367357 on Build GRCh38
6377811 c.1183C>A p.P395T Chr.7: 117367444-117367444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035040624 Chr.7: 117367264-117367264 on Build GRCh38
rs775953490 Chr.7: 117367411-117367411 on Build GRCh38
rs10267003 Chr.7: 117367272-117367272 on Build GRCh38
rs999585271 Chr.7: 117367242-117367242 on Build GRCh38
rs753403646 Chr.7: 117367487-117367487 on Build GRCh38
rs990053767 Chr.7: 117367372-117367372 on Build GRCh38
rs779739765 Chr.7: 117367325-117367325 on Build GRCh38
rs771957825 Chr.7: 117367378-117367378 on Build GRCh38
rs759616826 Chr.7: 117367393-117367393 on Build GRCh38
rs755624948 Chr.7: 117367323-117367323 on Build GRCh38
rs973791907 Chr.7: 117367564-117367564 on Build GRCh38
rs765575245 Chr.7: 117367405-117367405 on Build GRCh38
rs747958047 Chr.7: 117367341-117367341 on Build GRCh38
rs1015052778 Chr.7: 117367262-117367262 on Build GRCh38
rs938324246 Chr.7: 117367586-117367586 on Build GRCh38
rs748957936 Chr.7: 117367326-117367326 on Build GRCh38
rs770985427 Chr.7: 117367385-117367385 on Build GRCh38
rs765742708 Chr.7: 117367479-117367479 on Build GRCh38
rs10237738 Chr.7: 117367632-117367632 on Build GRCh38
rs372367007 Chr.7: 117367432-117367432 on Build GRCh38
rs773182626 Chr.7: 117367381-117367381 on Build GRCh38
rs977131814 Chr.7: 117367518-117367518 on Build GRCh38
rs551382056 Chr.7: 117367577-117367577 on Build GRCh38
rs188189009 Chr.7: 117367505-117367505 on Build GRCh38
rs953332440 Chr.7: 117367363-117367363 on Build GRCh38
rs368462441 Chr.7: 117367387-117367387 on Build GRCh38
rs768358316 Chr.7: 117367327-117367327 on Build GRCh38
rs541234914 Chr.7: 117367277-117367277 on Build GRCh38
rs1029299410 Chr.7: 117367331-117367331 on Build GRCh38
rs749913493 Chr.7: 117367319-117367319 on Build GRCh38
rs887971408 Chr.7: 117367231-117367231 on Build GRCh38
rs35806172 Chr.7: 117367493-117367494 on Build GRCh38
rs559240651 Chr.7: 117367297-117367297 on Build GRCh38
rs191306118 Chr.7: 117367606-117367606 on Build GRCh38
rs966404922 Chr.7: 117367324-117367324 on Build GRCh38
rs879567660 Chr.7: 117367614-117367614 on Build GRCh38
rs918227028 Chr.7: 117367569-117367569 on Build GRCh38
rs902620871 Chr.7: 117367218-117367218 on Build GRCh38
rs756655644 Chr.7: 117367187-117367187 on Build GRCh38
rs754576966 Chr.7: 117367499-117367499 on Build GRCh38
rs147994188 Chr.7: 117367442-117367442 on Build GRCh38
rs778660056 Chr.7: 117367330-117367330 on Build GRCh38
rs755648802 Chr.7: 117367454-117367454 on Build GRCh38
rs747916408 Chr.7: 117367516-117367516 on Build GRCh38
rs951003439 Chr.7: 117367571-117367571 on Build GRCh38
rs960867787 Chr.7: 117367307-117367307 on Build GRCh38

View Full Product Details