Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42768230-42768804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA6
Entrez Gene ID
8973
Gene Name
cholinergic receptor nicotinic alpha 6 subunit
Gene Aliases
CHNRA6
UniGene
Hs.103128
Gene Chromosome Location
Chr.8: 42752620-42768786 on Build GRCh38
Transcripts
NM_001199279.1 NM_004198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100072 c.64A>G p.T22A Chr.8: 42768367-42768367 on Build GRCh38
4775979 c.22G>A p.G8R Chr.8: 42768409-42768409 on Build GRCh38
5633218 c.79+2T>C p.? Chr.8: 42768350-42768350 on Build GRCh38
6628736 c.43T>C p.C15R Chr.8: 42768388-42768388 on Build GRCh38
3649369 c.79G>A p.G27S Chr.8: 42768352-42768352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577944510 Chr.8: 42768521-42768521 on Build GRCh38
rs138325954 Chr.8: 42768544-42768544 on Build GRCh38
rs746907748 Chr.8: 42768464-42768465 on Build GRCh38
rs756800424 Chr.8: 42768251-42768251 on Build GRCh38
rs371777471 Chr.8: 42768329-42768329 on Build GRCh38
rs991049194 Chr.8: 42768546-42768546 on Build GRCh38
rs1011951746 Chr.8: 42768754-42768754 on Build GRCh38
rs780962662 Chr.8: 42768425-42768425 on Build GRCh38
rs200256551 Chr.8: 42768496-42768496 on Build GRCh38
rs952380979 Chr.8: 42768381-42768381 on Build GRCh38
rs200229186 Chr.8: 42768388-42768388 on Build GRCh38
rs777686049 Chr.8: 42768462-42768462 on Build GRCh38
rs199914382 Chr.8: 42768329-42768329 on Build GRCh38
rs1012412400 Chr.8: 42768385-42768385 on Build GRCh38
rs757266530 Chr.8: 42768468-42768468 on Build GRCh38
rs181221048 Chr.8: 42768643-42768643 on Build GRCh38
rs780406657 Chr.8: 42768710-42768710 on Build GRCh38
rs956954467 Chr.8: 42768433-42768433 on Build GRCh38
rs558773489 Chr.8: 42768685-42768685 on Build GRCh38
rs772320560 Chr.8: 42768476-42768476 on Build GRCh38
rs202025303 Chr.8: 42768387-42768387 on Build GRCh38
rs538869899 Chr.8: 42768668-42768668 on Build GRCh38
rs3217406 Chr.8: 42768330-42768330 on Build GRCh38
rs373533539 Chr.8: 42768351-42768351 on Build GRCh38
rs141199708 Chr.8: 42768353-42768353 on Build GRCh38
rs757808637 Chr.8: 42768460-42768460 on Build GRCh38
rs1025396279 Chr.8: 42768699-42768699 on Build GRCh38
rs889808335 Chr.8: 42768644-42768644 on Build GRCh38
rs115773120 Chr.8: 42768543-42768543 on Build GRCh38
rs1040842914 Chr.8: 42768257-42768257 on Build GRCh38
rs201227941 Chr.8: 42768462-42768462 on Build GRCh38
rs202231701 Chr.8: 42768373-42768373 on Build GRCh38
rs889660385 Chr.8: 42768260-42768260 on Build GRCh38
rs779430380 Chr.8: 42768576-42768576 on Build GRCh38
rs779268752 Chr.8: 42768469-42768469 on Build GRCh38
rs751417062 Chr.8: 42768397-42768397 on Build GRCh38
rs758191044 Chr.8: 42768447-42768448 on Build GRCh38
rs571650824 Chr.8: 42768532-42768532 on Build GRCh38
rs200609466 Chr.8: 42768648-42768650 on Build GRCh38
rs746287408 Chr.8: 42768471-42768471 on Build GRCh38
rs569402191 Chr.8: 42768493-42768493 on Build GRCh38
rs187077675 Chr.8: 42768708-42768708 on Build GRCh38
rs144055326 Chr.8: 42768588-42768588 on Build GRCh38
rs147326892 Chr.8: 42768628-42768628 on Build GRCh38
rs551545999 Chr.8: 42768443-42768443 on Build GRCh38
rs754696130 Chr.8: 42768399-42768399 on Build GRCh38
rs1034405693 Chr.8: 42768768-42768768 on Build GRCh38
rs984153198 Chr.8: 42768535-42768535 on Build GRCh38
rs201844732 Chr.8: 42768341-42768341 on Build GRCh38
rs760813110 Chr.8: 42768347-42768347 on Build GRCh38
rs888359123 Chr.8: 42768749-42768749 on Build GRCh38
rs942590083 Chr.8: 42768647-42768647 on Build GRCh38
rs369103835 Chr.8: 42768306-42768306 on Build GRCh38
rs752429073 Chr.8: 42768374-42768385 on Build GRCh38
rs1031157903 Chr.8: 42768432-42768432 on Build GRCh38
rs199545170 Chr.8: 42768506-42768506 on Build GRCh38
rs752382512 Chr.8: 42768429-42768429 on Build GRCh38
rs879915495 Chr.8: 42768571-42768571 on Build GRCh38
rs1025428841 Chr.8: 42768474-42768474 on Build GRCh38
rs1006814432 Chr.8: 42768298-42768298 on Build GRCh38
rs748770519 Chr.8: 42768454-42768454 on Build GRCh38
rs755766383 Chr.8: 42768442-42768442 on Build GRCh38
rs150939999 Chr.8: 42768382-42768382 on Build GRCh38
rs80020392 Chr.8: 42768348-42768348 on Build GRCh38
rs201800849 Chr.8: 42768494-42768494 on Build GRCh38
rs200023923 Chr.8: 42768375-42768375 on Build GRCh38
rs200827899 Chr.8: 42768330-42768330 on Build GRCh38
rs34444353 Chr.8: 42768578-42768579 on Build GRCh38
rs201329722 Chr.8: 42768500-42768500 on Build GRCh38
rs970878049 Chr.8: 42768420-42768420 on Build GRCh38
rs1023711467 Chr.8: 42768758-42768758 on Build GRCh38
rs900994735 Chr.8: 42768674-42768674 on Build GRCh38
rs1005313228 Chr.8: 42768715-42768715 on Build GRCh38
rs1054165048 Chr.8: 42768630-42768630 on Build GRCh38
rs924064806 Chr.8: 42768614-42768614 on Build GRCh38

View Full Product Details