Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 43069525-43069982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNTA
Entrez Gene ID
2339
Gene Name
farnesyltransferase, CAAX box, alpha
Gene Aliases
FPTA, PGGT1A, PTAR2
UniGene
Hs.370312
Gene Chromosome Location
Chr.8: 43056299-43085788 on Build GRCh38
Transcripts
NM_002027.2 NR_033698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
750300 c.418C>G p.L140V Chr.8: 43069571-43069571 on Build GRCh38
1457205 c.495C>A p.N165K Chr.8: 43069648-43069648 on Build GRCh38
750299 c.434A>T p.Q145L Chr.8: 43069587-43069587 on Build GRCh38
328728 c.497A>G p.Y166C Chr.8: 43069650-43069650 on Build GRCh38
4767923 c.407T>C p.F136S Chr.8: 43069560-43069560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040201787 Chr.8: 43069782-43069782 on Build GRCh38
rs759745062 Chr.8: 43069624-43069624 on Build GRCh38
rs183381340 Chr.8: 43069864-43069864 on Build GRCh38
rs554850498 Chr.8: 43069883-43069883 on Build GRCh38
rs778274499 Chr.8: 43069592-43069592 on Build GRCh38
rs868738223 Chr.8: 43069609-43069609 on Build GRCh38
rs1034119333 Chr.8: 43069833-43069833 on Build GRCh38
rs556242132 Chr.8: 43069625-43069625 on Build GRCh38
rs751361863 Chr.8: 43069677-43069677 on Build GRCh38
rs1034197858 Chr.8: 43069942-43069942 on Build GRCh38
rs780631274 Chr.8: 43069579-43069579 on Build GRCh38
rs565962358 Chr.8: 43069861-43069861 on Build GRCh38
rs552520151 Chr.8: 43069810-43069810 on Build GRCh38
rs538788108 Chr.8: 43069733-43069733 on Build GRCh38
rs779959783 Chr.8: 43069551-43069551 on Build GRCh38
rs866848968 Chr.8: 43069832-43069832 on Build GRCh38
rs1054518251 Chr.8: 43069791-43069791 on Build GRCh38
rs377283893 Chr.8: 43069685-43069685 on Build GRCh38
rs771334589 Chr.8: 43069599-43069599 on Build GRCh38
rs754660986 Chr.8: 43069678-43069678 on Build GRCh38
rs201464015 Chr.8: 43069577-43069577 on Build GRCh38
rs895217195 Chr.8: 43069835-43069835 on Build GRCh38
rs750430244 Chr.8: 43069667-43069667 on Build GRCh38
rs778886995 Chr.8: 43069771-43069771 on Build GRCh38
rs775519626 Chr.8: 43069641-43069641 on Build GRCh38
rs377369408 Chr.8: 43069755-43069755 on Build GRCh38
rs200780088 Chr.8: 43069562-43069562 on Build GRCh38
rs929185641 Chr.8: 43069792-43069792 on Build GRCh38
rs754643572 Chr.8: 43069561-43069561 on Build GRCh38
rs750443496 Chr.8: 43069633-43069633 on Build GRCh38
rs369603001 Chr.8: 43069690-43069690 on Build GRCh38
rs766141475 Chr.8: 43069676-43069676 on Build GRCh38
rs762958614 Chr.8: 43069670-43069670 on Build GRCh38
rs978378765 Chr.8: 43069713-43069713 on Build GRCh38
rs559113793 Chr.8: 43069595-43069595 on Build GRCh38
rs949851202 Chr.8: 43069752-43069752 on Build GRCh38
rs780909637 Chr.8: 43069680-43069680 on Build GRCh38
rs760863217 Chr.8: 43069651-43069651 on Build GRCh38
rs143829409 Chr.8: 43069556-43069556 on Build GRCh38
rs957001904 Chr.8: 43069935-43069935 on Build GRCh38
rs1006197588 Chr.8: 43069821-43069821 on Build GRCh38
rs764718374 Chr.8: 43069689-43069696 on Build GRCh38
rs190618065 Chr.8: 43069798-43069798 on Build GRCh38
rs576085708 Chr.8: 43069794-43069794 on Build GRCh38
rs777233633 Chr.8: 43069701-43069701 on Build GRCh38
rs561826929 Chr.8: 43069714-43069714 on Build GRCh38
rs550839435 Chr.8: 43069757-43069757 on Build GRCh38
rs189240086 Chr.8: 43069926-43069926 on Build GRCh38
rs769234919 Chr.8: 43069588-43069588 on Build GRCh38
rs76046042 Chr.8: 43069576-43069576 on Build GRCh38
rs747697107 Chr.8: 43069587-43069587 on Build GRCh38
rs774766251 Chr.8: 43069615-43069615 on Build GRCh38
rs143260490 Chr.8: 43069674-43069674 on Build GRCh38
rs899522835 Chr.8: 43069812-43069812 on Build GRCh38
rs1014121234 Chr.8: 43069859-43069859 on Build GRCh38
rs147010452 Chr.8: 43069654-43069654 on Build GRCh38
rs1032606366 Chr.8: 43069928-43069928 on Build GRCh38
rs901715354 Chr.8: 43069787-43069787 on Build GRCh38
rs955502009 Chr.8: 43069585-43069585 on Build GRCh38
rs982963861 Chr.8: 43069630-43069630 on Build GRCh38
rs943142783 Chr.8: 43069763-43069763 on Build GRCh38
rs891392654 Chr.8: 43069851-43069851 on Build GRCh38

View Full Product Details