Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 48716317-48716725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB1
Entrez Gene ID
79645
Gene Name
EF-hand calcium binding domain 1
Gene Aliases
-
UniGene
Hs.23245
Gene Chromosome Location
Chr.8: 48714915-48735843 on Build GRCh38
Transcripts
NM_001142857.1 XM_011517589.1

SNP Detail

SNP ID Chromosome Location
rs888907116 Chr.8: 48716430-48716430 on Build GRCh38
rs985971870 Chr.8: 48716539-48716539 on Build GRCh38
rs180782636 Chr.8: 48716336-48716336 on Build GRCh38
rs985644192 Chr.8: 48716346-48716346 on Build GRCh38
rs374136106 Chr.8: 48716454-48716454 on Build GRCh38
rs867663819 Chr.8: 48716501-48716501 on Build GRCh38
rs1045306120 Chr.8: 48716701-48716701 on Build GRCh38
rs79926266 Chr.8: 48716466-48716466 on Build GRCh38
rs1001729936 Chr.8: 48716422-48716422 on Build GRCh38
rs187275258 Chr.8: 48716367-48716367 on Build GRCh38
rs1013845595 Chr.8: 48716646-48716646 on Build GRCh38
rs888176099 Chr.8: 48716416-48716416 on Build GRCh38
rs965906843 Chr.8: 48716484-48716484 on Build GRCh38
rs935541750 Chr.8: 48716585-48716585 on Build GRCh38
rs911415749 Chr.8: 48716347-48716347 on Build GRCh38
rs531394796 Chr.8: 48716467-48716467 on Build GRCh38
rs958143117 Chr.8: 48716472-48716472 on Build GRCh38
rs911314830 Chr.8: 48716478-48716478 on Build GRCh38
rs954627648 Chr.8: 48716535-48716535 on Build GRCh38
rs756122785 Chr.8: 48716445-48716445 on Build GRCh38
rs1014089918 Chr.8: 48716465-48716465 on Build GRCh38
rs1041597854 Chr.8: 48716681-48716681 on Build GRCh38
rs117894061 Chr.8: 48716502-48716502 on Build GRCh38
rs976899200 Chr.8: 48716485-48716485 on Build GRCh38
rs1014491842 Chr.8: 48716624-48716624 on Build GRCh38
rs551798462 Chr.8: 48716468-48716468 on Build GRCh38
rs972017308 Chr.8: 48716386-48716386 on Build GRCh38
rs868337633 Chr.8: 48716606-48716606 on Build GRCh38
rs777469332 Chr.8: 48716570-48716570 on Build GRCh38
rs1055623695 Chr.8: 48716436-48716436 on Build GRCh38
rs896779674 Chr.8: 48716590-48716590 on Build GRCh38
rs377123760 Chr.8: 48716476-48716476 on Build GRCh38
rs373022717 Chr.8: 48716397-48716397 on Build GRCh38
rs541697954 Chr.8: 48716360-48716360 on Build GRCh38
rs183560029 Chr.8: 48716579-48716579 on Build GRCh38
rs192232188 Chr.8: 48716387-48716387 on Build GRCh38
rs531153374 Chr.8: 48716359-48716359 on Build GRCh38
rs1050045217 Chr.8: 48716408-48716408 on Build GRCh38
rs369370979 Chr.8: 48716348-48716348 on Build GRCh38
rs910227779 Chr.8: 48716574-48716574 on Build GRCh38
rs549633289 Chr.8: 48716469-48716469 on Build GRCh38
rs969491392 Chr.8: 48716337-48716337 on Build GRCh38
rs774726831 Chr.8: 48716531-48716531 on Build GRCh38
rs1052597021 Chr.8: 48716589-48716589 on Build GRCh38
rs1055911196 Chr.8: 48716578-48716578 on Build GRCh38
rs752069747 Chr.8: 48716398-48716398 on Build GRCh38
rs909585163 Chr.8: 48716508-48716508 on Build GRCh38
rs148729881 Chr.8: 48716507-48716507 on Build GRCh38

View Full Product Details