Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94739636-94740081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L4
Entrez Gene ID
286148
Gene Name
dpy-19 like 4 (C. elegans)
Gene Aliases
-
UniGene
Hs.567828
Gene Chromosome Location
Chr.8: 94719708-94793848 on Build GRCh38
Transcripts
NM_181787.2 XM_005250894.4 XM_005250895.3 XM_011516983.2 XM_011516984.2 XM_017013351.1 XM_017013352.1 XM_017013353.1 XM_017013354.1 XM_017013355.1 XM_017013356.1 XM_017013357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
244139 c.508G>T p.G170* Chr.8: 94739687-94739687 on Build GRCh38
6326557 c.574G>A p.A192T Chr.8: 94739753-94739753 on Build GRCh38
5063402 c.520A>G p.I174V Chr.8: 94739699-94739699 on Build GRCh38
1643662 c.520_521delAT p.Y175fs*28 Chr.8: 94739699-94739700 on Build GRCh38
6266611 c.468G>T p.E156D Chr.8: 94739647-94739647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954954058 Chr.8: 94739863-94739863 on Build GRCh38
rs200413522 Chr.8: 94739693-94739693 on Build GRCh38
rs879207375 Chr.8: 94739811-94739811 on Build GRCh38
rs371360285 Chr.8: 94739823-94739823 on Build GRCh38
rs566242745 Chr.8: 94739672-94739672 on Build GRCh38
rs761125239 Chr.8: 94739757-94739757 on Build GRCh38
rs772252298 Chr.8: 94739723-94739723 on Build GRCh38
rs777988895 Chr.8: 94739824-94739824 on Build GRCh38
rs920278090 Chr.8: 94739935-94739935 on Build GRCh38
rs867263750 Chr.8: 94740043-94740043 on Build GRCh38
rs765176151 Chr.8: 94739813-94739823 on Build GRCh38
rs78446062 Chr.8: 94739921-94739921 on Build GRCh38
rs759395019 Chr.8: 94739803-94739803 on Build GRCh38
rs750494976 Chr.8: 94739776-94739776 on Build GRCh38
rs780993609 Chr.8: 94739791-94739791 on Build GRCh38
rs981504943 Chr.8: 94739716-94739716 on Build GRCh38
rs769217312 Chr.8: 94739743-94739743 on Build GRCh38
rs886926061 Chr.8: 94739908-94739908 on Build GRCh38
rs1014164268 Chr.8: 94739995-94739995 on Build GRCh38
rs759215062 Chr.8: 94739839-94739839 on Build GRCh38
rs572232019 Chr.8: 94739902-94739902 on Build GRCh38
rs138410439 Chr.8: 94739767-94739767 on Build GRCh38
rs756738882 Chr.8: 94739685-94739685 on Build GRCh38
rs768030971 Chr.8: 94739675-94739675 on Build GRCh38
rs1039348523 Chr.8: 94739784-94739784 on Build GRCh38
rs36005749 Chr.8: 94739866-94739867 on Build GRCh38
rs762624070 Chr.8: 94739674-94739674 on Build GRCh38
rs769745502 Chr.8: 94739818-94739818 on Build GRCh38
rs879719615 Chr.8: 94739986-94739986 on Build GRCh38
rs201559403 Chr.8: 94739865-94739865 on Build GRCh38
rs778419277 Chr.8: 94739691-94739691 on Build GRCh38
rs746798710 Chr.8: 94739735-94739735 on Build GRCh38
rs748798907 Chr.8: 94739827-94739827 on Build GRCh38
rs75768977 Chr.8: 94739706-94739706 on Build GRCh38
rs779873478 Chr.8: 94739732-94739732 on Build GRCh38
rs1012793492 Chr.8: 94740046-94740046 on Build GRCh38
rs200013383 Chr.8: 94739665-94739665 on Build GRCh38
rs778999300 Chr.8: 94739698-94739698 on Build GRCh38
rs975029344 Chr.8: 94739831-94739831 on Build GRCh38
rs908037242 Chr.8: 94739895-94739895 on Build GRCh38
rs899509974 Chr.8: 94739787-94739787 on Build GRCh38
rs150513785 Chr.8: 94739832-94739832 on Build GRCh38
rs776441000 Chr.8: 94739739-94739739 on Build GRCh38
rs770643202 Chr.8: 94739699-94739700 on Build GRCh38
rs898378462 Chr.8: 94739994-94739994 on Build GRCh38
rs371835229 Chr.8: 94739670-94739670 on Build GRCh38
rs1017261506 Chr.8: 94739973-94739973 on Build GRCh38
rs1055456996 Chr.8: 94739975-94739975 on Build GRCh38
rs772745322 Chr.8: 94739745-94739745 on Build GRCh38
rs757295557 Chr.8: 94739696-94739696 on Build GRCh38
rs754321957 Chr.8: 94739772-94739772 on Build GRCh38
rs747170559 Chr.8: 94739789-94739789 on Build GRCh38
rs764396111 Chr.8: 94739841-94739841 on Build GRCh38
rs940464374 Chr.8: 94739900-94739900 on Build GRCh38
rs914864475 Chr.8: 94739779-94739779 on Build GRCh38
rs397702994 Chr.8: 94739870-94739871 on Build GRCh38
rs762559485 Chr.8: 94739746-94739746 on Build GRCh38
rs753085758 Chr.8: 94739679-94739679 on Build GRCh38
rs376044474 Chr.8: 94739773-94739773 on Build GRCh38
rs147079289 Chr.8: 94739754-94739754 on Build GRCh38
rs753179723 Chr.8: 94739755-94739755 on Build GRCh38
rs1041604314 Chr.8: 94740009-94740009 on Build GRCh38
rs780254181 Chr.8: 94739783-94739783 on Build GRCh38
rs538238560 Chr.8: 94739869-94739869 on Build GRCh38
rs201963084 Chr.8: 94739780-94739780 on Build GRCh38
rs924751137 Chr.8: 94739752-94739752 on Build GRCh38
rs558857903 Chr.8: 94739873-94739873 on Build GRCh38
rs776496091 Chr.8: 94739780-94739781 on Build GRCh38
rs367827075 Chr.8: 94739794-94739794 on Build GRCh38
rs183852655 Chr.8: 94739847-94739847 on Build GRCh38
rs147936923 Chr.8: 94739736-94739736 on Build GRCh38
rs936969548 Chr.8: 94739936-94739936 on Build GRCh38
rs775669104 Chr.8: 94739881-94739881 on Build GRCh38

View Full Product Details