Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103453553-103454063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALHM1
Entrez Gene ID
255022
Gene Name
calcium homeostasis modulator 1
Gene Aliases
FAM26C
UniGene
Hs.680365
Gene Chromosome Location
Chr.10: 103453387-103458891 on Build GRCh38
Transcripts
NM_001001412.3

SNP Detail

SNP ID Chromosome Location
rs998557693 Chr.10: 103453626-103453626 on Build GRCh38
rs900838996 Chr.10: 103453970-103453970 on Build GRCh38
rs376657138 Chr.10: 103453687-103453687 on Build GRCh38
rs975755468 Chr.10: 103453690-103453690 on Build GRCh38
rs926636957 Chr.10: 103453801-103453801 on Build GRCh38
rs894051975 Chr.10: 103453837-103453837 on Build GRCh38
rs983947720 Chr.10: 103453704-103453704 on Build GRCh38
rs1019338167 Chr.10: 103453954-103453954 on Build GRCh38
rs996481631 Chr.10: 103453981-103453981 on Build GRCh38
rs1042513899 Chr.10: 103453867-103453867 on Build GRCh38
rs775997782 Chr.10: 103453708-103453708 on Build GRCh38
rs182644857 Chr.10: 103453716-103453716 on Build GRCh38
rs938082056 Chr.10: 103453807-103453807 on Build GRCh38
rs557069103 Chr.10: 103453795-103453795 on Build GRCh38
rs1037756302 Chr.10: 103453882-103453882 on Build GRCh38
rs1019687682 Chr.10: 103453643-103453643 on Build GRCh38
rs139434516 Chr.10: 103454007-103454007 on Build GRCh38
rs538784268 Chr.10: 103453776-103453776 on Build GRCh38
rs946923755 Chr.10: 103453844-103453844 on Build GRCh38
rs11191692 Chr.10: 103454008-103454008 on Build GRCh38
rs746711532 Chr.10: 103453800-103453800 on Build GRCh38
rs952533047 Chr.10: 103453701-103453701 on Build GRCh38
rs927972072 Chr.10: 103453740-103453740 on Build GRCh38
rs534566737 Chr.10: 103453666-103453666 on Build GRCh38
rs999026308 Chr.10: 103453941-103453941 on Build GRCh38
rs553212121 Chr.10: 103453700-103453700 on Build GRCh38
rs1028206621 Chr.10: 103453698-103453698 on Build GRCh38
rs370649673 Chr.10: 103454014-103454014 on Build GRCh38
rs890406955 Chr.10: 103453955-103453955 on Build GRCh38
rs575162697 Chr.10: 103453799-103453799 on Build GRCh38
rs964516333 Chr.10: 103453578-103453578 on Build GRCh38
rs761463129 Chr.10: 103453715-103453715 on Build GRCh38
rs982189756 Chr.10: 103453739-103453739 on Build GRCh38
rs993328131 Chr.10: 103453912-103453912 on Build GRCh38
rs898962529 Chr.10: 103453902-103453902 on Build GRCh38
rs1008960860 Chr.10: 103453960-103453960 on Build GRCh38
rs377018886 Chr.10: 103453792-103453793 on Build GRCh38
rs1021361505 Chr.10: 103453594-103453594 on Build GRCh38
rs902935850 Chr.10: 103453607-103453607 on Build GRCh38
rs949403301 Chr.10: 103453738-103453738 on Build GRCh38
rs999908520 Chr.10: 103454038-103454040 on Build GRCh38
rs542621630 Chr.10: 103453914-103453914 on Build GRCh38

View Full Product Details