Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133362708-133363084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHS1
Entrez Gene ID
1892
Gene Name
enoyl-CoA hydratase, short chain 1
Gene Aliases
ECHS1D, SCEH
UniGene
Hs.76394
Gene Chromosome Location
Chr.10: 133362483-133373404 on Build GRCh38
Transcripts
NM_004092.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
374007 c.814C>G p.R272G Chr.10: 133362927-133362927 on Build GRCh38
6651373 c.808-2A>G p.? Chr.10: 133362935-133362935 on Build GRCh38
4777577 c.812A>G p.D271G Chr.10: 133362929-133362929 on Build GRCh38
1346947 c.815G>A p.R272Q Chr.10: 133362926-133362926 on Build GRCh38
1675685 c.861C>A p.F287L Chr.10: 133362880-133362880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565090080 Chr.10: 133362924-133362924 on Build GRCh38
rs45506491 Chr.10: 133362969-133362969 on Build GRCh38
rs772585791 Chr.10: 133362927-133362927 on Build GRCh38
rs764304433 Chr.10: 133362832-133362832 on Build GRCh38
rs758144044 Chr.10: 133362973-133362973 on Build GRCh38
rs560296011 Chr.10: 133363005-133363005 on Build GRCh38
rs148875965 Chr.10: 133362915-133362915 on Build GRCh38
rs1002785131 Chr.10: 133362997-133362997 on Build GRCh38
rs754957782 Chr.10: 133362871-133362871 on Build GRCh38
rs755635585 Chr.10: 133362855-133362855 on Build GRCh38
rs372224994 Chr.10: 133362968-133362968 on Build GRCh38
rs747933229 Chr.10: 133362886-133362886 on Build GRCh38
rs753653919 Chr.10: 133362869-133362869 on Build GRCh38
rs765943111 Chr.10: 133362863-133362863 on Build GRCh38
rs367754078 Chr.10: 133362967-133362967 on Build GRCh38
rs758234030 Chr.10: 133362887-133362887 on Build GRCh38
rs769725255 Chr.10: 133362910-133362910 on Build GRCh38
rs767247598 Chr.10: 133362840-133362840 on Build GRCh38
rs1039094531 Chr.10: 133362767-133362767 on Build GRCh38
rs34784356 Chr.10: 133363047-133363048 on Build GRCh38
rs372081807 Chr.10: 133362828-133362828 on Build GRCh38
rs923554493 Chr.10: 133362908-133362908 on Build GRCh38
rs145541090 Chr.10: 133362907-133362907 on Build GRCh38
rs983943076 Chr.10: 133362747-133362747 on Build GRCh38
rs373661920 Chr.10: 133362732-133362732 on Build GRCh38
rs576889001 Chr.10: 133362739-133362739 on Build GRCh38
rs183588815 Chr.10: 133362733-133362733 on Build GRCh38
rs769781968 Chr.10: 133362982-133362982 on Build GRCh38
rs975242355 Chr.10: 133363032-133363032 on Build GRCh38
rs1031532710 Chr.10: 133363002-133363002 on Build GRCh38
rs774760601 Chr.10: 133362838-133362838 on Build GRCh38
rs376401874 Chr.10: 133362814-133362814 on Build GRCh38
rs909736957 Chr.10: 133363001-133363001 on Build GRCh38
rs112236366 Chr.10: 133363055-133363055 on Build GRCh38
rs965123282 Chr.10: 133363061-133363061 on Build GRCh38
rs914611110 Chr.10: 133362858-133362858 on Build GRCh38
rs906870754 Chr.10: 133362979-133362979 on Build GRCh38
rs772958061 Chr.10: 133362947-133362947 on Build GRCh38
rs573418441 Chr.10: 133362746-133362746 on Build GRCh38
rs769198495 Chr.10: 133362919-133362919 on Build GRCh38
rs778775545 Chr.10: 133362878-133362878 on Build GRCh38
rs781373718 Chr.10: 133362895-133362895 on Build GRCh38
rs188884786 Chr.10: 133362738-133362738 on Build GRCh38
rs955903642 Chr.10: 133363044-133363044 on Build GRCh38
rs114654147 Chr.10: 133362950-133362950 on Build GRCh38
rs532299045 Chr.10: 133363006-133363006 on Build GRCh38
rs750584290 Chr.10: 133362983-133362983 on Build GRCh38
rs149289784 Chr.10: 133363054-133363054 on Build GRCh38
rs775650144 Chr.10: 133362911-133362911 on Build GRCh38
rs750148216 Chr.10: 133362845-133362845 on Build GRCh38
rs1050040 Chr.10: 133362754-133362754 on Build GRCh38
rs1023348724 Chr.10: 133362952-133362952 on Build GRCh38
rs762331214 Chr.10: 133362839-133362839 on Build GRCh38
rs116786579 Chr.10: 133362759-133362759 on Build GRCh38
rs933592081 Chr.10: 133362909-133362909 on Build GRCh38
rs15116 Chr.10: 133362756-133362756 on Build GRCh38
rs955870758 Chr.10: 133362783-133362783 on Build GRCh38
rs192497815 Chr.10: 133362970-133362970 on Build GRCh38
rs575638199 Chr.10: 133362926-133362926 on Build GRCh38

View Full Product Details