Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6482288-6482867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIMM10B
Entrez Gene ID
26515
Gene Name
translocase of inner mitochondrial membrane 10 homolog B (yeast)
Gene Aliases
FXC1, TIM10B, Tim9b
UniGene
Hs.54943
Gene Chromosome Location
Chr.11: 6481447-6484681 on Build GRCh38
Transcripts
NM_012192.3

SNP Detail

SNP ID Chromosome Location
rs540285423 Chr.11: 6482392-6482392 on Build GRCh38
rs925632426 Chr.11: 6482429-6482429 on Build GRCh38
rs553021064 Chr.11: 6482593-6482593 on Build GRCh38
rs570349858 Chr.11: 6482433-6482433 on Build GRCh38
rs548927460 Chr.11: 6482428-6482428 on Build GRCh38
rs747390668 Chr.11: 6482682-6482682 on Build GRCh38
rs1002050326 Chr.11: 6482457-6482457 on Build GRCh38
rs1048121 Chr.11: 6482345-6482345 on Build GRCh38
rs141524560 Chr.11: 6482562-6482562 on Build GRCh38
rs969889600 Chr.11: 6482388-6482388 on Build GRCh38
rs988533933 Chr.11: 6482650-6482650 on Build GRCh38
rs993948358 Chr.11: 6482604-6482604 on Build GRCh38
rs778357715 Chr.11: 6482663-6482663 on Build GRCh38
rs1052720043 Chr.11: 6482468-6482468 on Build GRCh38
rs961210483 Chr.11: 6482349-6482349 on Build GRCh38
rs986344226 Chr.11: 6482809-6482813 on Build GRCh38
rs757678812 Chr.11: 6482762-6482762 on Build GRCh38
rs969377394 Chr.11: 6482737-6482737 on Build GRCh38
rs950333033 Chr.11: 6482667-6482667 on Build GRCh38
rs541574364 Chr.11: 6482677-6482677 on Build GRCh38
rs1049035497 Chr.11: 6482472-6482472 on Build GRCh38
rs887789109 Chr.11: 6482482-6482482 on Build GRCh38
rs545667905 Chr.11: 6482333-6482333 on Build GRCh38
rs957120072 Chr.11: 6482779-6482779 on Build GRCh38
rs73400858 Chr.11: 6482500-6482500 on Build GRCh38
rs773000430 Chr.11: 6482350-6482350 on Build GRCh38
rs957269865 Chr.11: 6482619-6482619 on Build GRCh38
rs1029476547 Chr.11: 6482823-6482823 on Build GRCh38
rs73400854 Chr.11: 6482358-6482358 on Build GRCh38
rs73400857 Chr.11: 6482448-6482448 on Build GRCh38
rs190745852 Chr.11: 6482423-6482423 on Build GRCh38
rs961321766 Chr.11: 6482517-6482517 on Build GRCh38
rs1005436155 Chr.11: 6482490-6482490 on Build GRCh38
rs905943820 Chr.11: 6482455-6482455 on Build GRCh38
rs1029528751 Chr.11: 6482561-6482561 on Build GRCh38
rs1005287628 Chr.11: 6482773-6482773 on Build GRCh38
rs561685079 Chr.11: 6482402-6482402 on Build GRCh38
rs546034469 Chr.11: 6482821-6482821 on Build GRCh38
rs994531100 Chr.11: 6482317-6482317 on Build GRCh38
rs759003518 Chr.11: 6482613-6482613 on Build GRCh38
rs913367882 Chr.11: 6482445-6482445 on Build GRCh38
rs901036322 Chr.11: 6482596-6482596 on Build GRCh38
rs1036395801 Chr.11: 6482801-6482801 on Build GRCh38
rs115297972 Chr.11: 6482808-6482808 on Build GRCh38
rs897060032 Chr.11: 6482803-6482803 on Build GRCh38
rs573062300 Chr.11: 6482777-6482777 on Build GRCh38
rs916556733 Chr.11: 6482364-6482364 on Build GRCh38
rs985631642 Chr.11: 6482588-6482588 on Build GRCh38
rs572961748 Chr.11: 6482838-6482838 on Build GRCh38
rs754086371 Chr.11: 6482443-6482443 on Build GRCh38
rs998009142 Chr.11: 6482519-6482519 on Build GRCh38
rs937414600 Chr.11: 6482730-6482730 on Build GRCh38
rs900903783 Chr.11: 6482836-6482836 on Build GRCh38
rs927486132 Chr.11: 6482689-6482689 on Build GRCh38
rs184241439 Chr.11: 6482591-6482591 on Build GRCh38
rs1045911690 Chr.11: 6482451-6482451 on Build GRCh38
rs1040798977 Chr.11: 6482594-6482594 on Build GRCh38
rs535439357 Chr.11: 6482741-6482741 on Build GRCh38
rs953808121 Chr.11: 6482427-6482427 on Build GRCh38
rs942373945 Chr.11: 6482824-6482824 on Build GRCh38
rs187683891 Chr.11: 6482755-6482755 on Build GRCh38
rs1025541241 Chr.11: 6482624-6482624 on Build GRCh38
rs752669277 Chr.11: 6482535-6482535 on Build GRCh38
rs9802 Chr.11: 6482831-6482831 on Build GRCh38
rs915385301 Chr.11: 6482807-6482808 on Build GRCh38
rs568142296 Chr.11: 6482608-6482608 on Build GRCh38

View Full Product Details