Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27349076-27349622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC34
Entrez Gene ID
91057
Gene Name
coiled-coil domain containing 34
Gene Aliases
L15, NY-REN-41, RAMA3
UniGene
Hs.143733
Gene Chromosome Location
Chr.11: 27338514-27363248 on Build GRCh38
Transcripts
NM_030771.1 NM_080654.2

SNP Detail

SNP ID Chromosome Location
rs201879455 Chr.11: 27349414-27349414 on Build GRCh38
rs894135210 Chr.11: 27349134-27349134 on Build GRCh38
rs1055438330 Chr.11: 27349115-27349115 on Build GRCh38
rs570776566 Chr.11: 27349121-27349121 on Build GRCh38
rs951296783 Chr.11: 27349349-27349349 on Build GRCh38
rs982614645 Chr.11: 27349355-27349355 on Build GRCh38
rs999183374 Chr.11: 27349329-27349329 on Build GRCh38
rs552929042 Chr.11: 27349233-27349233 on Build GRCh38
rs16916893 Chr.11: 27349466-27349466 on Build GRCh38
rs552517414 Chr.11: 27349103-27349103 on Build GRCh38
rs1012796502 Chr.11: 27349276-27349276 on Build GRCh38
rs948333444 Chr.11: 27349167-27349167 on Build GRCh38
rs986963163 Chr.11: 27349423-27349423 on Build GRCh38
rs1018315344 Chr.11: 27349325-27349325 on Build GRCh38
rs557388807 Chr.11: 27349410-27349410 on Build GRCh38
rs16916892 Chr.11: 27349218-27349218 on Build GRCh38
rs575655786 Chr.11: 27349463-27349463 on Build GRCh38
rs920176926 Chr.11: 27349471-27349471 on Build GRCh38
rs897487010 Chr.11: 27349216-27349216 on Build GRCh38
rs964020972 Chr.11: 27349326-27349326 on Build GRCh38
rs904064695 Chr.11: 27349212-27349212 on Build GRCh38
rs954125316 Chr.11: 27349421-27349421 on Build GRCh38
rs535570906 Chr.11: 27349306-27349306 on Build GRCh38
rs990094529 Chr.11: 27349416-27349416 on Build GRCh38
rs930329447 Chr.11: 27349570-27349570 on Build GRCh38
rs745880750 Chr.11: 27349467-27349467 on Build GRCh38
rs191796156 Chr.11: 27349303-27349303 on Build GRCh38
rs991631851 Chr.11: 27349436-27349436 on Build GRCh38
rs778804590 Chr.11: 27349362-27349362 on Build GRCh38
rs894212035 Chr.11: 27349273-27349273 on Build GRCh38
rs35700702 Chr.11: 27349520-27349521 on Build GRCh38
rs1043953649 Chr.11: 27349178-27349178 on Build GRCh38
rs928388121 Chr.11: 27349364-27349364 on Build GRCh38
rs773234941 Chr.11: 27349533-27349533 on Build GRCh38
rs369199448 Chr.11: 27349535-27349535 on Build GRCh38
rs912737820 Chr.11: 27349428-27349428 on Build GRCh38
rs1028387018 Chr.11: 27349419-27349419 on Build GRCh38
rs1037250801 Chr.11: 27349166-27349166 on Build GRCh38
rs957612452 Chr.11: 27349404-27349404 on Build GRCh38
rs771051525 Chr.11: 27349331-27349331 on Build GRCh38
rs772100627 Chr.11: 27349541-27349541 on Build GRCh38
rs907004065 Chr.11: 27349313-27349313 on Build GRCh38
rs940163994 Chr.11: 27349445-27349445 on Build GRCh38

View Full Product Details