Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65868119-65868713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFEMP2
Entrez Gene ID
30008
Gene Name
EGF containing fibulin like extracellular matrix protein 2
Gene Aliases
ARCL1B, FBLN4, MBP1, UPH1
UniGene
Hs.731454
Gene Chromosome Location
Chr.11: 65866441-65872934 on Build GRCh38
Transcripts
NM_016938.4 NR_037718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4035562 c.932A>G p.D311G Chr.11: 65868337-65868337 on Build GRCh38
6336840 c.945C>T p.C315C Chr.11: 65868324-65868324 on Build GRCh38
467249 c.946G>A p.V316M Chr.11: 65868323-65868323 on Build GRCh38
3980569 c.912T>C p.H304H Chr.11: 65868357-65868357 on Build GRCh38
3451909 c.891C>T p.A297A Chr.11: 65868378-65868378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559896273 Chr.11: 65868489-65868489 on Build GRCh38
rs1022753423 Chr.11: 65868470-65868470 on Build GRCh38
rs1036879024 Chr.11: 65868144-65868144 on Build GRCh38
rs761154932 Chr.11: 65868657-65868657 on Build GRCh38
rs193302866 Chr.11: 65868557-65868557 on Build GRCh38
rs777080474 Chr.11: 65868300-65868300 on Build GRCh38
rs780653985 Chr.11: 65868563-65868563 on Build GRCh38
rs756121658 Chr.11: 65868258-65868258 on Build GRCh38
rs200340088 Chr.11: 65868576-65868576 on Build GRCh38
rs373937692 Chr.11: 65868451-65868451 on Build GRCh38
rs769829583 Chr.11: 65868364-65868364 on Build GRCh38
rs564554534 Chr.11: 65868351-65868351 on Build GRCh38
rs780379870 Chr.11: 65868455-65868455 on Build GRCh38
rs200768579 Chr.11: 65868255-65868255 on Build GRCh38
rs752273286 Chr.11: 65868407-65868407 on Build GRCh38
rs767404922 Chr.11: 65868253-65868253 on Build GRCh38
rs369517593 Chr.11: 65868651-65868651 on Build GRCh38
rs756396652 Chr.11: 65868561-65868561 on Build GRCh38
rs756905509 Chr.11: 65868521-65868521 on Build GRCh38
rs972435635 Chr.11: 65868588-65868588 on Build GRCh38
rs746147986 Chr.11: 65868453-65868453 on Build GRCh38
rs771450901 Chr.11: 65868296-65868296 on Build GRCh38
rs573550578 Chr.11: 65868212-65868212 on Build GRCh38
rs532989312 Chr.11: 65868395-65868395 on Build GRCh38
rs766452746 Chr.11: 65868329-65868329 on Build GRCh38
rs375869389 Chr.11: 65868467-65868467 on Build GRCh38
rs371085261 Chr.11: 65868288-65868288 on Build GRCh38
rs142551972 Chr.11: 65868193-65868193 on Build GRCh38
rs759635079 Chr.11: 65868175-65868177 on Build GRCh38
rs760560174 Chr.11: 65868448-65868448 on Build GRCh38
rs113167523 Chr.11: 65868323-65868323 on Build GRCh38
rs901998017 Chr.11: 65868151-65868151 on Build GRCh38
rs544423133 Chr.11: 65868341-65868341 on Build GRCh38
rs750260446 Chr.11: 65868324-65868324 on Build GRCh38
rs967073714 Chr.11: 65868229-65868229 on Build GRCh38
rs119489102 Chr.11: 65868522-65868522 on Build GRCh38
rs553406481 Chr.11: 65868199-65868199 on Build GRCh38
rs1003856168 Chr.11: 65868297-65868297 on Build GRCh38
rs373825149 Chr.11: 65868656-65868656 on Build GRCh38
rs766642842 Chr.11: 65868677-65868677 on Build GRCh38
rs765815867 Chr.11: 65868449-65868451 on Build GRCh38
rs780085115 Chr.11: 65868260-65868260 on Build GRCh38
rs773386551 Chr.11: 65868437-65868437 on Build GRCh38
rs140784880 Chr.11: 65868396-65868396 on Build GRCh38
rs931224413 Chr.11: 65868267-65868267 on Build GRCh38
rs142509316 Chr.11: 65868384-65868384 on Build GRCh38
rs772378278 Chr.11: 65868283-65868283 on Build GRCh38
rs765407883 Chr.11: 65868246-65868246 on Build GRCh38
rs760501413 Chr.11: 65868328-65868328 on Build GRCh38
rs749794461 Chr.11: 65868566-65868566 on Build GRCh38
rs762497696 Chr.11: 65868497-65868497 on Build GRCh38
rs759195625 Chr.11: 65868490-65868490 on Build GRCh38
rs751277215 Chr.11: 65868505-65868505 on Build GRCh38
rs550977800 Chr.11: 65868346-65868346 on Build GRCh38
rs371712739 Chr.11: 65868387-65868387 on Build GRCh38
rs200285623 Chr.11: 65868417-65868417 on Build GRCh38
rs775639842 Chr.11: 65868374-65868374 on Build GRCh38
rs754474632 Chr.11: 65868272-65868272 on Build GRCh38
rs758052109 Chr.11: 65868338-65868338 on Build GRCh38
rs763774695 Chr.11: 65868433-65868433 on Build GRCh38
rs2234469 Chr.11: 65868313-65868313 on Build GRCh38
rs570960149 Chr.11: 65868161-65868161 on Build GRCh38
rs1044449024 Chr.11: 65868408-65868408 on Build GRCh38
rs770919025 Chr.11: 65868486-65868486 on Build GRCh38
rs371680911 Chr.11: 65868461-65868461 on Build GRCh38
rs755167697 Chr.11: 65868568-65868568 on Build GRCh38
rs763580525 Chr.11: 65868498-65868498 on Build GRCh38
rs748369626 Chr.11: 65868479-65868479 on Build GRCh38
rs202220871 Chr.11: 65868235-65868235 on Build GRCh38
rs775555812 Chr.11: 65868600-65868600 on Build GRCh38
rs1053067210 Chr.11: 65868156-65868156 on Build GRCh38
rs118107504 Chr.11: 65868248-65868248 on Build GRCh38
rs202124723 Chr.11: 65868587-65868587 on Build GRCh38
rs533449135 Chr.11: 65868457-65868457 on Build GRCh38
rs373585608 Chr.11: 65868257-65868257 on Build GRCh38
rs753199874 Chr.11: 65868620-65868622 on Build GRCh38
rs926184701 Chr.11: 65868350-65868350 on Build GRCh38
rs779131358 Chr.11: 65868574-65868574 on Build GRCh38
rs767233775 Chr.11: 65868654-65868654 on Build GRCh38
rs368296311 Chr.11: 65868358-65868358 on Build GRCh38
rs762880509 Chr.11: 65868383-65868383 on Build GRCh38
rs781700061 Chr.11: 65868347-65868347 on Build GRCh38
rs144780990 Chr.11: 65868619-65868619 on Build GRCh38
rs1050571636 Chr.11: 65868639-65868639 on Build GRCh38
rs747815940 Chr.11: 65868273-65868273 on Build GRCh38
rs148410446 Chr.11: 65868335-65868335 on Build GRCh38
rs1011843137 Chr.11: 65868198-65868198 on Build GRCh38
rs765251023 Chr.11: 65868496-65868496 on Build GRCh38
rs757673906 Chr.11: 65868449-65868449 on Build GRCh38
rs144256865 Chr.11: 65868583-65868583 on Build GRCh38
rs113520206 Chr.11: 65868421-65868421 on Build GRCh38
rs1029181102 Chr.11: 65868184-65868184 on Build GRCh38
rs766494495 Chr.11: 65868342-65868342 on Build GRCh38
rs763948572 Chr.11: 65868646-65868646 on Build GRCh38
rs769892560 Chr.11: 65868314-65868314 on Build GRCh38
rs776698728 Chr.11: 65868432-65868432 on Build GRCh38
rs895871146 Chr.11: 65868183-65868183 on Build GRCh38
rs757048147 Chr.11: 65868250-65868250 on Build GRCh38
rs761260850 Chr.11: 65868653-65868653 on Build GRCh38
rs377139656 Chr.11: 65868632-65868632 on Build GRCh38
rs977178424 Chr.11: 65868236-65868236 on Build GRCh38
rs750819363 Chr.11: 65868559-65868559 on Build GRCh38
rs372722886 Chr.11: 65868605-65868605 on Build GRCh38
rs150546623 Chr.11: 65868547-65868547 on Build GRCh38
rs542517910 Chr.11: 65868244-65868244 on Build GRCh38
rs778475610 Chr.11: 65868332-65868332 on Build GRCh38
rs953815379 Chr.11: 65868501-65868501 on Build GRCh38
rs754497556 Chr.11: 65868330-65868330 on Build GRCh38
rs759573856 Chr.11: 65868305-65868305 on Build GRCh38
rs772353731 Chr.11: 65868577-65868577 on Build GRCh38
rs774765721 Chr.11: 65868478-65868478 on Build GRCh38
rs601314 Chr.11: 65868582-65868582 on Build GRCh38
rs950192179 Chr.11: 65868190-65868190 on Build GRCh38
rs1011833179 Chr.11: 65868390-65868390 on Build GRCh38
rs367770866 Chr.11: 65868454-65868454 on Build GRCh38
rs753424389 Chr.11: 65868265-65868265 on Build GRCh38
rs759514025 Chr.11: 65868405-65868405 on Build GRCh38
rs868407759 Chr.11: 65868569-65868569 on Build GRCh38
rs753778921 Chr.11: 65868397-65868397 on Build GRCh38

View Full Product Details