Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 110435446-110435977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDX1
Entrez Gene ID
2230
Gene Name
ferredoxin 1
Gene Aliases
ADX, FDX, LOH11CR1D
UniGene
Hs.744
Gene Chromosome Location
Chr.11: 110429937-110464884 on Build GRCh38
Transcripts
NM_004109.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5749493 c.280G>T p.V94F Chr.11: 110435928-110435928 on Build GRCh38
1206963 c.283G>T p.E95* Chr.11: 110435931-110435931 on Build GRCh38
7178550 c.188C>T p.S63L Chr.11: 110435836-110435836 on Build GRCh38
4988586 c.239C>A p.T80K Chr.11: 110435887-110435887 on Build GRCh38
3738857 c.287A>G p.N96S Chr.11: 110435935-110435935 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1017386438 Chr.11: 110435816-110435817 on Build GRCh38
rs1009511955 Chr.11: 110435664-110435664 on Build GRCh38
rs779949588 Chr.11: 110435762-110435762 on Build GRCh38
rs535925663 Chr.11: 110435565-110435565 on Build GRCh38
rs773338470 Chr.11: 110435829-110435829 on Build GRCh38
rs923287469 Chr.11: 110435653-110435653 on Build GRCh38
rs139446247 Chr.11: 110435902-110435902 on Build GRCh38
rs780150621 Chr.11: 110435794-110435794 on Build GRCh38
rs906075720 Chr.11: 110435710-110435710 on Build GRCh38
rs909264831 Chr.11: 110435858-110435858 on Build GRCh38
rs757253507 Chr.11: 110435820-110435821 on Build GRCh38
rs952385264 Chr.11: 110435699-110435699 on Build GRCh38
rs768532188 Chr.11: 110435847-110435847 on Build GRCh38
rs1034718982 Chr.11: 110435716-110435716 on Build GRCh38
rs144228166 Chr.11: 110435951-110435951 on Build GRCh38
rs764822594 Chr.11: 110435872-110435872 on Build GRCh38
rs761182980 Chr.11: 110435577-110435577 on Build GRCh38
rs896134146 Chr.11: 110435718-110435718 on Build GRCh38
rs927098262 Chr.11: 110435550-110435550 on Build GRCh38
rs777923921 Chr.11: 110435703-110435703 on Build GRCh38
rs755903344 Chr.11: 110435759-110435759 on Build GRCh38
rs754755113 Chr.11: 110435817-110435817 on Build GRCh38
rs1026664129 Chr.11: 110435696-110435696 on Build GRCh38
rs572827954 Chr.11: 110435697-110435698 on Build GRCh38
rs1026693472 Chr.11: 110435511-110435511 on Build GRCh38
rs778972195 Chr.11: 110435621-110435621 on Build GRCh38
rs978706269 Chr.11: 110435810-110435810 on Build GRCh38
rs781457182 Chr.11: 110435819-110435819 on Build GRCh38
rs759894054 Chr.11: 110435855-110435855 on Build GRCh38
rs10891105 Chr.11: 110435605-110435605 on Build GRCh38
rs753029771 Chr.11: 110435671-110435671 on Build GRCh38
rs769913106 Chr.11: 110435825-110435825 on Build GRCh38
rs765304776 Chr.11: 110435821-110435821 on Build GRCh38
rs372905368 Chr.11: 110435731-110435731 on Build GRCh38
rs375254527 Chr.11: 110435887-110435887 on Build GRCh38
rs568402619 Chr.11: 110435544-110435544 on Build GRCh38
rs185852628 Chr.11: 110435867-110435867 on Build GRCh38
rs540180016 Chr.11: 110435709-110435709 on Build GRCh38
rs182106488 Chr.11: 110435684-110435684 on Build GRCh38
rs374154404 Chr.11: 110435669-110435669 on Build GRCh38
rs774868154 Chr.11: 110435851-110435851 on Build GRCh38
rs756272504 Chr.11: 110435944-110435944 on Build GRCh38
rs375998812 Chr.11: 110435922-110435922 on Build GRCh38
rs186325541 Chr.11: 110435572-110435572 on Build GRCh38
rs979902406 Chr.11: 110435534-110435534 on Build GRCh38
rs769887967 Chr.11: 110435747-110435747 on Build GRCh38
rs150960595 Chr.11: 110435891-110435891 on Build GRCh38
rs1034141652 Chr.11: 110435711-110435711 on Build GRCh38
rs992757205 Chr.11: 110435570-110435570 on Build GRCh38
rs780919632 Chr.11: 110435919-110435919 on Build GRCh38
rs749385109 Chr.11: 110435833-110435833 on Build GRCh38
rs189887469 Chr.11: 110435663-110435663 on Build GRCh38
rs925944834 Chr.11: 110435820-110435820 on Build GRCh38
rs58842739 Chr.11: 110435799-110435799 on Build GRCh38
rs773868601 Chr.11: 110435924-110435924 on Build GRCh38
rs929411985 Chr.11: 110435657-110435657 on Build GRCh38
rs199894632 Chr.11: 110435868-110435868 on Build GRCh38
rs748363171 Chr.11: 110435823-110435823 on Build GRCh38
rs952458738 Chr.11: 110435526-110435526 on Build GRCh38
rs142469641 Chr.11: 110435715-110435715 on Build GRCh38
rs761435469 Chr.11: 110435869-110435869 on Build GRCh38
rs755143922 Chr.11: 110435611-110435611 on Build GRCh38
rs1021229696 Chr.11: 110435489-110435489 on Build GRCh38
rs79237613 Chr.11: 110435602-110435602 on Build GRCh38
rs972816579 Chr.11: 110435504-110435504 on Build GRCh38
rs749892432 Chr.11: 110435879-110435879 on Build GRCh38
rs751058251 Chr.11: 110435569-110435569 on Build GRCh38
rs1020673718 Chr.11: 110435732-110435732 on Build GRCh38
rs898071813 Chr.11: 110435677-110435677 on Build GRCh38
rs946035707 Chr.11: 110435582-110435582 on Build GRCh38

View Full Product Details