Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134378842-134379406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GAT1
Entrez Gene ID
27087
Gene Name
beta-1,3-glucuronyltransferase 1
Gene Aliases
CD57, GLCATP, GLCUATP, HNK1, LEU7, NK-1, NK1
UniGene
Hs.381050
Gene Chromosome Location
Chr.11: 134378504-134411986 on Build GRCh38
Transcripts
NM_018644.3 NM_054025.2 XM_005271506.3 XM_011542751.2 XM_017017551.1 XM_017017552.1

SNP Detail

SNP ID Chromosome Location
rs1024287511 Chr.11: 134379041-134379041 on Build GRCh38
rs373101961 Chr.11: 134378880-134378880 on Build GRCh38
rs1045331503 Chr.11: 134379336-134379336 on Build GRCh38
rs1006149968 Chr.11: 134378946-134378946 on Build GRCh38
rs986385145 Chr.11: 134379093-134379093 on Build GRCh38
rs956362177 Chr.11: 134379089-134379089 on Build GRCh38
rs888675158 Chr.11: 134378882-134378882 on Build GRCh38
rs908966551 Chr.11: 134379233-134379233 on Build GRCh38
rs867684327 Chr.11: 134379370-134379370 on Build GRCh38
rs918963407 Chr.11: 134379121-134379121 on Build GRCh38
rs1033069517 Chr.11: 134379372-134379372 on Build GRCh38
rs372052866 Chr.11: 134379271-134379271 on Build GRCh38
rs998935221 Chr.11: 134379054-134379054 on Build GRCh38
rs554890324 Chr.11: 134379124-134379124 on Build GRCh38
rs551297234 Chr.11: 134379341-134379341 on Build GRCh38
rs1047033614 Chr.11: 134379292-134379292 on Build GRCh38
rs1056389573 Chr.11: 134379263-134379263 on Build GRCh38
rs186085326 Chr.11: 134379284-134379284 on Build GRCh38
rs1056019385 Chr.11: 134379005-134379005 on Build GRCh38
rs967968805 Chr.11: 134379118-134379118 on Build GRCh38
rs957194226 Chr.11: 134379076-134379076 on Build GRCh38
rs1047766806 Chr.11: 134378863-134378863 on Build GRCh38
rs561766391 Chr.11: 134379123-134379123 on Build GRCh38
rs34747914 Chr.11: 134378906-134378906 on Build GRCh38
rs967426306 Chr.11: 134379047-134379047 on Build GRCh38
rs1003673120 Chr.11: 134379376-134379376 on Build GRCh38
rs1014493713 Chr.11: 134379329-134379329 on Build GRCh38
rs910448885 Chr.11: 134379096-134379096 on Build GRCh38
rs998820585 Chr.11: 134379364-134379366 on Build GRCh38
rs1045706459 Chr.11: 134379331-134379331 on Build GRCh38
rs929976249 Chr.11: 134379291-134379291 on Build GRCh38
rs557767311 Chr.11: 134378997-134378997 on Build GRCh38
rs115630273 Chr.11: 134379161-134379161 on Build GRCh38
rs1033515494 Chr.11: 134379062-134379062 on Build GRCh38
rs888425973 Chr.11: 134379327-134379327 on Build GRCh38
rs879592181 Chr.11: 134379029-134379029 on Build GRCh38
rs759782782 Chr.11: 134379378-134379378 on Build GRCh38
rs905399958 Chr.11: 134379343-134379343 on Build GRCh38
rs915157546 Chr.11: 134379101-134379101 on Build GRCh38
rs117400758 Chr.11: 134379100-134379100 on Build GRCh38
rs1023826535 Chr.11: 134378976-134378976 on Build GRCh38
rs182419474 Chr.11: 134379039-134379039 on Build GRCh38
rs910414198 Chr.11: 134379129-134379129 on Build GRCh38
rs868202944 Chr.11: 134379257-134379257 on Build GRCh38
rs1034764811 Chr.11: 134379082-134379082 on Build GRCh38
rs976846985 Chr.11: 134379185-134379185 on Build GRCh38
rs112591124 Chr.11: 134379332-134379332 on Build GRCh38

View Full Product Details