Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48957020-48957591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF3
Entrez Gene ID
377
Gene Name
ADP ribosylation factor 3
Gene Aliases
-
UniGene
Hs.119177
Gene Chromosome Location
Chr.12: 48928836-48957526 on Build GRCh38
Transcripts
NM_001659.2 XM_005268856.1 XM_006719391.3

SNP Detail

SNP ID Chromosome Location
rs775758276 Chr.12: 48957405-48957405 on Build GRCh38
rs1043083525 Chr.12: 48957426-48957426 on Build GRCh38
rs948316849 Chr.12: 48957300-48957300 on Build GRCh38
rs962827980 Chr.12: 48957185-48957185 on Build GRCh38
rs863855 Chr.12: 48957540-48957540 on Build GRCh38
rs899058533 Chr.12: 48957524-48957524 on Build GRCh38
rs890405129 Chr.12: 48957294-48957294 on Build GRCh38
rs1014969385 Chr.12: 48957347-48957347 on Build GRCh38
rs891515822 Chr.12: 48957467-48957467 on Build GRCh38
rs960803489 Chr.12: 48957543-48957543 on Build GRCh38
rs1007166755 Chr.12: 48957142-48957142 on Build GRCh38
rs982325669 Chr.12: 48957369-48957369 on Build GRCh38
rs554461727 Chr.12: 48957290-48957290 on Build GRCh38
rs544237256 Chr.12: 48957441-48957441 on Build GRCh38
rs996035619 Chr.12: 48957249-48957249 on Build GRCh38
rs948248947 Chr.12: 48957069-48957069 on Build GRCh38
rs972815508 Chr.12: 48957220-48957220 on Build GRCh38
rs962545021 Chr.12: 48957528-48957528 on Build GRCh38
rs76101427 Chr.12: 48957392-48957392 on Build GRCh38
rs890280373 Chr.12: 48957484-48957484 on Build GRCh38
rs576883121 Chr.12: 48957483-48957483 on Build GRCh38
rs79092274 Chr.12: 48957286-48957286 on Build GRCh38
rs973363972 Chr.12: 48957355-48957355 on Build GRCh38
rs891453245 Chr.12: 48957145-48957145 on Build GRCh38
rs1014853384 Chr.12: 48957499-48957500 on Build GRCh38
rs998285040 Chr.12: 48957470-48957470 on Build GRCh38
rs758793371 Chr.12: 48957063-48957063 on Build GRCh38
rs935459714 Chr.12: 48957408-48957408 on Build GRCh38
rs573690121 Chr.12: 48957305-48957305 on Build GRCh38
rs947018561 Chr.12: 48957196-48957196 on Build GRCh38
rs935563776 Chr.12: 48957123-48957123 on Build GRCh38
rs889737414 Chr.12: 48957497-48957497 on Build GRCh38
rs953970753 Chr.12: 48957137-48957137 on Build GRCh38
rs928266300 Chr.12: 48957107-48957107 on Build GRCh38
rs992148115 Chr.12: 48957235-48957235 on Build GRCh38
rs1042789949 Chr.12: 48957215-48957215 on Build GRCh38
rs569040286 Chr.12: 48957150-48957150 on Build GRCh38
rs576641422 Chr.12: 48957558-48957558 on Build GRCh38
rs1051644588 Chr.12: 48957442-48957442 on Build GRCh38
rs1004801504 Chr.12: 48957296-48957296 on Build GRCh38
rs533829202 Chr.12: 48957406-48957406 on Build GRCh38
rs1044317802 Chr.12: 48957349-48957349 on Build GRCh38
rs1050772444 Chr.12: 48957474-48957474 on Build GRCh38
rs918642525 Chr.12: 48957223-48957223 on Build GRCh38
rs562621137 Chr.12: 48957498-48957498 on Build GRCh38
rs532294047 Chr.12: 48957095-48957095 on Build GRCh38
rs1016821688 Chr.12: 48957509-48957509 on Build GRCh38
rs376141389 Chr.12: 48957471-48957471 on Build GRCh38
rs904310419 Chr.12: 48957072-48957072 on Build GRCh38
rs1052756554 Chr.12: 48957110-48957110 on Build GRCh38
rs552413852 Chr.12: 48957135-48957135 on Build GRCh38
rs900277161 Chr.12: 48957431-48957431 on Build GRCh38
rs1023739589 Chr.12: 48957365-48957365 on Build GRCh38
rs947071159 Chr.12: 48957421-48957421 on Build GRCh38
rs993973205 Chr.12: 48957529-48957529 on Build GRCh38
rs546537739 Chr.12: 48957502-48957502 on Build GRCh38
rs969850134 Chr.12: 48957365-48957365 on Build GRCh38
rs916619987 Chr.12: 48957048-48957048 on Build GRCh38
rs751735633 Chr.12: 48957301-48957302 on Build GRCh38
rs992595667 Chr.12: 48957553-48957553 on Build GRCh38
rs1016761535 Chr.12: 48957179-48957179 on Build GRCh38
rs997826241 Chr.12: 48957132-48957132 on Build GRCh38
rs960711115 Chr.12: 48957232-48957232 on Build GRCh38
rs891463436 Chr.12: 48957122-48957122 on Build GRCh38

View Full Product Details