Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 63146645-63147143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR1A
Entrez Gene ID
552
Gene Name
arginine vasopressin receptor 1A
Gene Aliases
AVPR V1a, AVPR1, V1aR
UniGene
Hs.2131
Gene Chromosome Location
Chr.12: 63142759-63152810 on Build GRCh38
Transcripts
NM_000706.4

SNP Detail

SNP ID Chromosome Location
rs547981122 Chr.12: 63146786-63146786 on Build GRCh38
rs959982723 Chr.12: 63146691-63146691 on Build GRCh38
rs972344397 Chr.12: 63146847-63146847 on Build GRCh38
rs570678571 Chr.12: 63146897-63146897 on Build GRCh38
rs924676448 Chr.12: 63147041-63147041 on Build GRCh38
rs528214854 Chr.12: 63146735-63146735 on Build GRCh38
rs986850990 Chr.12: 63147027-63147027 on Build GRCh38
rs3803107 Chr.12: 63147054-63147054 on Build GRCh38
rs751123526 Chr.12: 63146994-63146994 on Build GRCh38
rs75197306 Chr.12: 63146723-63146723 on Build GRCh38
rs373659196 Chr.12: 63146799-63146799 on Build GRCh38
rs766381211 Chr.12: 63146950-63146950 on Build GRCh38
rs1035977597 Chr.12: 63146680-63146680 on Build GRCh38
rs948961572 Chr.12: 63147077-63147077 on Build GRCh38
rs1050079371 Chr.12: 63146987-63146987 on Build GRCh38
rs557186765 Chr.12: 63146844-63146844 on Build GRCh38
rs999134421 Chr.12: 63147039-63147039 on Build GRCh38
rs189748175 Chr.12: 63147098-63147098 on Build GRCh38
rs921235338 Chr.12: 63147022-63147022 on Build GRCh38
rs909393877 Chr.12: 63146802-63146802 on Build GRCh38
rs541584059 Chr.12: 63147067-63147067 on Build GRCh38
rs950583737 Chr.12: 63146846-63146847 on Build GRCh38
rs765411884 Chr.12: 63147115-63147115 on Build GRCh38
rs763426926 Chr.12: 63147089-63147089 on Build GRCh38
rs536931353 Chr.12: 63146807-63146807 on Build GRCh38
rs1006035082 Chr.12: 63146885-63146885 on Build GRCh38
rs538977382 Chr.12: 63146995-63146995 on Build GRCh38
rs940818330 Chr.12: 63146843-63146843 on Build GRCh38
rs11174811 Chr.12: 63146696-63146696 on Build GRCh38
rs559000770 Chr.12: 63147050-63147050 on Build GRCh38
rs1023628977 Chr.12: 63146707-63146707 on Build GRCh38
rs975827362 Chr.12: 63147004-63147004 on Build GRCh38
rs947156149 Chr.12: 63147001-63147001 on Build GRCh38
rs78249943 Chr.12: 63147023-63147023 on Build GRCh38
rs933565899 Chr.12: 63146930-63146930 on Build GRCh38
rs903019628 Chr.12: 63147026-63147026 on Build GRCh38
rs1044035226 Chr.12: 63146720-63146720 on Build GRCh38
rs895765729 Chr.12: 63147062-63147062 on Build GRCh38
rs563118991 Chr.12: 63146672-63146672 on Build GRCh38
rs532096187 Chr.12: 63146690-63146690 on Build GRCh38

View Full Product Details