Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36322388-36322856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVA1B
Entrez Gene ID
55194
Gene Name
eva-1 homolog B (C. elegans)
Gene Aliases
C1orf78, FAM176B
UniGene
Hs.87016
Gene Chromosome Location
Chr.1: 36322030-36324154 on Build GRCh38
Transcripts
NM_001304762.1 NM_018166.2 NR_130899.1 XM_017001627.1 XM_017002340.1 XM_017002341.1 XM_017002344.1 XM_017002345.1

Gene Symbol
SH3D21
Entrez Gene ID
79729
Gene Name
SH3 domain containing 21
Gene Aliases
C1orf113
UniGene
Hs.524496
Gene Chromosome Location
Chr.1: 36306348-36329221 on Build GRCh38
Transcripts
NM_001304762.1 NM_018166.2 NR_130899.1 XM_017001627.1 XM_017002340.1 XM_017002341.1 XM_017002344.1 XM_017002345.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682736 c.266T>C p.L89P Chr.1: 36322527-36322527 on Build GRCh38
4889648 c.175C>A p.P59T Chr.1: 36322618-36322618 on Build GRCh38
7289471 c.172C>T p.R58W Chr.1: 36322621-36322621 on Build GRCh38
4435822 c.393C>T p.I131I Chr.1: 36322400-36322400 on Build GRCh38
4682735 c.270C>G p.G90G Chr.1: 36322523-36322523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749064456 Chr.1: 36322577-36322577 on Build GRCh38
rs532076353 Chr.1: 36322453-36322453 on Build GRCh38
rs377723027 Chr.1: 36322498-36322498 on Build GRCh38
rs776974741 Chr.1: 36322597-36322597 on Build GRCh38
rs756560795 Chr.1: 36322416-36322416 on Build GRCh38
rs754549399 Chr.1: 36322543-36322543 on Build GRCh38
rs7536394 Chr.1: 36322755-36322755 on Build GRCh38
rs201954279 Chr.1: 36322553-36322553 on Build GRCh38
rs1034370838 Chr.1: 36322725-36322725 on Build GRCh38
rs565642335 Chr.1: 36322559-36322559 on Build GRCh38
rs766073439 Chr.1: 36322638-36322638 on Build GRCh38
rs768703283 Chr.1: 36322752-36322752 on Build GRCh38
rs762436034 Chr.1: 36322607-36322607 on Build GRCh38
rs760727308 Chr.1: 36322592-36322592 on Build GRCh38
rs766889776 Chr.1: 36322775-36322775 on Build GRCh38
rs745604875 Chr.1: 36322441-36322441 on Build GRCh38
rs985813761 Chr.1: 36322798-36322798 on Build GRCh38
rs779763338 Chr.1: 36322464-36322464 on Build GRCh38
rs535852437 Chr.1: 36322626-36322626 on Build GRCh38
rs201068811 Chr.1: 36322567-36322567 on Build GRCh38
rs774404441 Chr.1: 36322489-36322489 on Build GRCh38
rs141040011 Chr.1: 36322504-36322504 on Build GRCh38
rs145646808 Chr.1: 36322547-36322547 on Build GRCh38
rs897627064 Chr.1: 36322463-36322463 on Build GRCh38
rs565482818 Chr.1: 36322510-36322510 on Build GRCh38
rs747841324 Chr.1: 36322478-36322478 on Build GRCh38
rs536998405 Chr.1: 36322751-36322751 on Build GRCh38
rs746542181 Chr.1: 36322473-36322473 on Build GRCh38
rs765689469 Chr.1: 36322615-36322615 on Build GRCh38
rs1000965000 Chr.1: 36322797-36322797 on Build GRCh38
rs778420180 Chr.1: 36322437-36322437 on Build GRCh38
rs576634477 Chr.1: 36322819-36322819 on Build GRCh38
rs542369630 Chr.1: 36322637-36322637 on Build GRCh38
rs769697190 Chr.1: 36322488-36322488 on Build GRCh38
rs756037850 Chr.1: 36322701-36322701 on Build GRCh38
rs757650928 Chr.1: 36322431-36322431 on Build GRCh38
rs1026452051 Chr.1: 36322687-36322687 on Build GRCh38
rs1049403603 Chr.1: 36322736-36322736 on Build GRCh38
rs759849054 Chr.1: 36322490-36322490 on Build GRCh38
rs910288595 Chr.1: 36322816-36322816 on Build GRCh38
rs1021471434 Chr.1: 36322546-36322546 on Build GRCh38
rs868607127 Chr.1: 36322644-36322644 on Build GRCh38
rs200217271 Chr.1: 36322422-36322422 on Build GRCh38
rs749050710 Chr.1: 36322746-36322746 on Build GRCh38
rs754506123 Chr.1: 36322652-36322652 on Build GRCh38
rs751315847 Chr.1: 36322645-36322645 on Build GRCh38
rs753179253 Chr.1: 36322415-36322415 on Build GRCh38
rs779886034 Chr.1: 36322521-36322521 on Build GRCh38
rs766781878 Chr.1: 36322554-36322559 on Build GRCh38
rs764317016 Chr.1: 36322600-36322617 on Build GRCh38
rs113936116 Chr.1: 36322827-36322827 on Build GRCh38
rs754130794 Chr.1: 36322429-36322429 on Build GRCh38
rs767623016 Chr.1: 36322409-36322409 on Build GRCh38
rs201498075 Chr.1: 36322477-36322477 on Build GRCh38
rs1040163248 Chr.1: 36322430-36322430 on Build GRCh38
rs764523478 Chr.1: 36322501-36322501 on Build GRCh38
rs888540820 Chr.1: 36322675-36322675 on Build GRCh38
rs746452686 Chr.1: 36322530-36322530 on Build GRCh38
rs768316044 Chr.1: 36322688-36322688 on Build GRCh38
rs757562480 Chr.1: 36322506-36322506 on Build GRCh38
rs929154659 Chr.1: 36322733-36322733 on Build GRCh38
rs550387297 Chr.1: 36322499-36322499 on Build GRCh38
rs773010275 Chr.1: 36322561-36322561 on Build GRCh38
rs758069931 Chr.1: 36322748-36322748 on Build GRCh38
rs910186004 Chr.1: 36322568-36322568 on Build GRCh38
rs532193645 Chr.1: 36322512-36322512 on Build GRCh38
rs896463343 Chr.1: 36322765-36322765 on Build GRCh38
rs777572883 Chr.1: 36322745-36322745 on Build GRCh38
rs758023174 Chr.1: 36322518-36322518 on Build GRCh38
rs1005591187 Chr.1: 36322474-36322474 on Build GRCh38
rs751232112 Chr.1: 36322777-36322777 on Build GRCh38
rs887621770 Chr.1: 36322743-36322743 on Build GRCh38
rs977555243 Chr.1: 36322545-36322545 on Build GRCh38
rs1002037077 Chr.1: 36322744-36322744 on Build GRCh38
rs746798396 Chr.1: 36322800-36322800 on Build GRCh38
rs547818675 Chr.1: 36322630-36322630 on Build GRCh38
rs773606790 Chr.1: 36322770-36322770 on Build GRCh38
rs967971768 Chr.1: 36322517-36322517 on Build GRCh38
rs780801449 Chr.1: 36322654-36322654 on Build GRCh38
rs775700586 Chr.1: 36322588-36322588 on Build GRCh38
rs965682593 Chr.1: 36322836-36322836 on Build GRCh38
rs932856047 Chr.1: 36322574-36322574 on Build GRCh38
rs763397436 Chr.1: 36322771-36322771 on Build GRCh38
rs954209080 Chr.1: 36322810-36322810 on Build GRCh38
rs576702559 Chr.1: 36322555-36322555 on Build GRCh38
rs754190302 Chr.1: 36322603-36322620 on Build GRCh38
rs929163411 Chr.1: 36322633-36322633 on Build GRCh38
rs770270546 Chr.1: 36322764-36322764 on Build GRCh38
rs752170638 Chr.1: 36322700-36322700 on Build GRCh38

View Full Product Details