Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85729799-85730223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL24A1
Entrez Gene ID
255631
Gene Name
collagen type XXIV alpha 1
Gene Aliases
-
UniGene
Hs.659516
Gene Chromosome Location
Chr.1: 85728507-86156987 on Build GRCh38
Transcripts
NM_152890.5 XM_017000924.1 XM_017000925.1 XM_017000926.1 XM_017000927.1 XM_017000928.1 XM_017000929.1

SNP Detail

SNP ID Chromosome Location
rs1002323518 Chr.1: 85730138-85730138 on Build GRCh38
rs775466927 Chr.1: 85730037-85730037 on Build GRCh38
rs1016500014 Chr.1: 85729915-85729915 on Build GRCh38
rs993277435 Chr.1: 85730083-85730083 on Build GRCh38
rs926321400 Chr.1: 85729828-85729828 on Build GRCh38
rs115657728 Chr.1: 85729874-85729874 on Build GRCh38
rs749543822 Chr.1: 85730018-85730018 on Build GRCh38
rs989166121 Chr.1: 85730198-85730198 on Build GRCh38
rs542633479 Chr.1: 85730176-85730176 on Build GRCh38
rs934300154 Chr.1: 85730100-85730100 on Build GRCh38
rs146223257 Chr.1: 85730050-85730050 on Build GRCh38
rs116292974 Chr.1: 85730030-85730030 on Build GRCh38
rs1026818594 Chr.1: 85729875-85729875 on Build GRCh38
rs984800962 Chr.1: 85729907-85729907 on Build GRCh38
rs950660889 Chr.1: 85729890-85729890 on Build GRCh38
rs574331642 Chr.1: 85729835-85729835 on Build GRCh38
rs148263455 Chr.1: 85730179-85730179 on Build GRCh38
rs914842085 Chr.1: 85729886-85729886 on Build GRCh38
rs997766411 Chr.1: 85730199-85730199 on Build GRCh38
rs924363929 Chr.1: 85730092-85730092 on Build GRCh38
rs914473195 Chr.1: 85730132-85730132 on Build GRCh38
rs946175460 Chr.1: 85729897-85729897 on Build GRCh38
rs1050534449 Chr.1: 85730017-85730017 on Build GRCh38
rs746815037 Chr.1: 85730081-85730081 on Build GRCh38
rs990971356 Chr.1: 85730022-85730022 on Build GRCh38
rs1041933866 Chr.1: 85729906-85729906 on Build GRCh38
rs41289765 Chr.1: 85730121-85730121 on Build GRCh38
rs901916386 Chr.1: 85729975-85729975 on Build GRCh38
rs776645405 Chr.1: 85730185-85730185 on Build GRCh38
rs943323329 Chr.1: 85730151-85730151 on Build GRCh38
rs1025261810 Chr.1: 85730090-85730090 on Build GRCh38
rs933437918 Chr.1: 85729993-85729993 on Build GRCh38
rs564016408 Chr.1: 85730106-85730106 on Build GRCh38

View Full Product Details