Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86468674-86469205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA1
Entrez Gene ID
1179
Gene Name
chloride channel accessory 1
Gene Aliases
CACC, CACC1, CLCRG1, CaCC-1, GOB5, hCLCA1, hCaCC-1
UniGene
Hs.194659
Gene Chromosome Location
Chr.1: 86468843-86500294 on Build GRCh38
Transcripts
NM_001285.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3985275 c.52G>A p.G18R Chr.1: 86469023-86469023 on Build GRCh38
7422867 c.55G>A p.A19T Chr.1: 86469026-86469026 on Build GRCh38
5001198 c.107T>C p.V36A Chr.1: 86469078-86469078 on Build GRCh38
4423069 c.109G>A p.V37I Chr.1: 86469080-86469080 on Build GRCh38
912781 c.20C>A p.S7Y Chr.1: 86468991-86468991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569922132 Chr.1: 86469091-86469091 on Build GRCh38
rs376749837 Chr.1: 86469146-86469146 on Build GRCh38
rs901149405 Chr.1: 86468808-86468808 on Build GRCh38
rs770258919 Chr.1: 86468972-86468972 on Build GRCh38
rs761846884 Chr.1: 86468888-86468888 on Build GRCh38
rs368956046 Chr.1: 86469021-86469021 on Build GRCh38
rs751061300 Chr.1: 86469033-86469033 on Build GRCh38
rs890074708 Chr.1: 86468857-86468857 on Build GRCh38
rs772480629 Chr.1: 86469129-86469129 on Build GRCh38
rs1046330704 Chr.1: 86468721-86468721 on Build GRCh38
rs565663181 Chr.1: 86468736-86468736 on Build GRCh38
rs763510318 Chr.1: 86468975-86468975 on Build GRCh38
rs749193711 Chr.1: 86468944-86468944 on Build GRCh38
rs76928978 Chr.1: 86468787-86468787 on Build GRCh38
rs190336859 Chr.1: 86468932-86468932 on Build GRCh38
rs777666738 Chr.1: 86468955-86468955 on Build GRCh38
rs535366156 Chr.1: 86469120-86469120 on Build GRCh38
rs754939887 Chr.1: 86468946-86468946 on Build GRCh38
rs1017081578 Chr.1: 86469159-86469159 on Build GRCh38
rs371305518 Chr.1: 86469076-86469076 on Build GRCh38
rs762283718 Chr.1: 86469075-86469075 on Build GRCh38
rs746527117 Chr.1: 86469139-86469139 on Build GRCh38
rs781475493 Chr.1: 86468967-86468967 on Build GRCh38
rs755354335 Chr.1: 86469107-86469107 on Build GRCh38
rs1014356274 Chr.1: 86468848-86468848 on Build GRCh38
rs377703691 Chr.1: 86469023-86469023 on Build GRCh38
rs746959458 Chr.1: 86468900-86468900 on Build GRCh38
rs893280412 Chr.1: 86469168-86469168 on Build GRCh38
rs749679890 Chr.1: 86469066-86469066 on Build GRCh38
rs762896434 Chr.1: 86469166-86469166 on Build GRCh38
rs998169365 Chr.1: 86468822-86468822 on Build GRCh38
rs949262259 Chr.1: 86468703-86468703 on Build GRCh38
rs199987194 Chr.1: 86469162-86469162 on Build GRCh38
rs550010719 Chr.1: 86469088-86469088 on Build GRCh38
rs760273429 Chr.1: 86469080-86469080 on Build GRCh38
rs189550563 Chr.1: 86469180-86469180 on Build GRCh38
rs549849538 Chr.1: 86468790-86468790 on Build GRCh38
rs760363228 Chr.1: 86469014-86469014 on Build GRCh38
rs770168920 Chr.1: 86469047-86469047 on Build GRCh38
rs373512851 Chr.1: 86469096-86469096 on Build GRCh38
rs542367977 Chr.1: 86469004-86469004 on Build GRCh38
rs779193119 Chr.1: 86469118-86469118 on Build GRCh38
rs1045954929 Chr.1: 86468849-86468849 on Build GRCh38
rs55960532 Chr.1: 86468924-86468924 on Build GRCh38
rs759582156 Chr.1: 86468698-86468698 on Build GRCh38
rs192477945 Chr.1: 86468856-86468856 on Build GRCh38
rs201221009 Chr.1: 86469017-86469017 on Build GRCh38
rs753566864 Chr.1: 86469084-86469084 on Build GRCh38
rs765003183 Chr.1: 86469089-86469089 on Build GRCh38
rs922504951 Chr.1: 86469009-86469009 on Build GRCh38
rs35155356 Chr.1: 86468931-86468931 on Build GRCh38
rs370725300 Chr.1: 86469177-86469177 on Build GRCh38
rs1017525486 Chr.1: 86468893-86468893 on Build GRCh38
rs772651975 Chr.1: 86469133-86469133 on Build GRCh38
rs143958809 Chr.1: 86468925-86468925 on Build GRCh38
rs776035113 Chr.1: 86468973-86468973 on Build GRCh38
rs112643007 Chr.1: 86469058-86469058 on Build GRCh38
rs747130094 Chr.1: 86468961-86468961 on Build GRCh38
rs909629810 Chr.1: 86468750-86468750 on Build GRCh38
rs750075037 Chr.1: 86469008-86469008 on Build GRCh38
rs762906880 Chr.1: 86469017-86469018 on Build GRCh38
rs976495008 Chr.1: 86468980-86468980 on Build GRCh38
rs375519423 Chr.1: 86468927-86468927 on Build GRCh38
rs375577694 Chr.1: 86468934-86468934 on Build GRCh38
rs2791518 Chr.1: 86468949-86468949 on Build GRCh38
rs183772879 Chr.1: 86468753-86468753 on Build GRCh38
rs189623745 Chr.1: 86468781-86468781 on Build GRCh38
rs998996191 Chr.1: 86468896-86468896 on Build GRCh38
rs756342469 Chr.1: 86469098-86469098 on Build GRCh38
rs918933484 Chr.1: 86468701-86468701 on Build GRCh38
rs770995884 Chr.1: 86468963-86468963 on Build GRCh38
rs775177648 Chr.1: 86469164-86469164 on Build GRCh38
rs200977918 Chr.1: 86469041-86469041 on Build GRCh38
rs535479637 Chr.1: 86468908-86468908 on Build GRCh38
rs143108847 Chr.1: 86469074-86469074 on Build GRCh38
rs773042182 Chr.1: 86468961-86468962 on Build GRCh38
rs781181374 Chr.1: 86469034-86469034 on Build GRCh38
rs201493645 Chr.1: 86468993-86468993 on Build GRCh38
rs749587140 Chr.1: 86469103-86469103 on Build GRCh38
rs756258522 Chr.1: 86469178-86469178 on Build GRCh38
rs778917601 Chr.1: 86469022-86469022 on Build GRCh38
rs375828178 Chr.1: 86468986-86468986 on Build GRCh38
rs780514083 Chr.1: 86469062-86469062 on Build GRCh38
rs746137570 Chr.1: 86468968-86468968 on Build GRCh38
rs759239138 Chr.1: 86469161-86469161 on Build GRCh38
rs770498864 Chr.1: 86469144-86469144 on Build GRCh38
rs926556489 Chr.1: 86468752-86468752 on Build GRCh38
rs146601251 Chr.1: 86468929-86468929 on Build GRCh38
rs1054067816 Chr.1: 86468769-86468769 on Build GRCh38
rs145304242 Chr.1: 86469015-86469015 on Build GRCh38
rs780428205 Chr.1: 86469101-86469101 on Build GRCh38
rs763948147 Chr.1: 86469176-86469176 on Build GRCh38
rs139168398 Chr.1: 86469079-86469079 on Build GRCh38

View Full Product Details