Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156243290-156243882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BGLAP
Entrez Gene ID
632
Gene Name
bone gamma-carboxyglutamate protein
Gene Aliases
BGP, OC, OCN
UniGene
Hs.654541
Gene Chromosome Location
Chr.1: 156241962-156243332 on Build GRCh38
Transcripts
NM_199173.5 NM_001272104.1 NM_001272105.1 NM_001272106.1 NM_001272107.1 NM_001272108.1 NM_001272109.1 NM_001272110.1 NM_001272111.1 NM_001272112.1 NM_001272113.1 NM_024897.3 NM_198406.2 NR_073610.1 XM_005245494.3 XM_006711546.2 XM_006711547.3 XM_006711548.3 XM_006711552.2 XM_006711553.3 XM_011510000.2 XM_017002385.1 NM_001199661.1 NM_001199662.1 NM_001199663.1 NM_001199664.1

Gene Symbol
PAQR6
Entrez Gene ID
79957
Gene Name
progestin and adipoQ receptor family member 6
Gene Aliases
PRdelta
UniGene
Hs.235873
Gene Chromosome Location
Chr.1: 156243321-156248488 on Build GRCh38
Transcripts
NM_199173.5 NM_001272104.1 NM_001272105.1 NM_001272106.1 NM_001272107.1 NM_001272108.1 NM_001272109.1 NM_001272110.1 NM_001272111.1 NM_001272112.1 NM_001272113.1 NM_024897.3 NM_198406.2 NR_073610.1 XM_005245494.3 XM_006711546.2 XM_006711547.3 XM_006711548.3 XM_006711552.2 XM_006711553.3 XM_011510000.2 XM_017002385.1 NM_001199661.1 NM_001199662.1 NM_001199663.1 NM_001199664.1

Gene Symbol
PMF1-BGLAP
Entrez Gene ID
100527963
Gene Name
PMF1-BGLAP readthrough
Gene Aliases
PMF1
UniGene
Hs.530479
Gene Chromosome Location
Chr.1: 156212988-156243332 on Build GRCh38
Transcripts
NM_199173.5 NM_001272104.1 NM_001272105.1 NM_001272106.1 NM_001272107.1 NM_001272108.1 NM_001272109.1 NM_001272110.1 NM_001272111.1 NM_001272112.1 NM_001272113.1 NM_024897.3 NM_198406.2 NR_073610.1 XM_005245494.3 XM_006711546.2 XM_006711547.3 XM_006711548.3 XM_006711552.2 XM_006711553.3 XM_011510000.2 XM_017002385.1 NM_001199661.1 NM_001199662.1 NM_001199663.1 NM_001199664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6324098 c.1049C>T p.P350L Chr.1: 156243868-156243868 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762301843 Chr.1: 156243797-156243797 on Build GRCh38
rs776986359 Chr.1: 156243501-156243504 on Build GRCh38
rs541835004 Chr.1: 156243659-156243659 on Build GRCh38
rs749245135 Chr.1: 156243703-156243703 on Build GRCh38
rs775805230 Chr.1: 156243561-156243561 on Build GRCh38
rs1041457028 Chr.1: 156243555-156243555 on Build GRCh38
rs568568157 Chr.1: 156243831-156243831 on Build GRCh38
rs900424912 Chr.1: 156243796-156243796 on Build GRCh38
rs768782784 Chr.1: 156243747-156243747 on Build GRCh38
rs1037481314 Chr.1: 156243389-156243389 on Build GRCh38
rs550288776 Chr.1: 156243844-156243844 on Build GRCh38
rs750083705 Chr.1: 156243826-156243826 on Build GRCh38
rs1015593644 Chr.1: 156243523-156243523 on Build GRCh38
rs1010156201 Chr.1: 156243586-156243586 on Build GRCh38
rs774352842 Chr.1: 156243759-156243759 on Build GRCh38
rs527984261 Chr.1: 156243704-156243704 on Build GRCh38
rs377649430 Chr.1: 156243825-156243825 on Build GRCh38
rs763122545 Chr.1: 156243578-156243578 on Build GRCh38
rs1032427777 Chr.1: 156243575-156243575 on Build GRCh38
rs903638482 Chr.1: 156243379-156243379 on Build GRCh38
rs138743544 Chr.1: 156243692-156243692 on Build GRCh38
rs1024990392 Chr.1: 156243471-156243471 on Build GRCh38
rs371351556 Chr.1: 156243815-156243815 on Build GRCh38
rs4350194 Chr.1: 156243386-156243386 on Build GRCh38
rs754303155 Chr.1: 156243820-156243820 on Build GRCh38
rs764856916 Chr.1: 156243507-156243516 on Build GRCh38
rs1032867680 Chr.1: 156243821-156243821 on Build GRCh38
rs775512023 Chr.1: 156243679-156243680 on Build GRCh38
rs184955812 Chr.1: 156243728-156243728 on Build GRCh38
rs974136537 Chr.1: 156243541-156243541 on Build GRCh38
rs756776427 Chr.1: 156243822-156243822 on Build GRCh38
rs1021081294 Chr.1: 156243628-156243628 on Build GRCh38
rs765581949 Chr.1: 156243824-156243825 on Build GRCh38
rs562639150 Chr.1: 156243580-156243580 on Build GRCh38
rs953900653 Chr.1: 156243604-156243604 on Build GRCh38
rs570705427 Chr.1: 156243734-156243734 on Build GRCh38
rs746804301 Chr.1: 156243442-156243442 on Build GRCh38
rs945309392 Chr.1: 156243683-156243683 on Build GRCh38
rs893892631 Chr.1: 156243818-156243818 on Build GRCh38
rs996784489 Chr.1: 156243568-156243568 on Build GRCh38
rs1008793178 Chr.1: 156243516-156243516 on Build GRCh38
rs370993197 Chr.1: 156243829-156243829 on Build GRCh38
rs944526055 Chr.1: 156243779-156243779 on Build GRCh38
rs764615228 Chr.1: 156243817-156243817 on Build GRCh38
rs920342447 Chr.1: 156243404-156243404 on Build GRCh38
rs937754584 Chr.1: 156243380-156243380 on Build GRCh38
rs936279672 Chr.1: 156243707-156243707 on Build GRCh38
rs982354669 Chr.1: 156243680-156243680 on Build GRCh38
rs577157502 Chr.1: 156243497-156243497 on Build GRCh38
rs531825847 Chr.1: 156243756-156243756 on Build GRCh38
rs773499849 Chr.1: 156243838-156243839 on Build GRCh38
rs770414810 Chr.1: 156243502-156243516 on Build GRCh38
rs572880549 Chr.1: 156243529-156243529 on Build GRCh38
rs535875639 Chr.1: 156243383-156243383 on Build GRCh38
rs894648894 Chr.1: 156243411-156243411 on Build GRCh38
rs554545336 Chr.1: 156243400-156243400 on Build GRCh38
rs775809055 Chr.1: 156243812-156243812 on Build GRCh38
rs4316359 Chr.1: 156243331-156243331 on Build GRCh38
rs748100466 Chr.1: 156243839-156243839 on Build GRCh38
rs1038973469 Chr.1: 156243789-156243789 on Build GRCh38
rs912500029 Chr.1: 156243680-156243680 on Build GRCh38
rs770981340 Chr.1: 156243847-156243847 on Build GRCh38
rs962802842 Chr.1: 156243537-156243537 on Build GRCh38
rs187592701 Chr.1: 156243455-156243455 on Build GRCh38
rs759330 Chr.1: 156243466-156243466 on Build GRCh38
rs374026893 Chr.1: 156243830-156243830 on Build GRCh38
rs970968842 Chr.1: 156243319-156243319 on Build GRCh38
rs982185882 Chr.1: 156243322-156243322 on Build GRCh38
rs192668510 Chr.1: 156243504-156243504 on Build GRCh38
rs529638145 Chr.1: 156243644-156243644 on Build GRCh38
rs926283204 Chr.1: 156243353-156243353 on Build GRCh38
rs549845883 Chr.1: 156243773-156243773 on Build GRCh38
rs1003836043 Chr.1: 156243789-156243800 on Build GRCh38
rs781009160 Chr.1: 156243836-156243836 on Build GRCh38
rs540389894 Chr.1: 156243462-156243462 on Build GRCh38

View Full Product Details