Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111139185-111139773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEPT1
Entrez Gene ID
10390
Gene Name
choline/ethanolamine phosphotransferase 1
Gene Aliases
-
UniGene
Hs.363572
Gene Chromosome Location
Chr.1: 111139436-111185102 on Build GRCh38
Transcripts
NM_001007794.1 NM_178454.4 XM_005270469.2 XM_005270470.1 XM_006710361.1 XM_006710362.1 XM_011540707.1 XM_011540708.1 XM_017000320.1 XM_017000321.1

Gene Symbol
DRAM2
Entrez Gene ID
128338
Gene Name
DNA damage regulated autophagy modulator 2
Gene Aliases
CORD21, PRO180, TMEM77, WWFQ154
UniGene
Hs.485606
Gene Chromosome Location
Chr.1: 111117332-111140216 on Build GRCh38
Transcripts
NM_001007794.1 NM_178454.4 XM_005270469.2 XM_005270470.1 XM_006710361.1 XM_006710362.1 XM_011540707.1 XM_011540708.1 XM_017000320.1 XM_017000321.1

SNP Detail

SNP ID Chromosome Location
rs1050028672 Chr.1: 111139750-111139750 on Build GRCh38
rs991266804 Chr.1: 111139677-111139677 on Build GRCh38
rs756950704 Chr.1: 111139457-111139457 on Build GRCh38
rs557483184 Chr.1: 111139519-111139519 on Build GRCh38
rs945350981 Chr.1: 111139587-111139587 on Build GRCh38
rs137970027 Chr.1: 111139386-111139386 on Build GRCh38
rs150737151 Chr.1: 111139321-111139321 on Build GRCh38
rs1044282591 Chr.1: 111139225-111139225 on Build GRCh38
rs3762374 Chr.1: 111139496-111139496 on Build GRCh38
rs971916780 Chr.1: 111139330-111139330 on Build GRCh38
rs980035570 Chr.1: 111139522-111139522 on Build GRCh38
rs187969701 Chr.1: 111139434-111139434 on Build GRCh38
rs181233662 Chr.1: 111139482-111139482 on Build GRCh38
rs934659355 Chr.1: 111139492-111139492 on Build GRCh38
rs144025898 Chr.1: 111139586-111139586 on Build GRCh38
rs996999819 Chr.1: 111139508-111139508 on Build GRCh38
rs1053879430 Chr.1: 111139287-111139287 on Build GRCh38
rs760199908 Chr.1: 111139218-111139218 on Build GRCh38
rs899304618 Chr.1: 111139689-111139689 on Build GRCh38
rs548552914 Chr.1: 111139227-111139227 on Build GRCh38
rs551613178 Chr.1: 111139296-111139296 on Build GRCh38
rs1043096 Chr.1: 111139685-111139685 on Build GRCh38
rs1026070441 Chr.1: 111139471-111139471 on Build GRCh38
rs764816254 Chr.1: 111139496-111139512 on Build GRCh38
rs950205511 Chr.1: 111139415-111139415 on Build GRCh38
rs924569592 Chr.1: 111139485-111139485 on Build GRCh38
rs1051801068 Chr.1: 111139603-111139603 on Build GRCh38
rs114806973 Chr.1: 111139618-111139618 on Build GRCh38
rs963530516 Chr.1: 111139453-111139453 on Build GRCh38
rs890731672 Chr.1: 111139394-111139394 on Build GRCh38
rs183703398 Chr.1: 111139269-111139269 on Build GRCh38
rs951985744 Chr.1: 111139670-111139670 on Build GRCh38
rs1049951388 Chr.1: 111139514-111139514 on Build GRCh38
rs750817782 Chr.1: 111139606-111139606 on Build GRCh38
rs563966465 Chr.1: 111139605-111139605 on Build GRCh38
rs892428646 Chr.1: 111139303-111139303 on Build GRCh38
rs149492217 Chr.1: 111139581-111139581 on Build GRCh38
rs549152891 Chr.1: 111139715-111139715 on Build GRCh38
rs935829178 Chr.1: 111139555-111139555 on Build GRCh38
rs981902123 Chr.1: 111139695-111139695 on Build GRCh38
rs79428887 Chr.1: 111139339-111139339 on Build GRCh38
rs545926755 Chr.1: 111139684-111139684 on Build GRCh38
rs961755416 Chr.1: 111139313-111139313 on Build GRCh38
rs925738474 Chr.1: 111139544-111139544 on Build GRCh38
rs924755454 Chr.1: 111139700-111139700 on Build GRCh38
rs622103 Chr.1: 111139566-111139566 on Build GRCh38
rs576875414 Chr.1: 111139628-111139628 on Build GRCh38
rs991319157 Chr.1: 111139382-111139382 on Build GRCh38
rs539180066 Chr.1: 111139385-111139385 on Build GRCh38
rs780435032 Chr.1: 111139659-111139659 on Build GRCh38
rs930469691 Chr.1: 111139371-111139371 on Build GRCh38
rs992543604 Chr.1: 111139498-111139498 on Build GRCh38
rs932861134 Chr.1: 111139730-111139730 on Build GRCh38
rs536682102 Chr.1: 111139240-111139240 on Build GRCh38
rs915697655 Chr.1: 111139679-111139679 on Build GRCh38
rs995048649 Chr.1: 111139711-111139711 on Build GRCh38
rs1024632132 Chr.1: 111139500-111139500 on Build GRCh38
rs969936135 Chr.1: 111139513-111139513 on Build GRCh38
rs1017697355 Chr.1: 111139421-111139421 on Build GRCh38
rs948530991 Chr.1: 111139222-111139222 on Build GRCh38
rs1018998542 Chr.1: 111139294-111139294 on Build GRCh38
rs763554736 Chr.1: 111139242-111139242 on Build GRCh38
rs914814629 Chr.1: 111139716-111139716 on Build GRCh38
rs1007668095 Chr.1: 111139397-111139397 on Build GRCh38
rs567645597 Chr.1: 111139229-111139229 on Build GRCh38
rs1028744695 Chr.1: 111139220-111139220 on Build GRCh38

View Full Product Details