Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151097881-151098192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABPB2
Entrez Gene ID
126626
Gene Name
GA binding protein transcription factor beta subunit 2
Gene Aliases
GABPB-2
UniGene
Hs.15671
Gene Chromosome Location
Chr.1: 151070180-151125545 on Build GRCh38
Transcripts
NM_001323906.1 NM_001323907.1 NM_001323908.1 NM_001323909.1 NM_001323910.1 NM_001323911.1 NM_001323912.1 NM_001323913.1 NM_144618.2 XM_017000246.1 XM_017000247.1 XM_017000248.1 XM_017000249.1 XM_017000250.1 XM_017000251.1 XM_017000252.1 XM_017000253.1 XM_017000254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3976115 c.508A>G p.R170G Chr.1: 151097888-151097888 on Build GRCh38
7281820 c.560C>T p.T187M Chr.1: 151097940-151097940 on Build GRCh38
3473582 c.511G>A p.A171T Chr.1: 151097891-151097891 on Build GRCh38
5377581 c.518C>T p.P173L Chr.1: 151097898-151097898 on Build GRCh38
7417006 c.616A>T p.T206S Chr.1: 151097996-151097996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960668650 Chr.1: 151098144-151098144 on Build GRCh38
rs749118987 Chr.1: 151097933-151097933 on Build GRCh38
rs774372293 Chr.1: 151097979-151097982 on Build GRCh38
rs772347612 Chr.1: 151098039-151098039 on Build GRCh38
rs748101564 Chr.1: 151097923-151097923 on Build GRCh38
rs923945945 Chr.1: 151098059-151098059 on Build GRCh38
rs904840787 Chr.1: 151098050-151098050 on Build GRCh38
rs934742844 Chr.1: 151097976-151097976 on Build GRCh38
rs753128987 Chr.1: 151098090-151098090 on Build GRCh38
rs978000727 Chr.1: 151098036-151098036 on Build GRCh38
rs751191805 Chr.1: 151097912-151097912 on Build GRCh38
rs747119533 Chr.1: 151098049-151098049 on Build GRCh38
rs1049677763 Chr.1: 151097904-151097904 on Build GRCh38
rs139033835 Chr.1: 151097947-151097947 on Build GRCh38
rs1022534454 Chr.1: 151098000-151098000 on Build GRCh38
rs587619026 Chr.1: 151097983-151097983 on Build GRCh38
rs587774461 Chr.1: 151098055-151098055 on Build GRCh38
rs961731154 Chr.1: 151098149-151098149 on Build GRCh38
rs374570194 Chr.1: 151097962-151097962 on Build GRCh38
rs751227478 Chr.1: 151097964-151097964 on Build GRCh38
rs777468273 Chr.1: 151098020-151098020 on Build GRCh38
rs111628112 Chr.1: 151097929-151097929 on Build GRCh38
rs772105540 Chr.1: 151097946-151097946 on Build GRCh38
rs764605178 Chr.1: 151097951-151097951 on Build GRCh38
rs587637546 Chr.1: 151098165-151098165 on Build GRCh38
rs138089875 Chr.1: 151097981-151097981 on Build GRCh38
rs368892897 Chr.1: 151098031-151098031 on Build GRCh38
rs888065509 Chr.1: 151097916-151097916 on Build GRCh38
rs754495576 Chr.1: 151097918-151097918 on Build GRCh38
rs770812873 Chr.1: 151097941-151097941 on Build GRCh38
rs896785154 Chr.1: 151097940-151097940 on Build GRCh38
rs777080160 Chr.1: 151097960-151097960 on Build GRCh38
rs780739967 Chr.1: 151097921-151097921 on Build GRCh38
rs369896527 Chr.1: 151097908-151097908 on Build GRCh38
rs767011654 Chr.1: 151097985-151097985 on Build GRCh38
rs993225256 Chr.1: 151098150-151098150 on Build GRCh38
rs765611292 Chr.1: 151097963-151097963 on Build GRCh38
rs368996122 Chr.1: 151098001-151098001 on Build GRCh38
rs768689361 Chr.1: 151098053-151098053 on Build GRCh38
rs369983790 Chr.1: 151098045-151098045 on Build GRCh38
rs968026778 Chr.1: 151098019-151098019 on Build GRCh38
rs756125769 Chr.1: 151097925-151097925 on Build GRCh38
rs910538308 Chr.1: 151097926-151097926 on Build GRCh38
rs150081114 Chr.1: 151097997-151097997 on Build GRCh38
rs746084688 Chr.1: 151097944-151097944 on Build GRCh38
rs774480090 Chr.1: 151097943-151097943 on Build GRCh38
rs778759880 Chr.1: 151098021-151098021 on Build GRCh38
rs1009413152 Chr.1: 151097952-151097952 on Build GRCh38
rs777890508 Chr.1: 151098009-151098009 on Build GRCh38
rs771332503 Chr.1: 151098096-151098096 on Build GRCh38
rs149526187 Chr.1: 151098166-151098166 on Build GRCh38

View Full Product Details