Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35455555-35455884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP250
Entrez Gene ID
11190
Gene Name
centrosomal protein 250
Gene Aliases
C-NAP1, CEP2, CNAP1
UniGene
Hs.443976
Gene Chromosome Location
Chr.20: 35455138-35517531 on Build GRCh38
Transcripts
NM_007186.5 XM_005260262.4 XM_005260263.4 XM_005260264.4 XM_006723690.3 XM_006723691.1 XM_006723692.3 XM_006723693.3 XM_006723694.3 XM_011528517.2 XM_011528518.2 XM_017027617.1

SNP Detail

SNP ID Chromosome Location
rs1055688934 Chr.20: 35455588-35455588 on Build GRCh38
rs551772318 Chr.20: 35455607-35455607 on Build GRCh38
rs1041811282 Chr.20: 35455628-35455628 on Build GRCh38
rs1036907035 Chr.20: 35455759-35455759 on Build GRCh38
rs574569359 Chr.20: 35455830-35455830 on Build GRCh38
rs766664468 Chr.20: 35455669-35455669 on Build GRCh38
rs149130070 Chr.20: 35455777-35455777 on Build GRCh38
rs907832190 Chr.20: 35455771-35455771 on Build GRCh38
rs761356024 Chr.20: 35455581-35455581 on Build GRCh38
rs932518065 Chr.20: 35455633-35455633 on Build GRCh38
rs960593125 Chr.20: 35455578-35455578 on Build GRCh38
rs1011226928 Chr.20: 35455613-35455613 on Build GRCh38
rs1019380695 Chr.20: 35455666-35455666 on Build GRCh38
rs557178340 Chr.20: 35455850-35455850 on Build GRCh38
rs966367077 Chr.20: 35455598-35455598 on Build GRCh38
rs902857873 Chr.20: 35455648-35455648 on Build GRCh38
rs1008355354 Chr.20: 35455649-35455649 on Build GRCh38
rs776861419 Chr.20: 35455730-35455730 on Build GRCh38
rs1020669593 Chr.20: 35455586-35455586 on Build GRCh38
rs1029182454 Chr.20: 35455692-35455692 on Build GRCh38
rs6120949 Chr.20: 35455811-35455811 on Build GRCh38
rs897164554 Chr.20: 35455765-35455765 on Build GRCh38
rs570186145 Chr.20: 35455680-35455680 on Build GRCh38
rs985719765 Chr.20: 35455721-35455721 on Build GRCh38
rs905921038 Chr.20: 35455824-35455824 on Build GRCh38
rs537010990 Chr.20: 35455579-35455579 on Build GRCh38
rs966582295 Chr.20: 35455672-35455672 on Build GRCh38
rs952338173 Chr.20: 35455738-35455738 on Build GRCh38
rs943986039 Chr.20: 35455743-35455743 on Build GRCh38
rs183960290 Chr.20: 35455837-35455837 on Build GRCh38
rs912384402 Chr.20: 35455724-35455724 on Build GRCh38
rs534069745 Chr.20: 35455796-35455796 on Build GRCh38
rs534393740 Chr.20: 35455740-35455740 on Build GRCh38
rs1048476337 Chr.20: 35455783-35455783 on Build GRCh38
rs979182346 Chr.20: 35455603-35455603 on Build GRCh38
rs928746966 Chr.20: 35455794-35455794 on Build GRCh38

View Full Product Details