Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33016410-33016973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1
Entrez Gene ID
2720
Gene Name
galactosidase beta 1
Gene Aliases
EBP, ELNR1, MPS4B
UniGene
Hs.443031
Gene Chromosome Location
Chr.3: 32996608-33097230 on Build GRCh38
Transcripts
NM_000404.3 NM_001079811.2 NM_001135602.2 NM_001317040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7369509 c.1408G>C p.G470R Chr.3: 33016780-33016780 on Build GRCh38
311468 c.1361T>C p.V454A Chr.3: 33016827-33016827 on Build GRCh38
1043696 c.1391A>G p.N464S Chr.3: 33016797-33016797 on Build GRCh38
5444068 c.1370G>A p.R457Q Chr.3: 33016818-33016818 on Build GRCh38
4874009 c.1535A>G p.N512S Chr.3: 33016797-33016797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764603384 Chr.3: 33016780-33016780 on Build GRCh38
rs28934886 Chr.3: 33016818-33016818 on Build GRCh38
rs374547468 Chr.3: 33016758-33016758 on Build GRCh38
rs750591988 Chr.3: 33016825-33016825 on Build GRCh38
rs267599771 Chr.3: 33016836-33016836 on Build GRCh38
rs1004005486 Chr.3: 33016611-33016611 on Build GRCh38
rs113534264 Chr.3: 33016762-33016762 on Build GRCh38
rs398123349 Chr.3: 33016721-33016722 on Build GRCh38
rs757102439 Chr.3: 33016867-33016867 on Build GRCh38
rs751683034 Chr.3: 33016850-33016850 on Build GRCh38
rs948119045 Chr.3: 33016564-33016564 on Build GRCh38
rs946759204 Chr.3: 33016913-33016913 on Build GRCh38
rs148860108 Chr.3: 33016862-33016862 on Build GRCh38
rs72555359 Chr.3: 33016819-33016819 on Build GRCh38
rs915394572 Chr.3: 33016461-33016461 on Build GRCh38
rs775103521 Chr.3: 33016765-33016765 on Build GRCh38
rs888255591 Chr.3: 33016583-33016583 on Build GRCh38
rs886767390 Chr.3: 33016599-33016599 on Build GRCh38
rs369789589 Chr.3: 33016698-33016698 on Build GRCh38
rs780232995 Chr.3: 33016717-33016717 on Build GRCh38
rs267599772 Chr.3: 33016837-33016837 on Build GRCh38
rs1049639204 Chr.3: 33016569-33016569 on Build GRCh38
rs773868843 Chr.3: 33016747-33016747 on Build GRCh38
rs758688455 Chr.3: 33016838-33016838 on Build GRCh38
rs1022476871 Chr.3: 33016708-33016708 on Build GRCh38
rs765454500 Chr.3: 33016664-33016664 on Build GRCh38
rs946518578 Chr.3: 33016508-33016508 on Build GRCh38
rs375870036 Chr.3: 33016877-33016877 on Build GRCh38
rs1056141313 Chr.3: 33016579-33016579 on Build GRCh38
rs895189075 Chr.3: 33016560-33016560 on Build GRCh38
rs909288468 Chr.3: 33016946-33016946 on Build GRCh38
rs769973733 Chr.3: 33016655-33016655 on Build GRCh38
rs1029095516 Chr.3: 33016672-33016672 on Build GRCh38
rs769290024 Chr.3: 33016941-33016941 on Build GRCh38
rs1015474270 Chr.3: 33016591-33016591 on Build GRCh38
rs767949963 Chr.3: 33016811-33016813 on Build GRCh38
rs201164962 Chr.3: 33016884-33016884 on Build GRCh38
rs946904871 Chr.3: 33016474-33016474 on Build GRCh38
rs747812191 Chr.3: 33016746-33016746 on Build GRCh38
rs777663814 Chr.3: 33016732-33016732 on Build GRCh38
rs72555365 Chr.3: 33016744-33016744 on Build GRCh38
rs72555391 Chr.3: 33016743-33016743 on Build GRCh38
rs756059173 Chr.3: 33016851-33016851 on Build GRCh38
rs1016621364 Chr.3: 33016645-33016649 on Build GRCh38
rs560770425 Chr.3: 33016947-33016947 on Build GRCh38
rs1049118700 Chr.3: 33016938-33016938 on Build GRCh38
rs770656643 Chr.3: 33016467-33016467 on Build GRCh38
rs939320656 Chr.3: 33016532-33016532 on Build GRCh38
rs948067459 Chr.3: 33016897-33016897 on Build GRCh38
rs905128003 Chr.3: 33016531-33016531 on Build GRCh38
rs369156171 Chr.3: 33016808-33016808 on Build GRCh38
rs1005814361 Chr.3: 33016588-33016588 on Build GRCh38
rs897084693 Chr.3: 33016643-33016643 on Build GRCh38
rs370951326 Chr.3: 33016873-33016873 on Build GRCh38
rs748007307 Chr.3: 33016718-33016718 on Build GRCh38
rs776918632 Chr.3: 33016795-33016795 on Build GRCh38
rs750727671 Chr.3: 33016668-33016668 on Build GRCh38
rs778815380 Chr.3: 33016868-33016868 on Build GRCh38
rs968221254 Chr.3: 33016736-33016736 on Build GRCh38
rs1056382443 Chr.3: 33016544-33016544 on Build GRCh38
rs145457348 Chr.3: 33016476-33016476 on Build GRCh38
rs554132901 Chr.3: 33016437-33016437 on Build GRCh38
rs777856183 Chr.3: 33016856-33016856 on Build GRCh38
rs557275388 Chr.3: 33016811-33016811 on Build GRCh38
rs761126107 Chr.3: 33016775-33016775 on Build GRCh38
rs181973764 Chr.3: 33016653-33016653 on Build GRCh38
rs911845901 Chr.3: 33016442-33016442 on Build GRCh38
rs774732235 Chr.3: 33016847-33016847 on Build GRCh38
rs575202375 Chr.3: 33016882-33016882 on Build GRCh38
rs756154087 Chr.3: 33016725-33016725 on Build GRCh38
rs998138342 Chr.3: 33016647-33016647 on Build GRCh38
rs769139481 Chr.3: 33016812-33016812 on Build GRCh38
rs987510039 Chr.3: 33016441-33016441 on Build GRCh38
rs12054351 Chr.3: 33016505-33016505 on Build GRCh38

View Full Product Details