Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74480656-74481200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC142
Entrez Gene ID
84865
Gene Name
coiled-coil domain containing 142
Gene Aliases
-
UniGene
Hs.430199
Gene Chromosome Location
Chr.2: 74472832-74483230 on Build GRCh38
Transcripts
NM_032779.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4920326 c.1464G>A p.K488K Chr.2: 74480787-74480787 on Build GRCh38
3583370 c.1479C>T p.I493I Chr.2: 74480793-74480793 on Build GRCh38
4964260 c.1259-7G>A p.? Chr.2: 74481072-74481072 on Build GRCh38
4869379 c.1417T>C p.Y473H Chr.2: 74480855-74480855 on Build GRCh38
5430087 c.1474G>C p.E492Q Chr.2: 74480798-74480798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754575651 Chr.2: 74480941-74480941 on Build GRCh38
rs889586999 Chr.2: 74480702-74480702 on Build GRCh38
rs779593492 Chr.2: 74480796-74480796 on Build GRCh38
rs757038062 Chr.2: 74480749-74480749 on Build GRCh38
rs781518911 Chr.2: 74481097-74481097 on Build GRCh38
rs751478095 Chr.2: 74480745-74480745 on Build GRCh38
rs933670040 Chr.2: 74481108-74481108 on Build GRCh38
rs780078465 Chr.2: 74481125-74481125 on Build GRCh38
rs752034477 Chr.2: 74481082-74481082 on Build GRCh38
rs547820352 Chr.2: 74480710-74480710 on Build GRCh38
rs989454599 Chr.2: 74480741-74480741 on Build GRCh38
rs754488087 Chr.2: 74480793-74480793 on Build GRCh38
rs759258324 Chr.2: 74481062-74481062 on Build GRCh38
rs750098140 Chr.2: 74480776-74480776 on Build GRCh38
rs267599455 Chr.2: 74480838-74480838 on Build GRCh38
rs202065967 Chr.2: 74480997-74480997 on Build GRCh38
rs768577583 Chr.2: 74481178-74481178 on Build GRCh38
rs776697702 Chr.2: 74480729-74480729 on Build GRCh38
rs901332013 Chr.2: 74480679-74480679 on Build GRCh38
rs749406101 Chr.2: 74481128-74481128 on Build GRCh38
rs898523133 Chr.2: 74480696-74480696 on Build GRCh38
rs775631815 Chr.2: 74480999-74480999 on Build GRCh38
rs758949639 Chr.2: 74480955-74480955 on Build GRCh38
rs776787688 Chr.2: 74480861-74480861 on Build GRCh38
rs746151233 Chr.2: 74481118-74481118 on Build GRCh38
rs986941111 Chr.2: 74481146-74481146 on Build GRCh38
rs773241030 Chr.2: 74481180-74481180 on Build GRCh38
rs747424634 Chr.2: 74480980-74480980 on Build GRCh38
rs144617711 Chr.2: 74481073-74481073 on Build GRCh38
rs1023734785 Chr.2: 74480867-74480867 on Build GRCh38
rs369773742 Chr.2: 74480739-74480739 on Build GRCh38
rs770131464 Chr.2: 74481045-74481045 on Build GRCh38
rs769011108 Chr.2: 74481152-74481152 on Build GRCh38
rs539965559 Chr.2: 74481124-74481124 on Build GRCh38
rs928073391 Chr.2: 74480862-74480862 on Build GRCh38
rs3806608 Chr.2: 74481079-74481079 on Build GRCh38
rs753500232 Chr.2: 74480935-74480935 on Build GRCh38
rs771158655 Chr.2: 74480857-74480857 on Build GRCh38
rs766246005 Chr.2: 74481047-74481047 on Build GRCh38
rs1025618643 Chr.2: 74480742-74480742 on Build GRCh38
rs753212783 Chr.2: 74480948-74480948 on Build GRCh38
rs767279330 Chr.2: 74480898-74480898 on Build GRCh38
rs529276763 Chr.2: 74481025-74481025 on Build GRCh38
rs760486978 Chr.2: 74480918-74480918 on Build GRCh38
rs757671917 Chr.2: 74481084-74481084 on Build GRCh38
rs568441583 Chr.2: 74480855-74480855 on Build GRCh38
rs900275114 Chr.2: 74481066-74481066 on Build GRCh38
rs762872361 Chr.2: 74481020-74481020 on Build GRCh38
rs886113523 Chr.2: 74481001-74481001 on Build GRCh38
rs764085877 Chr.2: 74481089-74481089 on Build GRCh38
rs769838239 Chr.2: 74480882-74480882 on Build GRCh38
rs755783828 Chr.2: 74480783-74480783 on Build GRCh38
rs759556268 Chr.2: 74480732-74480732 on Build GRCh38
rs766150849 Chr.2: 74480919-74480919 on Build GRCh38
rs528470385 Chr.2: 74480711-74480711 on Build GRCh38
rs114473749 Chr.2: 74480810-74480810 on Build GRCh38
rs759647821 Chr.2: 74480865-74480865 on Build GRCh38
rs527566494 Chr.2: 74480728-74480728 on Build GRCh38
rs764696996 Chr.2: 74481080-74481080 on Build GRCh38
rs371687760 Chr.2: 74480888-74480888 on Build GRCh38
rs751663649 Chr.2: 74480777-74480778 on Build GRCh38
rs997055192 Chr.2: 74480686-74480686 on Build GRCh38
rs779784180 Chr.2: 74480788-74480788 on Build GRCh38
rs766635612 Chr.2: 74481115-74481115 on Build GRCh38
rs750234372 Chr.2: 74480915-74480915 on Build GRCh38
rs748666840 Chr.2: 74480806-74480806 on Build GRCh38
rs778197713 Chr.2: 74480826-74480826 on Build GRCh38
rs990637012 Chr.2: 74480932-74480932 on Build GRCh38
rs1033003879 Chr.2: 74480992-74480992 on Build GRCh38
rs753410207 Chr.2: 74480791-74480791 on Build GRCh38
rs778473573 Chr.2: 74480946-74480946 on Build GRCh38
rs376679728 Chr.2: 74480892-74480892 on Build GRCh38
rs867932667 Chr.2: 74480786-74480786 on Build GRCh38
rs930579320 Chr.2: 74480944-74480944 on Build GRCh38
rs757012417 Chr.2: 74481131-74481132 on Build GRCh38
rs774199883 Chr.2: 74480887-74480887 on Build GRCh38
rs772972873 Chr.2: 74481035-74481035 on Build GRCh38
rs778085168 Chr.2: 74480959-74480959 on Build GRCh38
rs771248932 Chr.2: 74480986-74480986 on Build GRCh38
rs551986523 Chr.2: 74481159-74481159 on Build GRCh38
rs1020974820 Chr.2: 74480747-74480747 on Build GRCh38
rs368091653 Chr.2: 74480763-74480763 on Build GRCh38
rs761964589 Chr.2: 74480995-74480995 on Build GRCh38
rs753602658 Chr.2: 74481060-74481060 on Build GRCh38
rs753774752 Chr.2: 74481105-74481108 on Build GRCh38
rs767317592 Chr.2: 74480772-74480772 on Build GRCh38
rs759727096 Chr.2: 74481001-74481013 on Build GRCh38
rs34186281 Chr.2: 74480886-74480887 on Build GRCh38

View Full Product Details