Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55543689-55544271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP36
Entrez Gene ID
112942
Gene Name
cilia and flagella associated protein 36
Gene Aliases
BARTL1, CCDC104
UniGene
Hs.264208
Gene Chromosome Location
Chr.2: 55519595-55545080 on Build GRCh38
Transcripts
NM_001282761.1 NM_080667.6 XM_011532499.1 XM_017003254.1 XM_017003255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4094910 c.810G>A p.R270R Chr.2: 55544252-55544252 on Build GRCh38
1021883 c.673G>T p.E225* Chr.2: 55543970-55543970 on Build GRCh38
1021886 c.808C>T p.R270W Chr.2: 55544250-55544250 on Build GRCh38
5058970 c.902T>C p.L301P Chr.2: 55544269-55544269 on Build GRCh38
4133876 c.787G>C p.V263L Chr.2: 55544229-55544229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753451365 Chr.2: 55544028-55544028 on Build GRCh38
rs775871264 Chr.2: 55544086-55544095 on Build GRCh38
rs547197074 Chr.2: 55543902-55543902 on Build GRCh38
rs756192692 Chr.2: 55544241-55544241 on Build GRCh38
rs1024039413 Chr.2: 55543863-55543873 on Build GRCh38
rs1041255322 Chr.2: 55544099-55544099 on Build GRCh38
rs747283308 Chr.2: 55544045-55544045 on Build GRCh38
rs368236290 Chr.2: 55543828-55543828 on Build GRCh38
rs536189267 Chr.2: 55543920-55543920 on Build GRCh38
rs770751548 Chr.2: 55544102-55544102 on Build GRCh38
rs542538922 Chr.2: 55544133-55544133 on Build GRCh38
rs772924174 Chr.2: 55544011-55544011 on Build GRCh38
rs776648413 Chr.2: 55543958-55543958 on Build GRCh38
rs777362685 Chr.2: 55544043-55544043 on Build GRCh38
rs200486714 Chr.2: 55543890-55543890 on Build GRCh38
rs750629388 Chr.2: 55544239-55544239 on Build GRCh38
rs964502539 Chr.2: 55543814-55543814 on Build GRCh38
rs759887331 Chr.2: 55543888-55543891 on Build GRCh38
rs377625368 Chr.2: 55543967-55543967 on Build GRCh38
rs749692801 Chr.2: 55544081-55544081 on Build GRCh38
rs3816489 Chr.2: 55543914-55543914 on Build GRCh38
rs547077963 Chr.2: 55543979-55543979 on Build GRCh38
rs775782530 Chr.2: 55543891-55543891 on Build GRCh38
rs962944203 Chr.2: 55544125-55544125 on Build GRCh38
rs373800467 Chr.2: 55544058-55544058 on Build GRCh38
rs767799026 Chr.2: 55544082-55544084 on Build GRCh38
rs761195645 Chr.2: 55544089-55544090 on Build GRCh38
rs1019679203 Chr.2: 55543759-55543759 on Build GRCh38
rs767700980 Chr.2: 55544139-55544139 on Build GRCh38
rs747999928 Chr.2: 55543926-55543926 on Build GRCh38
rs775943898 Chr.2: 55543976-55543976 on Build GRCh38
rs1039888706 Chr.2: 55543770-55543770 on Build GRCh38
rs781369745 Chr.2: 55544052-55544052 on Build GRCh38
rs373105185 Chr.2: 55543992-55543992 on Build GRCh38
rs201403227 Chr.2: 55544096-55544096 on Build GRCh38
rs758223598 Chr.2: 55544037-55544037 on Build GRCh38
rs752683322 Chr.2: 55544193-55544193 on Build GRCh38
rs758126750 Chr.2: 55543929-55543929 on Build GRCh38
rs373930835 Chr.2: 55543948-55543948 on Build GRCh38
rs199619720 Chr.2: 55543946-55543946 on Build GRCh38
rs375795436 Chr.2: 55543807-55543807 on Build GRCh38
rs539504950 Chr.2: 55543894-55543894 on Build GRCh38
rs562437727 Chr.2: 55544142-55544142 on Build GRCh38
rs763281596 Chr.2: 55543900-55543900 on Build GRCh38
rs769096648 Chr.2: 55544083-55544083 on Build GRCh38
rs766501054 Chr.2: 55544107-55544107 on Build GRCh38
rs373171383 Chr.2: 55544226-55544226 on Build GRCh38
rs770334893 Chr.2: 55543974-55543974 on Build GRCh38
rs757368168 Chr.2: 55544047-55544047 on Build GRCh38
rs762173970 Chr.2: 55544189-55544189 on Build GRCh38
rs570936043 Chr.2: 55543848-55543848 on Build GRCh38
rs199790709 Chr.2: 55544213-55544213 on Build GRCh38
rs780069678 Chr.2: 55544246-55544246 on Build GRCh38
rs530910274 Chr.2: 55543791-55543791 on Build GRCh38
rs1027438264 Chr.2: 55543838-55543838 on Build GRCh38
rs765936856 Chr.2: 55544027-55544027 on Build GRCh38
rs753859254 Chr.2: 55543915-55543915 on Build GRCh38
rs757045145 Chr.2: 55544217-55544217 on Build GRCh38
rs375585503 Chr.2: 55544194-55544194 on Build GRCh38
rs367730913 Chr.2: 55544070-55544070 on Build GRCh38
rs764111290 Chr.2: 55544092-55544093 on Build GRCh38
rs759790942 Chr.2: 55544138-55544138 on Build GRCh38
rs746502242 Chr.2: 55543808-55543810 on Build GRCh38
rs751355993 Chr.2: 55544216-55544216 on Build GRCh38
rs764785510 Chr.2: 55544178-55544178 on Build GRCh38
rs370286928 Chr.2: 55544036-55544036 on Build GRCh38
rs1045920 Chr.2: 55544033-55544033 on Build GRCh38
rs934050814 Chr.2: 55543792-55543792 on Build GRCh38
rs1008384008 Chr.2: 55544097-55544097 on Build GRCh38
rs186765766 Chr.2: 55544084-55544084 on Build GRCh38
rs747140574 Chr.2: 55543947-55543947 on Build GRCh38
rs754915018 Chr.2: 55543916-55543916 on Build GRCh38
rs750037305 Chr.2: 55543912-55543912 on Build GRCh38
rs183896217 Chr.2: 55544074-55544074 on Build GRCh38
rs753921733 Chr.2: 55544172-55544173 on Build GRCh38
rs1045910 Chr.2: 55544025-55544025 on Build GRCh38
rs1008418086 Chr.2: 55543749-55543749 on Build GRCh38
rs899946295 Chr.2: 55543782-55543782 on Build GRCh38
rs759177608 Chr.2: 55544109-55544109 on Build GRCh38
rs372024307 Chr.2: 55544230-55544230 on Build GRCh38
rs150639486 Chr.2: 55543985-55543985 on Build GRCh38
rs561778262 Chr.2: 55543771-55543771 on Build GRCh38
rs557427363 Chr.2: 55544053-55544053 on Build GRCh38

View Full Product Details