Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162142446-162142989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCG
Entrez Gene ID
2641
Gene Name
glucagon
Gene Aliases
GLP1, GLP2, GRPP
UniGene
Hs.516494
Gene Chromosome Location
Chr.2: 162142869-162152404 on Build GRCh38
Transcripts
NM_002054.4 NR_110255.1

Gene Symbol
LOC101929532
Entrez Gene ID
101929532
Gene Name
uncharacterized LOC101929532
Gene Aliases
-
UniGene
Hs.639883
Gene Chromosome Location
Chr.2: 162114441-162172739 on Build GRCh38
Transcripts
NM_002054.4 NR_110255.1

SNP Detail

SNP ID Chromosome Location
rs937653356 Chr.2: 162142892-162142892 on Build GRCh38
rs887669458 Chr.2: 162142796-162142796 on Build GRCh38
rs960431573 Chr.2: 162142738-162142738 on Build GRCh38
rs914120580 Chr.2: 162142633-162142633 on Build GRCh38
rs973640383 Chr.2: 162142839-162142839 on Build GRCh38
rs6717609 Chr.2: 162142753-162142753 on Build GRCh38
rs929143625 Chr.2: 162142949-162142949 on Build GRCh38
rs573251943 Chr.2: 162142515-162142515 on Build GRCh38
rs185729970 Chr.2: 162142669-162142669 on Build GRCh38
rs11552332 Chr.2: 162142918-162142918 on Build GRCh38
rs1051505868 Chr.2: 162142581-162142581 on Build GRCh38
rs982376894 Chr.2: 162142699-162142699 on Build GRCh38
rs112307942 Chr.2: 162142543-162142543 on Build GRCh38
rs866663677 Chr.2: 162142576-162142576 on Build GRCh38
rs912071397 Chr.2: 162142875-162142875 on Build GRCh38
rs1017335173 Chr.2: 162142824-162142824 on Build GRCh38
rs762032866 Chr.2: 162142580-162142580 on Build GRCh38
rs983896025 Chr.2: 162142519-162142519 on Build GRCh38
rs115679255 Chr.2: 162142757-162142757 on Build GRCh38
rs979103389 Chr.2: 162142884-162142884 on Build GRCh38
rs149201782 Chr.2: 162142862-162142862 on Build GRCh38
rs773583775 Chr.2: 162142932-162142932 on Build GRCh38
rs969211909 Chr.2: 162142478-162142478 on Build GRCh38
rs150023184 Chr.2: 162142637-162142637 on Build GRCh38
rs1005578255 Chr.2: 162142799-162142799 on Build GRCh38
rs16846320 Chr.2: 162142490-162142490 on Build GRCh38
rs562407074 Chr.2: 162142565-162142565 on Build GRCh38
rs946344419 Chr.2: 162142475-162142475 on Build GRCh38
rs1035775189 Chr.2: 162142719-162142719 on Build GRCh38
rs937752145 Chr.2: 162142657-162142657 on Build GRCh38
rs373440443 Chr.2: 162142525-162142525 on Build GRCh38
rs533941455 Chr.2: 162142876-162142876 on Build GRCh38
rs1003367897 Chr.2: 162142931-162142931 on Build GRCh38
rs796510940 Chr.2: 162142491-162142491 on Build GRCh38
rs964195336 Chr.2: 162142844-162142844 on Build GRCh38
rs745784860 Chr.2: 162142682-162142686 on Build GRCh38
rs544676361 Chr.2: 162142622-162142622 on Build GRCh38
rs893444035 Chr.2: 162142880-162142880 on Build GRCh38
rs549016429 Chr.2: 162142643-162142643 on Build GRCh38
rs544023074 Chr.2: 162142550-162142550 on Build GRCh38
rs183105157 Chr.2: 162142917-162142917 on Build GRCh38
rs6717263 Chr.2: 162142545-162142545 on Build GRCh38
rs548912528 Chr.2: 162142856-162142856 on Build GRCh38
rs896438621 Chr.2: 162142521-162142521 on Build GRCh38
rs917646890 Chr.2: 162142920-162142920 on Build GRCh38
rs946406218 Chr.2: 162142634-162142634 on Build GRCh38
rs527299083 Chr.2: 162142638-162142638 on Build GRCh38
rs1023519723 Chr.2: 162142647-162142647 on Build GRCh38
rs904919107 Chr.2: 162142495-162142495 on Build GRCh38
rs1035230366 Chr.2: 162142934-162142934 on Build GRCh38
rs533290005 Chr.2: 162142583-162142583 on Build GRCh38
rs6732914 Chr.2: 162142878-162142878 on Build GRCh38
rs767524765 Chr.2: 162142679-162142679 on Build GRCh38
rs751657570 Chr.2: 162142573-162142573 on Build GRCh38
rs145343576 Chr.2: 162142855-162142855 on Build GRCh38
rs955686533 Chr.2: 162142591-162142591 on Build GRCh38
rs34204135 Chr.2: 162142733-162142734 on Build GRCh38
rs540759713 Chr.2: 162142542-162142542 on Build GRCh38
rs775728274 Chr.2: 162142527-162142527 on Build GRCh38
rs558163510 Chr.2: 162142498-162142498 on Build GRCh38
rs969181080 Chr.2: 162142650-162142650 on Build GRCh38
rs890242779 Chr.2: 162142593-162142593 on Build GRCh38
rs6717659 Chr.2: 162142845-162142845 on Build GRCh38
rs1029705764 Chr.2: 162142851-162142851 on Build GRCh38
rs926281851 Chr.2: 162142891-162142891 on Build GRCh38
rs1047691370 Chr.2: 162142773-162142773 on Build GRCh38
rs763538781 Chr.2: 162142529-162142529 on Build GRCh38

View Full Product Details