Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54509585-54510059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGRRF1
Entrez Gene ID
10668
Gene Name
cell growth regulator with ring finger domain 1
Gene Aliases
CGR19, RNF197
UniGene
Hs.59106
Gene Chromosome Location
Chr.14: 54509869-54539310 on Build GRCh38
Transcripts
NM_006568.2 XM_017020931.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5626206 c.95T>C p.V32A Chr.14: 54510054-54510054 on Build GRCh38
3729997 c.90delC p.L31fs*12 Chr.14: 54510049-54510049 on Build GRCh38
4051329 c.11T>C p.V4A Chr.14: 54509970-54509970 on Build GRCh38
4822409 c.31G>A p.E11K Chr.14: 54509990-54509990 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs896075800 Chr.14: 54509893-54509893 on Build GRCh38
rs935212360 Chr.14: 54509858-54509858 on Build GRCh38
rs1013429975 Chr.14: 54509980-54509980 on Build GRCh38
rs992526812 Chr.14: 54509890-54509890 on Build GRCh38
rs1029062539 Chr.14: 54509919-54509919 on Build GRCh38
rs778639631 Chr.14: 54509881-54509881 on Build GRCh38
rs753614608 Chr.14: 54509871-54509871 on Build GRCh38
rs746553994 Chr.14: 54509947-54509947 on Build GRCh38
rs904862651 Chr.14: 54509748-54509748 on Build GRCh38
rs760523448 Chr.14: 54510023-54510023 on Build GRCh38
rs777662762 Chr.14: 54509945-54509945 on Build GRCh38
rs375625346 Chr.14: 54509688-54509688 on Build GRCh38
rs756820986 Chr.14: 54509928-54509928 on Build GRCh38
rs1051350443 Chr.14: 54509842-54509842 on Build GRCh38
rs763490879 Chr.14: 54509925-54509925 on Build GRCh38
rs764789300 Chr.14: 54509956-54509956 on Build GRCh38
rs749268741 Chr.14: 54509984-54509984 on Build GRCh38
rs185999192 Chr.14: 54509808-54509808 on Build GRCh38
rs771671316 Chr.14: 54509913-54509931 on Build GRCh38
rs769104000 Chr.14: 54509627-54509627 on Build GRCh38
rs987994498 Chr.14: 54509862-54509862 on Build GRCh38
rs527242448 Chr.14: 54509789-54509789 on Build GRCh38
rs1002267340 Chr.14: 54509878-54509878 on Build GRCh38
rs966584873 Chr.14: 54509895-54509895 on Build GRCh38
rs973481825 Chr.14: 54509798-54509798 on Build GRCh38
rs756535378 Chr.14: 54509975-54509975 on Build GRCh38
rs943075872 Chr.14: 54509805-54509805 on Build GRCh38
rs370019996 Chr.14: 54509947-54509947 on Build GRCh38
rs1002225389 Chr.14: 54509755-54509755 on Build GRCh38
rs764124020 Chr.14: 54509954-54509954 on Build GRCh38
rs921887920 Chr.14: 54509681-54509681 on Build GRCh38
rs199796624 Chr.14: 54509952-54509952 on Build GRCh38
rs75987656 Chr.14: 54509926-54509926 on Build GRCh38
rs2298077 Chr.14: 54509628-54509628 on Build GRCh38
rs759894263 Chr.14: 54509950-54509950 on Build GRCh38
rs188852541 Chr.14: 54509880-54509880 on Build GRCh38
rs764022726 Chr.14: 54509870-54509870 on Build GRCh38
rs760481707 Chr.14: 54509953-54509953 on Build GRCh38
rs549538047 Chr.14: 54509898-54509898 on Build GRCh38
rs966636659 Chr.14: 54509999-54509999 on Build GRCh38
rs963426852 Chr.14: 54509667-54509667 on Build GRCh38
rs900101210 Chr.14: 54509684-54509684 on Build GRCh38
rs925026237 Chr.14: 54509804-54509804 on Build GRCh38
rs981018000 Chr.14: 54509738-54509738 on Build GRCh38
rs370411135 Chr.14: 54510035-54510035 on Build GRCh38
rs770379085 Chr.14: 54509944-54509944 on Build GRCh38
rs557381217 Chr.14: 54509935-54509935 on Build GRCh38
rs778343224 Chr.14: 54509978-54509978 on Build GRCh38
rs371219880 Chr.14: 54509955-54509955 on Build GRCh38
rs370809387 Chr.14: 54509657-54509657 on Build GRCh38
rs1015138108 Chr.14: 54509897-54509897 on Build GRCh38
rs182354124 Chr.14: 54509709-54509709 on Build GRCh38
rs763060847 Chr.14: 54509917-54509917 on Build GRCh38
rs373074430 Chr.14: 54509957-54509957 on Build GRCh38
rs962735776 Chr.14: 54509733-54509733 on Build GRCh38
rs1033497122 Chr.14: 54509741-54509741 on Build GRCh38
rs746273080 Chr.14: 54509894-54509894 on Build GRCh38
rs374674383 Chr.14: 54509768-54509768 on Build GRCh38
rs377205659 Chr.14: 54509967-54509967 on Build GRCh38
rs535232652 Chr.14: 54509649-54509649 on Build GRCh38
rs1035929824 Chr.14: 54509767-54509767 on Build GRCh38
rs375843158 Chr.14: 54509910-54509910 on Build GRCh38
rs778904566 Chr.14: 54509890-54509891 on Build GRCh38
rs774244099 Chr.14: 54509920-54509920 on Build GRCh38
rs371030398 Chr.14: 54509682-54509682 on Build GRCh38
rs774726687 Chr.14: 54509949-54509949 on Build GRCh38
rs987817543 Chr.14: 54509830-54509830 on Build GRCh38
rs776621009 Chr.14: 54510038-54510038 on Build GRCh38
rs1043491465 Chr.14: 54509638-54509638 on Build GRCh38
rs960922811 Chr.14: 54509744-54509744 on Build GRCh38
rs913542341 Chr.14: 54509618-54509618 on Build GRCh38
rs774520077 Chr.14: 54509802-54509802 on Build GRCh38
rs1036602587 Chr.14: 54509976-54509976 on Build GRCh38
rs1056504164 Chr.14: 54509891-54509891 on Build GRCh38
rs1001740933 Chr.14: 54509605-54509605 on Build GRCh38
rs78067093 Chr.14: 54509865-54509865 on Build GRCh38
rs753411776 Chr.14: 54509927-54509927 on Build GRCh38
rs117211305 Chr.14: 54509951-54509951 on Build GRCh38
rs538453749 Chr.14: 54509911-54509911 on Build GRCh38
rs772447063 Chr.14: 54510009-54510009 on Build GRCh38
rs545401376 Chr.14: 54509680-54509680 on Build GRCh38
rs778546173 Chr.14: 54509932-54509932 on Build GRCh38
rs35335497 Chr.14: 54509806-54509807 on Build GRCh38
rs766642984 Chr.14: 54509918-54509918 on Build GRCh38
rs887147952 Chr.14: 54509852-54509852 on Build GRCh38
rs544346685 Chr.14: 54509792-54509792 on Build GRCh38
rs770051658 Chr.14: 54509686-54509686 on Build GRCh38
rs779199761 Chr.14: 54509988-54509988 on Build GRCh38
rs779736832 Chr.14: 54509927-54509927 on Build GRCh38
rs148749832 Chr.14: 54509647-54509647 on Build GRCh38
rs979107037 Chr.14: 54509905-54509905 on Build GRCh38
rs748480661 Chr.14: 54509662-54509662 on Build GRCh38
rs562994103 Chr.14: 54509793-54509793 on Build GRCh38
rs1016966887 Chr.14: 54509713-54509713 on Build GRCh38
rs372448071 Chr.14: 54509940-54509940 on Build GRCh38
rs934260328 Chr.14: 54509838-54509838 on Build GRCh38
rs181249929 Chr.14: 54509922-54509922 on Build GRCh38
rs200619234 Chr.14: 54509924-54509924 on Build GRCh38
rs973406680 Chr.14: 54509680-54509681 on Build GRCh38
rs574868197 Chr.14: 54509664-54509664 on Build GRCh38
rs149160670 Chr.14: 54509992-54509992 on Build GRCh38
rs748714368 Chr.14: 54509909-54509909 on Build GRCh38
rs781606695 Chr.14: 54509938-54509938 on Build GRCh38
rs771028922 Chr.14: 54509987-54509987 on Build GRCh38
rs142371935 Chr.14: 54509995-54509995 on Build GRCh38
rs1036305124 Chr.14: 54509608-54509608 on Build GRCh38
rs899984996 Chr.14: 54509837-54509837 on Build GRCh38
rs998419978 Chr.14: 54509685-54509685 on Build GRCh38
rs76634616 Chr.14: 54509872-54509872 on Build GRCh38
rs772218060 Chr.14: 54510012-54510012 on Build GRCh38

View Full Product Details