Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52864181-52864779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FERMT2
Entrez Gene ID
10979
Gene Name
fermitin family member 2
Gene Aliases
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
UniGene
Hs.509343
Gene Chromosome Location
Chr.14: 52857271-52951192 on Build GRCh38
Transcripts
NM_001134999.1 NM_001135000.1 NM_006832.2 XM_005267285.2 XM_006720008.2 XM_006720009.2 XM_006720010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6663140 c.1442C>T p.A481V Chr.14: 52864561-52864561 on Build GRCh38
1370121 c.1477C>A p.L493I Chr.14: 52864526-52864526 on Build GRCh38
7233840 c.1581A>G p.L527L Chr.14: 52864422-52864422 on Build GRCh38
5927712 c.1381G>A p.E461K Chr.14: 52864622-52864622 on Build GRCh38
6663136 c.1444G>T p.D482Y Chr.14: 52864559-52864559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753163184 Chr.14: 52864717-52864717 on Build GRCh38
rs767088052 Chr.14: 52864378-52864378 on Build GRCh38
rs749026267 Chr.14: 52864408-52864408 on Build GRCh38
rs748225738 Chr.14: 52864444-52864444 on Build GRCh38
rs745919726 Chr.14: 52864698-52864698 on Build GRCh38
rs540092121 Chr.14: 52864296-52864296 on Build GRCh38
rs747382966 Chr.14: 52864560-52864560 on Build GRCh38
rs563474878 Chr.14: 52864238-52864238 on Build GRCh38
rs753077242 Chr.14: 52864362-52864362 on Build GRCh38
rs754998020 Chr.14: 52864377-52864377 on Build GRCh38
rs375366343 Chr.14: 52864393-52864393 on Build GRCh38
rs372507518 Chr.14: 52864540-52864540 on Build GRCh38
rs763470096 Chr.14: 52864657-52864657 on Build GRCh38
rs766248457 Chr.14: 52864352-52864352 on Build GRCh38
rs756666589 Chr.14: 52864732-52864732 on Build GRCh38
rs944691171 Chr.14: 52864705-52864705 on Build GRCh38
rs373948864 Chr.14: 52864633-52864633 on Build GRCh38
rs149596230 Chr.14: 52864566-52864566 on Build GRCh38
rs113911542 Chr.14: 52864343-52864343 on Build GRCh38
rs773374416 Chr.14: 52864622-52864622 on Build GRCh38
rs551427383 Chr.14: 52864345-52864345 on Build GRCh38
rs774487752 Chr.14: 52864368-52864369 on Build GRCh38
rs760732426 Chr.14: 52864656-52864656 on Build GRCh38
rs185396458 Chr.14: 52864628-52864628 on Build GRCh38
rs756401065 Chr.14: 52864429-52864429 on Build GRCh38
rs777658527 Chr.14: 52864757-52864757 on Build GRCh38
rs981827598 Chr.14: 52864598-52864598 on Build GRCh38
rs979304030 Chr.14: 52864647-52864647 on Build GRCh38
rs777285253 Chr.14: 52864397-52864397 on Build GRCh38
rs773950457 Chr.14: 52864632-52864632 on Build GRCh38
rs761310814 Chr.14: 52864663-52864663 on Build GRCh38
rs993996144 Chr.14: 52864446-52864446 on Build GRCh38
rs758666219 Chr.14: 52864418-52864418 on Build GRCh38
rs750137441 Chr.14: 52864672-52864672 on Build GRCh38
rs916354600 Chr.14: 52864301-52864301 on Build GRCh38
rs529361720 Chr.14: 52864250-52864250 on Build GRCh38
rs752746667 Chr.14: 52864526-52864526 on Build GRCh38
rs567338870 Chr.14: 52864439-52864439 on Build GRCh38
rs367808160 Chr.14: 52864652-52864652 on Build GRCh38
rs747430958 Chr.14: 52864430-52864430 on Build GRCh38
rs201646952 Chr.14: 52864659-52864659 on Build GRCh38
rs771198553 Chr.14: 52864629-52864629 on Build GRCh38
rs752268255 Chr.14: 52864381-52864381 on Build GRCh38
rs79804050 Chr.14: 52864465-52864465 on Build GRCh38
rs552889764 Chr.14: 52864464-52864464 on Build GRCh38
rs759399416 Chr.14: 52864643-52864643 on Build GRCh38
rs754358628 Chr.14: 52864565-52864565 on Build GRCh38
rs992891489 Chr.14: 52864685-52864685 on Build GRCh38
rs759488950 Chr.14: 52864504-52864504 on Build GRCh38
rs917250009 Chr.14: 52864414-52864414 on Build GRCh38
rs952393008 Chr.14: 52864282-52864282 on Build GRCh38
rs767533080 Chr.14: 52864453-52864454 on Build GRCh38
rs751423329 Chr.14: 52864369-52864369 on Build GRCh38
rs535966031 Chr.14: 52864457-52864457 on Build GRCh38
rs981854307 Chr.14: 52864326-52864326 on Build GRCh38
rs767566349 Chr.14: 52864512-52864512 on Build GRCh38
rs755670343 Chr.14: 52864533-52864533 on Build GRCh38
rs769351114 Chr.14: 52864627-52864627 on Build GRCh38
rs759782955 Chr.14: 52864450-52864450 on Build GRCh38
rs369784505 Chr.14: 52864485-52864485 on Build GRCh38
rs1027118672 Chr.14: 52864257-52864257 on Build GRCh38
rs781580024 Chr.14: 52864708-52864708 on Build GRCh38
rs752796550 Chr.14: 52864738-52864738 on Build GRCh38
rs757757441 Chr.14: 52864581-52864581 on Build GRCh38
rs549331675 Chr.14: 52864307-52864307 on Build GRCh38
rs528347511 Chr.14: 52864344-52864344 on Build GRCh38
rs73300786 Chr.14: 52864654-52864654 on Build GRCh38
rs759146052 Chr.14: 52864294-52864294 on Build GRCh38
rs1025608690 Chr.14: 52864456-52864456 on Build GRCh38
rs537272975 Chr.14: 52864384-52864384 on Build GRCh38
rs111420315 Chr.14: 52864350-52864350 on Build GRCh38
rs918475864 Chr.14: 52864278-52864278 on Build GRCh38
rs940646305 Chr.14: 52864704-52864704 on Build GRCh38
rs765857313 Chr.14: 52864212-52864212 on Build GRCh38
rs896975039 Chr.14: 52864210-52864210 on Build GRCh38
rs200299119 Chr.14: 52864548-52864548 on Build GRCh38
rs776630065 Chr.14: 52864441-52864441 on Build GRCh38
rs755315837 Chr.14: 52864696-52864696 on Build GRCh38
rs748705901 Chr.14: 52864594-52864594 on Build GRCh38
rs36108575 Chr.14: 52864735-52864736 on Build GRCh38
rs780229537 Chr.14: 52864557-52864557 on Build GRCh38
rs753398844 Chr.14: 52864546-52864546 on Build GRCh38
rs112020006 Chr.14: 52864608-52864608 on Build GRCh38
rs983831218 Chr.14: 52864467-52864467 on Build GRCh38
rs771297715 Chr.14: 52864367-52864369 on Build GRCh38
rs758714686 Chr.14: 52864723-52864723 on Build GRCh38
rs571066363 Chr.14: 52864269-52864269 on Build GRCh38
rs763706765 Chr.14: 52864545-52864545 on Build GRCh38
rs937365347 Chr.14: 52864390-52864390 on Build GRCh38

View Full Product Details