Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35081088-35081548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM177A1
Entrez Gene ID
283635
Gene Name
family with sequence similarity 177 member A1
Gene Aliases
C14orf24
UniGene
Hs.446357
Gene Chromosome Location
Chr.14: 35044907-35083383 on Build GRCh38
Transcripts
NM_001079519.1 NM_001289022.1 NM_173607.3 NR_110415.1

Gene Symbol
LOC101927178
Entrez Gene ID
101927178
Gene Name
uncharacterized LOC101927178
Gene Aliases
-
UniGene
Hs.617139
Gene Chromosome Location
Chr.14: 35081081-35113130 on Build GRCh38
Transcripts
NM_001079519.1 NM_001289022.1 NM_173607.3 NR_110415.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
955486 c.557C>T p.S186L Chr.14: 35081143-35081143 on Build GRCh38
6463485 c.647A>T p.E216V Chr.14: 35081164-35081164 on Build GRCh38
5672600 c.639A>G p.P213P Chr.14: 35081225-35081225 on Build GRCh38
6463486 c.578A>T p.E193V Chr.14: 35081164-35081164 on Build GRCh38
6418125 c.509C>A p.T170N Chr.14: 35081095-35081095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759533088 Chr.14: 35081262-35081263 on Build GRCh38
rs913479731 Chr.14: 35081472-35081473 on Build GRCh38
rs758431524 Chr.14: 35081513-35081513 on Build GRCh38
rs143519021 Chr.14: 35081222-35081222 on Build GRCh38
rs190938140 Chr.14: 35081169-35081169 on Build GRCh38
rs537120671 Chr.14: 35081311-35081311 on Build GRCh38
rs562184528 Chr.14: 35081262-35081262 on Build GRCh38
rs763393585 Chr.14: 35081278-35081278 on Build GRCh38
rs148849349 Chr.14: 35081158-35081158 on Build GRCh38
rs866868313 Chr.14: 35081258-35081260 on Build GRCh38
rs750895223 Chr.14: 35081157-35081157 on Build GRCh38
rs770250955 Chr.14: 35081256-35081257 on Build GRCh38
rs760909986 Chr.14: 35081115-35081115 on Build GRCh38
rs754213355 Chr.14: 35081130-35081130 on Build GRCh38
rs995825002 Chr.14: 35081519-35081519 on Build GRCh38
rs1041020820 Chr.14: 35081490-35081490 on Build GRCh38
rs139163092 Chr.14: 35081295-35081295 on Build GRCh38
rs900165047 Chr.14: 35081207-35081207 on Build GRCh38
rs780897288 Chr.14: 35081195-35081195 on Build GRCh38
rs201051719 Chr.14: 35081120-35081120 on Build GRCh38
rs748738169 Chr.14: 35081257-35081262 on Build GRCh38
rs79788636 Chr.14: 35081494-35081494 on Build GRCh38
rs775649167 Chr.14: 35081237-35081237 on Build GRCh38
rs139237361 Chr.14: 35081259-35081260 on Build GRCh38
rs762132442 Chr.14: 35081263-35081263 on Build GRCh38
rs758078667 Chr.14: 35081182-35081185 on Build GRCh38
rs570496776 Chr.14: 35081340-35081340 on Build GRCh38
rs764566848 Chr.14: 35081118-35081118 on Build GRCh38
rs768278595 Chr.14: 35081257-35081261 on Build GRCh38
rs746515534 Chr.14: 35081212-35081213 on Build GRCh38
rs761782158 Chr.14: 35081273-35081274 on Build GRCh38
rs1024712714 Chr.14: 35081392-35081392 on Build GRCh38
rs778568602 Chr.14: 35081241-35081241 on Build GRCh38
rs1034015501 Chr.14: 35081522-35081522 on Build GRCh38
rs892631056 Chr.14: 35081335-35081335 on Build GRCh38
rs926018507 Chr.14: 35081379-35081379 on Build GRCh38
rs537801469 Chr.14: 35081318-35081318 on Build GRCh38
rs747936148 Chr.14: 35081219-35081219 on Build GRCh38
rs374710644 Chr.14: 35081181-35081181 on Build GRCh38
rs750047021 Chr.14: 35081168-35081171 on Build GRCh38
rs757634919 Chr.14: 35081138-35081138 on Build GRCh38
rs900195361 Chr.14: 35081516-35081516 on Build GRCh38
rs112571405 Chr.14: 35081480-35081480 on Build GRCh38
rs371353956 Chr.14: 35081261-35081261 on Build GRCh38
rs1053926403 Chr.14: 35081325-35081325 on Build GRCh38
rs768827114 Chr.14: 35081262-35081262 on Build GRCh38
rs552492253 Chr.14: 35081113-35081113 on Build GRCh38
rs762261746 Chr.14: 35081258-35081259 on Build GRCh38
rs970416892 Chr.14: 35081112-35081112 on Build GRCh38
rs754687338 Chr.14: 35081191-35081191 on Build GRCh38
rs926530030 Chr.14: 35081137-35081137 on Build GRCh38
rs746678300 Chr.14: 35081178-35081178 on Build GRCh38
rs923287654 Chr.14: 35081243-35081243 on Build GRCh38
rs753700968 Chr.14: 35081262-35081265 on Build GRCh38
rs749194215 Chr.14: 35081227-35081227 on Build GRCh38
rs779492665 Chr.14: 35081182-35081186 on Build GRCh38
rs369006342 Chr.14: 35081151-35081151 on Build GRCh38
rs144052189 Chr.14: 35081353-35081353 on Build GRCh38
rs936188753 Chr.14: 35081414-35081414 on Build GRCh38
rs184025379 Chr.14: 35081322-35081322 on Build GRCh38
rs945018197 Chr.14: 35081476-35081476 on Build GRCh38
rs573408624 Chr.14: 35081515-35081515 on Build GRCh38
rs1004694915 Chr.14: 35081258-35081261 on Build GRCh38
rs769611643 Chr.14: 35081221-35081221 on Build GRCh38
rs757688937 Chr.14: 35081259-35081261 on Build GRCh38
rs199773964 Chr.14: 35081265-35081265 on Build GRCh38
rs750899979 Chr.14: 35081266-35081266 on Build GRCh38
rs1053183416 Chr.14: 35081460-35081460 on Build GRCh38
rs772278689 Chr.14: 35081235-35081235 on Build GRCh38
rs763215146 Chr.14: 35081261-35081261 on Build GRCh38
rs781535637 Chr.14: 35081257-35081260 on Build GRCh38
rs373465295 Chr.14: 35081261-35081262 on Build GRCh38

View Full Product Details