Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96451212-96451773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK7
Entrez Gene ID
122481
Gene Name
adenylate kinase 7
Gene Aliases
AK 7
UniGene
Hs.667462
Gene Chromosome Location
Chr.14: 96392111-96489427 on Build GRCh38
Transcripts
NM_152327.3 XM_006720021.2 XM_006720022.2 XM_011536403.2 XM_017020956.1 XM_017020957.1 XM_017020958.1 XM_017020959.1 XM_017020960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4796112 c.1085G>A p.S362N Chr.14: 96451557-96451557 on Build GRCh38
3987860 c.1022G>A p.R341Q Chr.14: 96451494-96451494 on Build GRCh38
2151877 c.997T>C p.F333L Chr.14: 96451469-96451469 on Build GRCh38
5681641 c.1031delC p.Q345fs*21 Chr.14: 96451503-96451503 on Build GRCh38
2199746 c.977A>G p.N326S Chr.14: 96451449-96451449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018902631 Chr.14: 96451255-96451255 on Build GRCh38
rs371692761 Chr.14: 96451393-96451393 on Build GRCh38
rs753912709 Chr.14: 96451481-96451481 on Build GRCh38
rs965053179 Chr.14: 96451257-96451257 on Build GRCh38
rs773562419 Chr.14: 96451576-96451576 on Build GRCh38
rs974800524 Chr.14: 96451275-96451275 on Build GRCh38
rs547973231 Chr.14: 96451327-96451327 on Build GRCh38
rs760992535 Chr.14: 96451582-96451582 on Build GRCh38
rs762152227 Chr.14: 96451232-96451232 on Build GRCh38
rs201349446 Chr.14: 96451543-96451543 on Build GRCh38
rs138075603 Chr.14: 96451566-96451566 on Build GRCh38
rs780991483 Chr.14: 96451512-96451512 on Build GRCh38
rs369963603 Chr.14: 96451427-96451427 on Build GRCh38
rs114559730 Chr.14: 96451309-96451309 on Build GRCh38
rs958625923 Chr.14: 96451281-96451281 on Build GRCh38
rs766696940 Chr.14: 96451609-96451609 on Build GRCh38
rs140034560 Chr.14: 96451550-96451550 on Build GRCh38
rs755693527 Chr.14: 96451530-96451530 on Build GRCh38
rs200892047 Chr.14: 96451482-96451482 on Build GRCh38
rs555152681 Chr.14: 96451670-96451670 on Build GRCh38
rs1002677923 Chr.14: 96451668-96451668 on Build GRCh38
rs374976503 Chr.14: 96451382-96451382 on Build GRCh38
rs143172089 Chr.14: 96451428-96451428 on Build GRCh38
rs758345212 Chr.14: 96451371-96451371 on Build GRCh38
rs114724004 Chr.14: 96451465-96451465 on Build GRCh38
rs764749876 Chr.14: 96451383-96451383 on Build GRCh38
rs907029626 Chr.14: 96451233-96451233 on Build GRCh38
rs756978825 Chr.14: 96451505-96451505 on Build GRCh38
rs780486577 Chr.14: 96451716-96451716 on Build GRCh38
rs140230412 Chr.14: 96451534-96451534 on Build GRCh38
rs972297310 Chr.14: 96451344-96451345 on Build GRCh38
rs764174660 Chr.14: 96451621-96451621 on Build GRCh38
rs758470168 Chr.14: 96451488-96451488 on Build GRCh38
rs766317292 Chr.14: 96451724-96451724 on Build GRCh38
rs748644130 Chr.14: 96451561-96451561 on Build GRCh38
rs571584215 Chr.14: 96451518-96451518 on Build GRCh38
rs772490713 Chr.14: 96451387-96451387 on Build GRCh38
rs369458683 Chr.14: 96451540-96451540 on Build GRCh38
rs1008813923 Chr.14: 96451254-96451254 on Build GRCh38
rs774833713 Chr.14: 96451441-96451441 on Build GRCh38
rs185565195 Chr.14: 96451579-96451579 on Build GRCh38
rs996515637 Chr.14: 96451739-96451739 on Build GRCh38
rs760914649 Chr.14: 96451469-96451469 on Build GRCh38
rs776917841 Chr.14: 96451617-96451617 on Build GRCh38
rs17853406 Chr.14: 96451453-96451453 on Build GRCh38
rs902347352 Chr.14: 96451565-96451565 on Build GRCh38
rs373425925 Chr.14: 96451464-96451464 on Build GRCh38
rs1034518581 Chr.14: 96451749-96451749 on Build GRCh38
rs1027621049 Chr.14: 96451742-96451742 on Build GRCh38
rs1019266793 Chr.14: 96451667-96451667 on Build GRCh38
rs945994072 Chr.14: 96451568-96451568 on Build GRCh38
rs769091755 Chr.14: 96451439-96451439 on Build GRCh38
rs552064132 Chr.14: 96451467-96451467 on Build GRCh38
rs777849703 Chr.14: 96451493-96451493 on Build GRCh38
rs773352505 Chr.14: 96451494-96451494 on Build GRCh38
rs745431750 Chr.14: 96451411-96451411 on Build GRCh38
rs770685461 Chr.14: 96451406-96451406 on Build GRCh38
rs774939631 Chr.14: 96451422-96451422 on Build GRCh38
rs749897626 Chr.14: 96451409-96451412 on Build GRCh38
rs1008759983 Chr.14: 96451645-96451645 on Build GRCh38
rs146653383 Chr.14: 96451472-96451472 on Build GRCh38
rs759658675 Chr.14: 96451619-96451619 on Build GRCh38
rs952123750 Chr.14: 96451310-96451310 on Build GRCh38
rs750637067 Chr.14: 96451468-96451468 on Build GRCh38
rs536787025 Chr.14: 96451644-96451644 on Build GRCh38
rs780676422 Chr.14: 96451410-96451410 on Build GRCh38
rs573484354 Chr.14: 96451719-96451719 on Build GRCh38
rs983506480 Chr.14: 96451319-96451319 on Build GRCh38
rs200320116 Chr.14: 96451421-96451421 on Build GRCh38

View Full Product Details