Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90407550-90408106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM1
Entrez Gene ID
801
Gene Name
calmodulin 1
Gene Aliases
CALML2, CAMI, CPVT4, DD132, LQT14, PHKD, caM
UniGene
Hs.282410
Gene Chromosome Location
Chr.14: 90396983-90408275 on Build GRCh38
Transcripts
NM_006888.4 XM_006720258.3

SNP Detail

SNP ID Chromosome Location
rs931533499 Chr.14: 90407931-90407931 on Build GRCh38
rs1052534481 Chr.14: 90407698-90407698 on Build GRCh38
rs969039684 Chr.14: 90408027-90408027 on Build GRCh38
rs899828474 Chr.14: 90407786-90407786 on Build GRCh38
rs943030494 Chr.14: 90407951-90407951 on Build GRCh38
rs759855927 Chr.14: 90407860-90407860 on Build GRCh38
rs560809535 Chr.14: 90407944-90407945 on Build GRCh38
rs769566588 Chr.14: 90407756-90407756 on Build GRCh38
rs142500788 Chr.14: 90407994-90407994 on Build GRCh38
rs1020893060 Chr.14: 90407589-90407589 on Build GRCh38
rs150072386 Chr.14: 90407575-90407578 on Build GRCh38
rs918859114 Chr.14: 90407840-90407840 on Build GRCh38
rs140025082 Chr.14: 90407664-90407664 on Build GRCh38
rs1005081735 Chr.14: 90407980-90407980 on Build GRCh38
rs916845728 Chr.14: 90407642-90407642 on Build GRCh38
rs982752644 Chr.14: 90407654-90407654 on Build GRCh38
rs562541013 Chr.14: 90407936-90407936 on Build GRCh38
rs967013904 Chr.14: 90407628-90407628 on Build GRCh38
rs902433074 Chr.14: 90408051-90408051 on Build GRCh38
rs144946670 Chr.14: 90408000-90408000 on Build GRCh38
rs973040050 Chr.14: 90407625-90407625 on Build GRCh38
rs887409660 Chr.14: 90407937-90407937 on Build GRCh38
rs371784290 Chr.14: 90408025-90408025 on Build GRCh38
rs963041554 Chr.14: 90407618-90407618 on Build GRCh38
rs370549807 Chr.14: 90407984-90407984 on Build GRCh38
rs578140742 Chr.14: 90407770-90407770 on Build GRCh38
rs973298333 Chr.14: 90407796-90407796 on Build GRCh38
rs760223179 Chr.14: 90407665-90407665 on Build GRCh38
rs115172487 Chr.14: 90407697-90407697 on Build GRCh38
rs979467641 Chr.14: 90407652-90407652 on Build GRCh38
rs776396739 Chr.14: 90407748-90407748 on Build GRCh38
rs963093954 Chr.14: 90407991-90407991 on Build GRCh38
rs997755007 Chr.14: 90407574-90407574 on Build GRCh38
rs147728175 Chr.14: 90407935-90407935 on Build GRCh38
rs1021175806 Chr.14: 90408017-90408017 on Build GRCh38
rs558383914 Chr.14: 90407896-90407896 on Build GRCh38
rs918004638 Chr.14: 90407757-90407757 on Build GRCh38
rs893869481 Chr.14: 90407724-90407724 on Build GRCh38
rs1015052373 Chr.14: 90407615-90407615 on Build GRCh38
rs753204876 Chr.14: 90408049-90408049 on Build GRCh38
rs949603622 Chr.14: 90407787-90407787 on Build GRCh38
rs145426711 Chr.14: 90407805-90407805 on Build GRCh38

View Full Product Details