Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95444324-95444856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE3
Entrez Gene ID
161176
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, NET53, Nesp3
UniGene
Hs.354645
Gene Chromosome Location
Chr.14: 95416083-95516693 on Build GRCh38
Transcripts
NM_152592.3 XM_005267377.3 XM_006720063.3 XM_011536515.2 XM_017021056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2253926 c.1738G>A p.D580N Chr.14: 95444523-95444523 on Build GRCh38
6794979 c.1699C>T p.L567F Chr.14: 95444562-95444562 on Build GRCh38
6794978 c.1699C>T p.L567F Chr.14: 95444562-95444562 on Build GRCh38
3936688 c.1727G>T p.G576V Chr.14: 95444534-95444534 on Build GRCh38
1493335 c.1659T>C p.F553F Chr.14: 95444602-95444602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780775179 Chr.14: 95444497-95444497 on Build GRCh38
rs760945215 Chr.14: 95444542-95444542 on Build GRCh38
rs564251914 Chr.14: 95444632-95444633 on Build GRCh38
rs556716725 Chr.14: 95444551-95444551 on Build GRCh38
rs9919887 Chr.14: 95444671-95444671 on Build GRCh38
rs374579596 Chr.14: 95444380-95444380 on Build GRCh38
rs748639681 Chr.14: 95444622-95444622 on Build GRCh38
rs992076816 Chr.14: 95444694-95444694 on Build GRCh38
rs768986406 Chr.14: 95444525-95444525 on Build GRCh38
rs913252205 Chr.14: 95444629-95444629 on Build GRCh38
rs978772064 Chr.14: 95444728-95444728 on Build GRCh38
rs533853262 Chr.14: 95444795-95444795 on Build GRCh38
rs373058185 Chr.14: 95444646-95444646 on Build GRCh38
rs60135668 Chr.14: 95444636-95444637 on Build GRCh38
rs912001410 Chr.14: 95444351-95444351 on Build GRCh38
rs749572025 Chr.14: 95444423-95444423 on Build GRCh38
rs372934775 Chr.14: 95444664-95444664 on Build GRCh38
rs554681012 Chr.14: 95444754-95444754 on Build GRCh38
rs901650930 Chr.14: 95444816-95444816 on Build GRCh38
rs571392982 Chr.14: 95444699-95444699 on Build GRCh38
rs776115855 Chr.14: 95444425-95444425 on Build GRCh38
rs1020408001 Chr.14: 95444829-95444829 on Build GRCh38
rs762139064 Chr.14: 95444444-95444444 on Build GRCh38
rs762955782 Chr.14: 95444566-95444566 on Build GRCh38
rs117511324 Chr.14: 95444724-95444724 on Build GRCh38
rs769129146 Chr.14: 95444435-95444435 on Build GRCh38
rs565608397 Chr.14: 95444597-95444597 on Build GRCh38
rs141984619 Chr.14: 95444487-95444487 on Build GRCh38
rs749710766 Chr.14: 95444384-95444384 on Build GRCh38
rs924232633 Chr.14: 95444732-95444732 on Build GRCh38
rs374250791 Chr.14: 95444450-95444450 on Build GRCh38
rs538551328 Chr.14: 95444719-95444719 on Build GRCh38
rs535952387 Chr.14: 95444507-95444507 on Build GRCh38
rs1056807277 Chr.14: 95444750-95444750 on Build GRCh38
rs1019196758 Chr.14: 95444806-95444806 on Build GRCh38
rs998281490 Chr.14: 95444824-95444824 on Build GRCh38
rs202009724 Chr.14: 95444636-95444636 on Build GRCh38
rs774782727 Chr.14: 95444526-95444526 on Build GRCh38
rs755662883 Chr.14: 95444505-95444505 on Build GRCh38
rs773614686 Chr.14: 95444541-95444541 on Build GRCh38
rs528322868 Chr.14: 95444679-95444679 on Build GRCh38
rs763902909 Chr.14: 95444583-95444583 on Build GRCh38
rs1048719609 Chr.14: 95444757-95444757 on Build GRCh38
rs9919918 Chr.14: 95444596-95444596 on Build GRCh38
rs202075336 Chr.14: 95444632-95444632 on Build GRCh38
rs192836578 Chr.14: 95444747-95444747 on Build GRCh38
rs776807420 Chr.14: 95444648-95444648 on Build GRCh38
rs779628602 Chr.14: 95444601-95444601 on Build GRCh38
rs531497600 Chr.14: 95444697-95444697 on Build GRCh38
rs779424527 Chr.14: 95444506-95444506 on Build GRCh38
rs773453052 Chr.14: 95444451-95444451 on Build GRCh38
rs759490079 Chr.14: 95444650-95444650 on Build GRCh38
rs8007302 Chr.14: 95444800-95444800 on Build GRCh38
rs995192063 Chr.14: 95444660-95444660 on Build GRCh38
rs150245374 Chr.14: 95444586-95444586 on Build GRCh38
rs758257602 Chr.14: 95444476-95444476 on Build GRCh38
rs772601950 Chr.14: 95444628-95444628 on Build GRCh38
rs752718635 Chr.14: 95444469-95444469 on Build GRCh38
rs144257709 Chr.14: 95444546-95444546 on Build GRCh38
rs766623957 Chr.14: 95444746-95444746 on Build GRCh38
rs974823687 Chr.14: 95444718-95444718 on Build GRCh38
rs748678129 Chr.14: 95444616-95444616 on Build GRCh38
rs754491640 Chr.14: 95444672-95444672 on Build GRCh38
rs200153210 Chr.14: 95444619-95444619 on Build GRCh38
rs370896209 Chr.14: 95444482-95444482 on Build GRCh38
rs763339517 Chr.14: 95444428-95444428 on Build GRCh38
rs766541476 Chr.14: 95444457-95444457 on Build GRCh38
rs987984176 Chr.14: 95444350-95444350 on Build GRCh38
rs36019358 Chr.14: 95444633-95444634 on Build GRCh38
rs8007261 Chr.14: 95444723-95444723 on Build GRCh38
rs774564608 Chr.14: 95444442-95444442 on Build GRCh38
rs771280899 Chr.14: 95444427-95444435 on Build GRCh38
rs374360574 Chr.14: 95444536-95444536 on Build GRCh38
rs760783647 Chr.14: 95444456-95444456 on Build GRCh38
rs562878238 Chr.14: 95444634-95444634 on Build GRCh38
rs1044112613 Chr.14: 95444793-95444793 on Build GRCh38
rs772347885 Chr.14: 95444590-95444590 on Build GRCh38
rs546491661 Chr.14: 95444680-95444680 on Build GRCh38
rs368339780 Chr.14: 95444652-95444652 on Build GRCh38
rs8007583 Chr.14: 95444786-95444786 on Build GRCh38
rs770308640 Chr.14: 95444422-95444422 on Build GRCh38
rs776518247 Chr.14: 95444549-95444549 on Build GRCh38
rs746077578 Chr.14: 95444526-95444527 on Build GRCh38
rs751249792 Chr.14: 95444484-95444484 on Build GRCh38
rs761736864 Chr.14: 95444662-95444662 on Build GRCh38
rs767355621 Chr.14: 95444663-95444663 on Build GRCh38
rs530103915 Chr.14: 95444642-95444642 on Build GRCh38
rs964794381 Chr.14: 95444705-95444705 on Build GRCh38
rs745499291 Chr.14: 95444503-95444503 on Build GRCh38
rs779216319 Chr.14: 95444476-95444477 on Build GRCh38
rs769039629 Chr.14: 95444409-95444409 on Build GRCh38
rs897472998 Chr.14: 95444534-95444534 on Build GRCh38
rs747251468 Chr.14: 95444640-95444640 on Build GRCh38
rs779234406 Chr.14: 95444620-95444620 on Build GRCh38

View Full Product Details