Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43793506-43794068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR1282
Entrez Gene ID
100302254
Gene Name
microRNA 1282
Gene Aliases
MIRN1282, hsa-mir-1282
UniGene
Gene Chromosome Location
Chr.15: 43793659-43793759 on Build GRCh38
Transcripts
NR_031695.1 NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1

Gene Symbol
SERF2
Entrez Gene ID
10169
Gene Name
small EDRK-rich factor 2
Gene Aliases
4F5REL, FAM2C, H4F5REL, HsT17089
UniGene
Hs.424126
Gene Chromosome Location
Chr.15: 43777096-43796089 on Build GRCh38
Transcripts
NR_031695.1 NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1

Gene Symbol
SERF2-C15ORF63
Entrez Gene ID
100529067
Gene Name
SERF2-C15orf63 readthrough
Gene Aliases
-
UniGene
Hs.730672
Gene Chromosome Location
Chr.15: 43791976-43802589 on Build GRCh38
Transcripts
NR_031695.1 NM_001018108.3 NM_001199875.1 NM_001199876.1 NM_001199877.1 NM_001199878.1 NR_037672.1 NR_037673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
433934 c.117-10C>G p.? Chr.15: 43793700-43793700 on Build GRCh38
3981516 c.256C>A p.R86R Chr.15: 43793715-43793715 on Build GRCh38
3981517 c.122C>A p.S41* Chr.15: 43793715-43793715 on Build GRCh38
3370180 c.288A>T p.Q96H Chr.15: 43793747-43793747 on Build GRCh38
3370182 c.295A>T p.R99* Chr.15: 43793754-43793754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538690173 Chr.15: 43793656-43793656 on Build GRCh38
rs750512118 Chr.15: 43793818-43793818 on Build GRCh38
rs751950090 Chr.15: 43793544-43793544 on Build GRCh38
rs764238650 Chr.15: 43793809-43793809 on Build GRCh38
rs928643399 Chr.15: 43794009-43794009 on Build GRCh38
rs775913330 Chr.15: 43793628-43793629 on Build GRCh38
rs778487274 Chr.15: 43793901-43793901 on Build GRCh38
rs770557996 Chr.15: 43793830-43793830 on Build GRCh38
rs764915359 Chr.15: 43793828-43793829 on Build GRCh38
rs367904215 Chr.15: 43793661-43793661 on Build GRCh38
rs749357108 Chr.15: 43793569-43793569 on Build GRCh38
rs768037613 Chr.15: 43793803-43793803 on Build GRCh38
rs879249287 Chr.15: 43793686-43793686 on Build GRCh38
rs750114075 Chr.15: 43793922-43793925 on Build GRCh38
rs11269 Chr.15: 43793711-43793711 on Build GRCh38
rs963310915 Chr.15: 43794016-43794016 on Build GRCh38
rs553559156 Chr.15: 43793619-43793619 on Build GRCh38
rs764583848 Chr.15: 43793850-43793850 on Build GRCh38
rs575152814 Chr.15: 43793811-43793811 on Build GRCh38
rs769017432 Chr.15: 43793923-43793923 on Build GRCh38
rs771499128 Chr.15: 43793848-43793848 on Build GRCh38
rs11555372 Chr.15: 43793817-43793817 on Build GRCh38
rs772478588 Chr.15: 43793836-43793836 on Build GRCh38
rs573381677 Chr.15: 43793628-43793628 on Build GRCh38
rs766290998 Chr.15: 43793994-43793995 on Build GRCh38
rs769462084 Chr.15: 43793618-43793618 on Build GRCh38
rs755840262 Chr.15: 43793568-43793568 on Build GRCh38
rs574350732 Chr.15: 43793941-43793941 on Build GRCh38
rs754908367 Chr.15: 43793821-43793821 on Build GRCh38
rs747914951 Chr.15: 43793823-43793823 on Build GRCh38
rs761170966 Chr.15: 43793702-43793702 on Build GRCh38
rs746226563 Chr.15: 43793528-43793529 on Build GRCh38
rs772869968 Chr.15: 43793783-43793783 on Build GRCh38
rs757746110 Chr.15: 43793877-43793877 on Build GRCh38
rs765520151 Chr.15: 43793854-43793854 on Build GRCh38
rs776485256 Chr.15: 43793603-43793603 on Build GRCh38
rs777347701 Chr.15: 43793587-43793587 on Build GRCh38
rs771542150 Chr.15: 43793907-43793907 on Build GRCh38
rs756987277 Chr.15: 43793964-43793964 on Build GRCh38
rs769358781 Chr.15: 43793778-43793778 on Build GRCh38
rs777603295 Chr.15: 43793894-43793894 on Build GRCh38
rs752003521 Chr.15: 43793638-43793638 on Build GRCh38
rs762171807 Chr.15: 43793853-43793853 on Build GRCh38
rs749061524 Chr.15: 43793895-43793895 on Build GRCh38
rs770359655 Chr.15: 43793787-43793787 on Build GRCh38
rs3180687 Chr.15: 43794046-43794046 on Build GRCh38
rs1802422 Chr.15: 43793838-43793838 on Build GRCh38
rs752794177 Chr.15: 43793561-43793561 on Build GRCh38
rs543848193 Chr.15: 43793857-43793857 on Build GRCh38
rs937562616 Chr.15: 43794011-43794011 on Build GRCh38
rs772634038 Chr.15: 43793625-43793625 on Build GRCh38
rs532242547 Chr.15: 43793959-43793959 on Build GRCh38
rs1012412136 Chr.15: 43793973-43793973 on Build GRCh38
rs753092831 Chr.15: 43793653-43793653 on Build GRCh38
rs759897421 Chr.15: 43793639-43793639 on Build GRCh38
rs773900330 Chr.15: 43793833-43793833 on Build GRCh38
rs372561860 Chr.15: 43793689-43793689 on Build GRCh38
rs760093532 Chr.15: 43793794-43793794 on Build GRCh38
rs762406090 Chr.15: 43793629-43793629 on Build GRCh38
rs758948985 Chr.15: 43793532-43793532 on Build GRCh38
rs773845378 Chr.15: 43793793-43793793 on Build GRCh38
rs780903077 Chr.15: 43793695-43793695 on Build GRCh38
rs755284896 Chr.15: 43793552-43793552 on Build GRCh38
rs753510458 Chr.15: 43793807-43793807 on Build GRCh38
rs772482463 Chr.15: 43793619-43793620 on Build GRCh38
rs756331229 Chr.15: 43793658-43793658 on Build GRCh38
rs749008549 Chr.15: 43793826-43793826 on Build GRCh38
rs776224437 Chr.15: 43793786-43793787 on Build GRCh38
rs757357108 Chr.15: 43793680-43793680 on Build GRCh38
rs918571408 Chr.15: 43793621-43793621 on Build GRCh38
rs758458926 Chr.15: 43793869-43793869 on Build GRCh38
rs551831965 Chr.15: 43793977-43793977 on Build GRCh38
rs11555370 Chr.15: 43793834-43793834 on Build GRCh38
rs369832525 Chr.15: 43793684-43793684 on Build GRCh38
rs745342456 Chr.15: 43793835-43793835 on Build GRCh38
rs1030020778 Chr.15: 43793560-43793560 on Build GRCh38
rs558401790 Chr.15: 43793670-43793670 on Build GRCh38
rs781404474 Chr.15: 43793553-43793553 on Build GRCh38
rs3180486 Chr.15: 43794045-43794045 on Build GRCh38
rs112115773 Chr.15: 43793820-43793820 on Build GRCh38
rs565283773 Chr.15: 43793990-43793990 on Build GRCh38
rs560265406 Chr.15: 43793958-43793958 on Build GRCh38
rs761280384 Chr.15: 43793814-43793815 on Build GRCh38
rs981889401 Chr.15: 43793987-43793987 on Build GRCh38
rs3180684 Chr.15: 43794044-43794044 on Build GRCh38
rs772168214 Chr.15: 43793574-43793574 on Build GRCh38
rs1041792142 Chr.15: 43793921-43793921 on Build GRCh38
rs766836436 Chr.15: 43793539-43793539 on Build GRCh38
rs1045969535 Chr.15: 43793640-43793640 on Build GRCh38
rs757971426 Chr.15: 43793998-43793998 on Build GRCh38
rs369902877 Chr.15: 43793813-43793813 on Build GRCh38
rs11555376 Chr.15: 43793886-43793886 on Build GRCh38
rs760950472 Chr.15: 43793841-43793841 on Build GRCh38
rs1049531 Chr.15: 43793843-43793843 on Build GRCh38
rs540137813 Chr.15: 43793949-43793949 on Build GRCh38
rs1034061455 Chr.15: 43793951-43793951 on Build GRCh38
rs745381980 Chr.15: 43793903-43793903 on Build GRCh38
rs760717391 Chr.15: 43793957-43793957 on Build GRCh38
rs763552438 Chr.15: 43793751-43793753 on Build GRCh38
rs768501330 Chr.15: 43793592-43793592 on Build GRCh38
rs756355019 Chr.15: 43793566-43793566 on Build GRCh38
rs747940819 Chr.15: 43793767-43793767 on Build GRCh38
rs145178206 Chr.15: 43793583-43793583 on Build GRCh38
rs201274196 Chr.15: 43793786-43793786 on Build GRCh38
rs375164328 Chr.15: 43793647-43793647 on Build GRCh38
rs753655469 Chr.15: 43793962-43793962 on Build GRCh38
rs766891421 Chr.15: 43793631-43793631 on Build GRCh38
rs778540129 Chr.15: 43793965-43793965 on Build GRCh38
rs11555369 Chr.15: 43793871-43793871 on Build GRCh38
rs747606015 Chr.15: 43793914-43793914 on Build GRCh38
rs756934168 Chr.15: 43793899-43793899 on Build GRCh38
rs776005780 Chr.15: 43793840-43793840 on Build GRCh38
rs750154803 Chr.15: 43793991-43793991 on Build GRCh38
rs762233187 Chr.15: 43793930-43793930 on Build GRCh38
rs753035270 Chr.15: 43793804-43793804 on Build GRCh38
rs761435279 Chr.15: 43793610-43793610 on Build GRCh38
rs1043887061 Chr.15: 43793819-43793819 on Build GRCh38
rs756003408 Chr.15: 43793893-43793893 on Build GRCh38
rs777171460 Chr.15: 43793929-43793929 on Build GRCh38
rs758259097 Chr.15: 43793953-43793953 on Build GRCh38
rs755878719 Chr.15: 43793824-43793824 on Build GRCh38
rs538292919 Chr.15: 43794007-43794008 on Build GRCh38
rs962753506 Chr.15: 43793611-43793611 on Build GRCh38
rs766549249 Chr.15: 43793947-43793947 on Build GRCh38
rs560839312 Chr.15: 43793888-43793888 on Build GRCh38
rs773514291 Chr.15: 43793946-43793946 on Build GRCh38
rs768705094 Chr.15: 43793690-43793690 on Build GRCh38
rs1025249440 Chr.15: 43793999-43793999 on Build GRCh38
rs765474877 Chr.15: 43793815-43793815 on Build GRCh38

View Full Product Details