Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34147322-34147845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNBL1
Entrez Gene ID
79768
Gene Name
katanin regulatory subunit B1 like 1
Gene Aliases
C15orf29
UniGene
Hs.633566
Gene Chromosome Location
Chr.15: 34140674-34210146 on Build GRCh38
Transcripts
NM_024713.2 XM_005254668.3 XM_017022571.1 XM_017022572.1 XM_017022573.1 XM_017022574.1 XM_017022575.1

SNP Detail

SNP ID Chromosome Location
rs770241291 Chr.15: 34147346-34147346 on Build GRCh38
rs201403507 Chr.15: 34147487-34147487 on Build GRCh38
rs756890707 Chr.15: 34147457-34147457 on Build GRCh38
rs773912306 Chr.15: 34147347-34147347 on Build GRCh38
rs935801562 Chr.15: 34147443-34147443 on Build GRCh38
rs950125268 Chr.15: 34147565-34147565 on Build GRCh38
rs1046020688 Chr.15: 34147581-34147581 on Build GRCh38
rs780394428 Chr.15: 34147463-34147463 on Build GRCh38
rs962567745 Chr.15: 34147492-34147492 on Build GRCh38
rs898476822 Chr.15: 34147684-34147684 on Build GRCh38
rs756015153 Chr.15: 34147352-34147352 on Build GRCh38
rs569691509 Chr.15: 34147678-34147678 on Build GRCh38
rs899482640 Chr.15: 34147655-34147655 on Build GRCh38
rs1028802260 Chr.15: 34147777-34147777 on Build GRCh38
rs536283658 Chr.15: 34147407-34147407 on Build GRCh38
rs916106090 Chr.15: 34147465-34147465 on Build GRCh38
rs1041536861 Chr.15: 34147592-34147592 on Build GRCh38
rs149971727 Chr.15: 34147620-34147620 on Build GRCh38
rs917313464 Chr.15: 34147563-34147563 on Build GRCh38
rs1003920853 Chr.15: 34147761-34147761 on Build GRCh38
rs777444047 Chr.15: 34147352-34147354 on Build GRCh38
rs563217484 Chr.15: 34147754-34147754 on Build GRCh38
rs1000518315 Chr.15: 34147628-34147628 on Build GRCh38
rs937355438 Chr.15: 34147543-34147543 on Build GRCh38
rs765021681 Chr.15: 34147432-34147432 on Build GRCh38
rs369718128 Chr.15: 34147414-34147414 on Build GRCh38
rs778719568 Chr.15: 34147763-34147763 on Build GRCh38
rs202128690 Chr.15: 34147488-34147488 on Build GRCh38
rs755141963 Chr.15: 34147480-34147480 on Build GRCh38
rs749063148 Chr.15: 34147351-34147352 on Build GRCh38
rs73376164 Chr.15: 34147725-34147725 on Build GRCh38
rs760585192 Chr.15: 34147405-34147405 on Build GRCh38
rs372467198 Chr.15: 34147449-34147449 on Build GRCh38
rs559182293 Chr.15: 34147601-34147601 on Build GRCh38
rs139279812 Chr.15: 34147454-34147454 on Build GRCh38
rs75426827 Chr.15: 34147558-34147558 on Build GRCh38
rs887316540 Chr.15: 34147587-34147587 on Build GRCh38
rs747173629 Chr.15: 34147478-34147478 on Build GRCh38
rs953225747 Chr.15: 34147796-34147796 on Build GRCh38
rs188976465 Chr.15: 34147814-34147814 on Build GRCh38
rs750533927 Chr.15: 34147441-34147441 on Build GRCh38
rs967026502 Chr.15: 34147371-34147371 on Build GRCh38
rs1015507296 Chr.15: 34147787-34147787 on Build GRCh38
rs767258189 Chr.15: 34147382-34147382 on Build GRCh38
rs758910903 Chr.15: 34147370-34147370 on Build GRCh38
rs920586224 Chr.15: 34147513-34147513 on Build GRCh38
rs757292160 Chr.15: 34147429-34147429 on Build GRCh38
rs774999787 Chr.15: 34147384-34147384 on Build GRCh38
rs530124944 Chr.15: 34147473-34147473 on Build GRCh38
rs1038404482 Chr.15: 34147656-34147656 on Build GRCh38
rs572894416 Chr.15: 34147507-34147507 on Build GRCh38

View Full Product Details