Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38473283-38473851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98B
Entrez Gene ID
283742
Gene Name
family with sequence similarity 98 member B
Gene Aliases
-
UniGene
Hs.625289
Gene Chromosome Location
Chr.15: 38454127-38487710 on Build GRCh38
Transcripts
NM_173611.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3500701 c.602C>T p.S201L Chr.15: 38473575-38473575 on Build GRCh38
960987 c.572G>A p.G191E Chr.15: 38473545-38473545 on Build GRCh38
7367565 c.552G>T p.K184N Chr.15: 38473525-38473525 on Build GRCh38
960988 c.572G>A p.G191E Chr.15: 38473545-38473545 on Build GRCh38
4499732 c.548C>T p.S183L Chr.15: 38473521-38473521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770147755 Chr.15: 38473458-38473458 on Build GRCh38
rs1011720559 Chr.15: 38473377-38473377 on Build GRCh38
rs761365550 Chr.15: 38473521-38473521 on Build GRCh38
rs1048341961 Chr.15: 38473511-38473511 on Build GRCh38
rs1011836647 Chr.15: 38473795-38473795 on Build GRCh38
rs771321555 Chr.15: 38473491-38473491 on Build GRCh38
rs55861557 Chr.15: 38473337-38473337 on Build GRCh38
rs1021330020 Chr.15: 38473335-38473335 on Build GRCh38
rs139669477 Chr.15: 38473505-38473505 on Build GRCh38
rs187994626 Chr.15: 38473460-38473460 on Build GRCh38
rs764696390 Chr.15: 38473550-38473550 on Build GRCh38
rs370979515 Chr.15: 38473636-38473636 on Build GRCh38
rs997628701 Chr.15: 38473673-38473673 on Build GRCh38
rs371947818 Chr.15: 38473602-38473602 on Build GRCh38
rs538197003 Chr.15: 38473693-38473693 on Build GRCh38
rs745988465 Chr.15: 38473386-38473386 on Build GRCh38
rs984551599 Chr.15: 38473640-38473640 on Build GRCh38
rs907723141 Chr.15: 38473663-38473663 on Build GRCh38
rs992688210 Chr.15: 38473445-38473445 on Build GRCh38
rs752908526 Chr.15: 38473608-38473608 on Build GRCh38
rs1050680642 Chr.15: 38473314-38473314 on Build GRCh38
rs572123590 Chr.15: 38473765-38473765 on Build GRCh38
rs573885858 Chr.15: 38473563-38473563 on Build GRCh38
rs1034089013 Chr.15: 38473797-38473797 on Build GRCh38
rs893405204 Chr.15: 38473350-38473351 on Build GRCh38
rs546377161 Chr.15: 38473356-38473356 on Build GRCh38
rs1040457381 Chr.15: 38473746-38473746 on Build GRCh38
rs938508798 Chr.15: 38473430-38473430 on Build GRCh38
rs569984690 Chr.15: 38473497-38473497 on Build GRCh38
rs992656811 Chr.15: 38473817-38473817 on Build GRCh38
rs749797212 Chr.15: 38473485-38473485 on Build GRCh38
rs75394677 Chr.15: 38473695-38473695 on Build GRCh38
rs959810884 Chr.15: 38473815-38473815 on Build GRCh38
rs979847470 Chr.15: 38473419-38473419 on Build GRCh38
rs201011129 Chr.15: 38473515-38473515 on Build GRCh38
rs766223410 Chr.15: 38473585-38473585 on Build GRCh38
rs537349939 Chr.15: 38473498-38473498 on Build GRCh38
rs988429310 Chr.15: 38473311-38473311 on Build GRCh38
rs145748473 Chr.15: 38473782-38473782 on Build GRCh38
rs755206687 Chr.15: 38473601-38473601 on Build GRCh38
rs748286460 Chr.15: 38473455-38473455 on Build GRCh38
rs767266136 Chr.15: 38473622-38473622 on Build GRCh38
rs999269455 Chr.15: 38473421-38473421 on Build GRCh38
rs542933524 Chr.15: 38473810-38473810 on Build GRCh38
rs923373933 Chr.15: 38473752-38473752 on Build GRCh38
rs892046431 Chr.15: 38473708-38473708 on Build GRCh38
rs751401448 Chr.15: 38473595-38473595 on Build GRCh38
rs780016334 Chr.15: 38473616-38473616 on Build GRCh38
rs62002939 Chr.15: 38473826-38473826 on Build GRCh38
rs980199854 Chr.15: 38473793-38473793 on Build GRCh38
rs750734369 Chr.15: 38473429-38473429 on Build GRCh38
rs375860714 Chr.15: 38473729-38473729 on Build GRCh38
rs751280530 Chr.15: 38473539-38473540 on Build GRCh38
rs918498504 Chr.15: 38473451-38473451 on Build GRCh38
rs769687413 Chr.15: 38473584-38473584 on Build GRCh38
rs887748802 Chr.15: 38473534-38473534 on Build GRCh38
rs1010405750 Chr.15: 38473738-38473738 on Build GRCh38
rs553168904 Chr.15: 38473725-38473725 on Build GRCh38
rs180855780 Chr.15: 38473792-38473792 on Build GRCh38
rs571349249 Chr.15: 38473413-38473413 on Build GRCh38
rs778318368 Chr.15: 38473484-38473484 on Build GRCh38
rs545047330 Chr.15: 38473756-38473756 on Build GRCh38
rs1030492530 Chr.15: 38473703-38473703 on Build GRCh38
rs754726883 Chr.15: 38473549-38473550 on Build GRCh38
rs538243180 Chr.15: 38473437-38473437 on Build GRCh38

View Full Product Details