Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59114443-59114909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB2
Entrez Gene ID
9133
Gene Name
cyclin B2
Gene Aliases
HsT17299
UniGene
Hs.194698
Gene Chromosome Location
Chr.15: 59105085-59125045 on Build GRCh38
Transcripts
NM_004701.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622924 c.497G>A p.R166H Chr.15: 59114776-59114776 on Build GRCh38
7417304 c.445C>T p.Q149* Chr.15: 59114724-59114724 on Build GRCh38
5849457 c.269G>A p.G90D Chr.15: 59114445-59114445 on Build GRCh38
7447940 c.362A>G p.D121G Chr.15: 59114538-59114538 on Build GRCh38
4570478 c.276T>G p.S92S Chr.15: 59114452-59114452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779994699 Chr.15: 59114546-59114546 on Build GRCh38
rs975888979 Chr.15: 59114664-59114664 on Build GRCh38
rs765464570 Chr.15: 59114802-59114802 on Build GRCh38
rs887489295 Chr.15: 59114621-59114621 on Build GRCh38
rs778079480 Chr.15: 59114638-59114638 on Build GRCh38
rs371430210 Chr.15: 59114648-59114648 on Build GRCh38
rs534109952 Chr.15: 59114586-59114586 on Build GRCh38
rs750456709 Chr.15: 59114617-59114617 on Build GRCh38
rs775841822 Chr.15: 59114595-59114595 on Build GRCh38
rs200052341 Chr.15: 59114737-59114737 on Build GRCh38
rs979385275 Chr.15: 59114735-59114735 on Build GRCh38
rs770327276 Chr.15: 59114787-59114787 on Build GRCh38
rs760877193 Chr.15: 59114608-59114608 on Build GRCh38
rs778950981 Chr.15: 59114660-59114660 on Build GRCh38
rs759942264 Chr.15: 59114483-59114483 on Build GRCh38
rs748353573 Chr.15: 59114468-59114468 on Build GRCh38
rs1048672738 Chr.15: 59114874-59114874 on Build GRCh38
rs749080700 Chr.15: 59114485-59114485 on Build GRCh38
rs865848383 Chr.15: 59114503-59114503 on Build GRCh38
rs200635946 Chr.15: 59114715-59114715 on Build GRCh38
rs776389426 Chr.15: 59114691-59114691 on Build GRCh38
rs754691646 Chr.15: 59114629-59114629 on Build GRCh38
rs781621511 Chr.15: 59114819-59114819 on Build GRCh38
rs779036918 Chr.15: 59114740-59114740 on Build GRCh38
rs762576167 Chr.15: 59114778-59114778 on Build GRCh38
rs937993546 Chr.15: 59114759-59114759 on Build GRCh38
rs760418573 Chr.15: 59114801-59114801 on Build GRCh38
rs550919161 Chr.15: 59114486-59114486 on Build GRCh38
rs145845320 Chr.15: 59114723-59114723 on Build GRCh38
rs760508792 Chr.15: 59114883-59114883 on Build GRCh38
rs200615810 Chr.15: 59114800-59114800 on Build GRCh38
rs1007712455 Chr.15: 59114624-59114624 on Build GRCh38
rs770999254 Chr.15: 59114745-59114745 on Build GRCh38
rs763591966 Chr.15: 59114507-59114507 on Build GRCh38
rs770530308 Chr.15: 59114866-59114866 on Build GRCh38
rs766450641 Chr.15: 59114718-59114718 on Build GRCh38
rs772482349 Chr.15: 59114594-59114594 on Build GRCh38
rs144432316 Chr.15: 59114769-59114769 on Build GRCh38
rs374481652 Chr.15: 59114706-59114706 on Build GRCh38
rs763836666 Chr.15: 59114609-59114609 on Build GRCh38
rs776096009 Chr.15: 59114610-59114610 on Build GRCh38
rs748017216 Chr.15: 59114687-59114687 on Build GRCh38
rs756373517 Chr.15: 59114557-59114557 on Build GRCh38
rs140205864 Chr.15: 59114592-59114592 on Build GRCh38
rs536661238 Chr.15: 59114537-59114537 on Build GRCh38
rs761438863 Chr.15: 59114614-59114614 on Build GRCh38
rs768878112 Chr.15: 59114689-59114689 on Build GRCh38
rs772568414 Chr.15: 59114666-59114666 on Build GRCh38
rs762486728 Chr.15: 59114716-59114716 on Build GRCh38
rs747420379 Chr.15: 59114685-59114685 on Build GRCh38
rs764812051 Chr.15: 59114616-59114616 on Build GRCh38
rs748469810 Chr.15: 59114772-59114772 on Build GRCh38
rs752169210 Chr.15: 59114844-59114844 on Build GRCh38
rs148986291 Chr.15: 59114760-59114760 on Build GRCh38
rs770234668 Chr.15: 59114860-59114860 on Build GRCh38
rs781400947 Chr.15: 59114771-59114771 on Build GRCh38
rs370067045 Chr.15: 59114814-59114814 on Build GRCh38
rs775189310 Chr.15: 59114494-59114494 on Build GRCh38
rs779995640 Chr.15: 59114843-59114843 on Build GRCh38
rs767211640 Chr.15: 59114728-59114728 on Build GRCh38
rs781678337 Chr.15: 59114850-59114850 on Build GRCh38
rs757393271 Chr.15: 59114650-59114650 on Build GRCh38
rs758400651 Chr.15: 59114618-59114618 on Build GRCh38
rs896571394 Chr.15: 59114547-59114547 on Build GRCh38
rs879247556 Chr.15: 59114695-59114695 on Build GRCh38
rs141219534 Chr.15: 59114804-59114804 on Build GRCh38
rs767834376 Chr.15: 59114493-59114493 on Build GRCh38
rs370099852 Chr.15: 59114880-59114880 on Build GRCh38
rs748553937 Chr.15: 59114855-59114855 on Build GRCh38
rs949483720 Chr.15: 59114790-59114790 on Build GRCh38
rs750619117 Chr.15: 59114832-59114832 on Build GRCh38
rs745308502 Chr.15: 59114876-59114876 on Build GRCh38
rs369976556 Chr.15: 59114708-59114708 on Build GRCh38
rs536086391 Chr.15: 59114871-59114871 on Build GRCh38
rs143904498 Chr.15: 59114722-59114722 on Build GRCh38
rs775816244 Chr.15: 59114683-59114683 on Build GRCh38
rs2306785 Chr.15: 59114579-59114579 on Build GRCh38
rs766362424 Chr.15: 59114619-59114619 on Build GRCh38
rs747445408 Chr.15: 59114765-59114765 on Build GRCh38
rs574882835 Chr.15: 59114729-59114729 on Build GRCh38
rs772960032 Chr.15: 59114776-59114776 on Build GRCh38
rs774800920 Chr.15: 59114481-59114481 on Build GRCh38
rs16941036 Chr.15: 59114475-59114475 on Build GRCh38
rs528221556 Chr.15: 59114865-59114865 on Build GRCh38
rs759377590 Chr.15: 59114788-59114788 on Build GRCh38
rs139296509 Chr.15: 59114476-59114476 on Build GRCh38
rs773329622 Chr.15: 59114474-59114474 on Build GRCh38
rs201147134 Chr.15: 59114528-59114528 on Build GRCh38
rs146057559 Chr.15: 59114578-59114578 on Build GRCh38
rs756679957 Chr.15: 59114527-59114527 on Build GRCh38
rs554757385 Chr.15: 59114548-59114548 on Build GRCh38
rs755457310 Chr.15: 59114849-59114849 on Build GRCh38
rs771930078 Chr.15: 59114878-59114878 on Build GRCh38
rs758420783 Chr.15: 59114758-59114758 on Build GRCh38
rs758510627 Chr.15: 59114837-59114837 on Build GRCh38
rs761370106 Chr.15: 59114704-59114704 on Build GRCh38
rs144226046 Chr.15: 59114511-59114511 on Build GRCh38
rs151178240 Chr.15: 59114789-59114789 on Build GRCh38
rs201017479 Chr.15: 59114628-59114628 on Build GRCh38
rs760361771 Chr.15: 59114499-59114499 on Build GRCh38
rs200017322 Chr.15: 59114470-59114470 on Build GRCh38
rs185419194 Chr.15: 59114663-59114663 on Build GRCh38
rs758310424 Chr.15: 59114542-59114542 on Build GRCh38
rs759811496 Chr.15: 59114775-59114775 on Build GRCh38

View Full Product Details