Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66846960-66847312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA7
Entrez Gene ID
766
Gene Name
carbonic anhydrase 7
Gene Aliases
CAVII
UniGene
Hs.37014
Gene Chromosome Location
Chr.16: 66844379-66854149 on Build GRCh38
Transcripts
NM_001014435.1 NM_005182.2 XM_005256136.3 XM_011523311.1 XM_011523312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6617128 c.238G>A p.V80M Chr.16: 66847227-66847227 on Build GRCh38
4515146 c.99C>T p.I33I Chr.16: 66847088-66847088 on Build GRCh38
1709270 c.67C>T p.P23S Chr.16: 66847056-66847056 on Build GRCh38
2993802 c.194G>A p.G65D Chr.16: 66847183-66847183 on Build GRCh38
4468738 c.155C>T p.S52F Chr.16: 66847144-66847144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773943619 Chr.16: 66847234-66847234 on Build GRCh38
rs376478290 Chr.16: 66847000-66847000 on Build GRCh38
rs924435928 Chr.16: 66847119-66847119 on Build GRCh38
rs756416928 Chr.16: 66847277-66847277 on Build GRCh38
rs767213597 Chr.16: 66847159-66847159 on Build GRCh38
rs759430331 Chr.16: 66847131-66847131 on Build GRCh38
rs758044639 Chr.16: 66847002-66847002 on Build GRCh38
rs780398265 Chr.16: 66847168-66847168 on Build GRCh38
rs117738342 Chr.16: 66847054-66847054 on Build GRCh38
rs35351220 Chr.16: 66847267-66847267 on Build GRCh38
rs756990569 Chr.16: 66847036-66847036 on Build GRCh38
rs749366504 Chr.16: 66847059-66847059 on Build GRCh38
rs967022072 Chr.16: 66847268-66847268 on Build GRCh38
rs770143403 Chr.16: 66847052-66847052 on Build GRCh38
rs752716714 Chr.16: 66847151-66847151 on Build GRCh38
rs762942456 Chr.16: 66847227-66847227 on Build GRCh38
rs373632795 Chr.16: 66846995-66846995 on Build GRCh38
rs777536968 Chr.16: 66847015-66847015 on Build GRCh38
rs750130377 Chr.16: 66847269-66847269 on Build GRCh38
rs759322502 Chr.16: 66846987-66846987 on Build GRCh38
rs773546271 Chr.16: 66847220-66847220 on Build GRCh38
rs772167245 Chr.16: 66847217-66847217 on Build GRCh38
rs574009034 Chr.16: 66847261-66847261 on Build GRCh38
rs751144147 Chr.16: 66847155-66847155 on Build GRCh38
rs761469094 Chr.16: 66847247-66847247 on Build GRCh38
rs372289581 Chr.16: 66847226-66847226 on Build GRCh38
rs199547005 Chr.16: 66847118-66847118 on Build GRCh38
rs765077126 Chr.16: 66847137-66847137 on Build GRCh38
rs776229316 Chr.16: 66847124-66847124 on Build GRCh38
rs762529587 Chr.16: 66847153-66847153 on Build GRCh38
rs774537213 Chr.16: 66847088-66847088 on Build GRCh38
rs1054633106 Chr.16: 66847207-66847207 on Build GRCh38
rs267604592 Chr.16: 66847222-66847222 on Build GRCh38
rs756567978 Chr.16: 66847166-66847166 on Build GRCh38
rs767073021 Chr.16: 66847265-66847265 on Build GRCh38
rs781369944 Chr.16: 66847037-66847037 on Build GRCh38
rs778926555 Chr.16: 66847214-66847214 on Build GRCh38
rs764129605 Chr.16: 66847233-66847233 on Build GRCh38
rs763648531 Chr.16: 66847154-66847154 on Build GRCh38
rs374514983 Chr.16: 66847021-66847021 on Build GRCh38
rs149281889 Chr.16: 66847187-66847187 on Build GRCh38
rs755505176 Chr.16: 66847206-66847206 on Build GRCh38
rs1017616874 Chr.16: 66847191-66847191 on Build GRCh38
rs868323251 Chr.16: 66847074-66847074 on Build GRCh38
rs746250502 Chr.16: 66847049-66847049 on Build GRCh38
rs761977983 Chr.16: 66847107-66847107 on Build GRCh38
rs577842491 Chr.16: 66847085-66847085 on Build GRCh38
rs750740487 Chr.16: 66847160-66847160 on Build GRCh38
rs138466773 Chr.16: 66847156-66847156 on Build GRCh38
rs748237884 Chr.16: 66847215-66847215 on Build GRCh38

View Full Product Details