Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70257815-70258370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARS
Entrez Gene ID
16
Gene Name
alanyl-tRNA synthetase
Gene Aliases
CMT2N, EIEE29
UniGene
Hs.315137
Gene Chromosome Location
Chr.16: 70252295-70289552 on Build GRCh38
Transcripts
NM_001605.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4910628 c.2013C>T p.C671C Chr.16: 70258197-70258197 on Build GRCh38
4062308 c.1995C>A p.A665A Chr.16: 70258215-70258215 on Build GRCh38
6394890 c.2039A>T p.Q680L Chr.16: 70258171-70258171 on Build GRCh38
1379274 c.2130_2131insC p.S711fs*11 Chr.16: 70258079-70258080 on Build GRCh38
6145193 c.2132C>G p.S711C Chr.16: 70258078-70258078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009856121 Chr.16: 70258339-70258339 on Build GRCh38
rs761043713 Chr.16: 70258127-70258127 on Build GRCh38
rs371048516 Chr.16: 70258162-70258162 on Build GRCh38
rs777219686 Chr.16: 70258166-70258166 on Build GRCh38
rs767814087 Chr.16: 70258071-70258071 on Build GRCh38
rs1048484771 Chr.16: 70257886-70257886 on Build GRCh38
rs767543083 Chr.16: 70258124-70258124 on Build GRCh38
rs199523744 Chr.16: 70258267-70258267 on Build GRCh38
rs745695764 Chr.16: 70258185-70258185 on Build GRCh38
rs115882953 Chr.16: 70258044-70258044 on Build GRCh38
rs769571418 Chr.16: 70258190-70258190 on Build GRCh38
rs746709009 Chr.16: 70258104-70258104 on Build GRCh38
rs952674709 Chr.16: 70257927-70257931 on Build GRCh38
rs775476419 Chr.16: 70258191-70258191 on Build GRCh38
rs936208264 Chr.16: 70258295-70258295 on Build GRCh38
rs780604407 Chr.16: 70258080-70258080 on Build GRCh38
rs756524514 Chr.16: 70258077-70258077 on Build GRCh38
rs145056270 Chr.16: 70258213-70258213 on Build GRCh38
rs781347519 Chr.16: 70258193-70258193 on Build GRCh38
rs932772553 Chr.16: 70258016-70258016 on Build GRCh38
rs749859755 Chr.16: 70257997-70257997 on Build GRCh38
rs558355775 Chr.16: 70258279-70258279 on Build GRCh38
rs757898507 Chr.16: 70258164-70258164 on Build GRCh38
rs901122160 Chr.16: 70257956-70257956 on Build GRCh38
rs768249695 Chr.16: 70258002-70258002 on Build GRCh38
rs374254987 Chr.16: 70258214-70258214 on Build GRCh38
rs926140963 Chr.16: 70258280-70258280 on Build GRCh38
rs771580598 Chr.16: 70258099-70258099 on Build GRCh38
rs114248390 Chr.16: 70257903-70257903 on Build GRCh38
rs190032705 Chr.16: 70257913-70257913 on Build GRCh38
rs1000059044 Chr.16: 70258324-70258324 on Build GRCh38
rs754391789 Chr.16: 70258081-70258081 on Build GRCh38
rs888421133 Chr.16: 70257901-70257901 on Build GRCh38
rs778226080 Chr.16: 70258088-70258088 on Build GRCh38
rs757357575 Chr.16: 70258066-70258066 on Build GRCh38
rs1049688571 Chr.16: 70258275-70258275 on Build GRCh38
rs201097032 Chr.16: 70258092-70258092 on Build GRCh38
rs908524380 Chr.16: 70257970-70257970 on Build GRCh38
rs889636771 Chr.16: 70258011-70258011 on Build GRCh38
rs182660362 Chr.16: 70258007-70258007 on Build GRCh38
rs761976449 Chr.16: 70258122-70258122 on Build GRCh38
rs760490353 Chr.16: 70258022-70258022 on Build GRCh38
rs762839576 Chr.16: 70258193-70258193 on Build GRCh38
rs756650795 Chr.16: 70257992-70257992 on Build GRCh38
rs776471315 Chr.16: 70258105-70258105 on Build GRCh38
rs777499923 Chr.16: 70258101-70258101 on Build GRCh38
rs750769245 Chr.16: 70258073-70258073 on Build GRCh38
rs766834986 Chr.16: 70258131-70258131 on Build GRCh38
rs780283444 Chr.16: 70257836-70257836 on Build GRCh38
rs1024713953 Chr.16: 70257845-70257845 on Build GRCh38
rs773483658 Chr.16: 70258207-70258207 on Build GRCh38
rs766959020 Chr.16: 70258272-70258272 on Build GRCh38
rs368191672 Chr.16: 70258121-70258121 on Build GRCh38
rs540154414 Chr.16: 70258276-70258276 on Build GRCh38
rs984065144 Chr.16: 70257957-70257957 on Build GRCh38
rs751676787 Chr.16: 70258065-70258065 on Build GRCh38
rs891576819 Chr.16: 70258337-70258337 on Build GRCh38
rs531257362 Chr.16: 70258157-70258157 on Build GRCh38
rs561892715 Chr.16: 70257918-70257918 on Build GRCh38
rs759539939 Chr.16: 70258051-70258051 on Build GRCh38
rs771766993 Chr.16: 70258013-70258013 on Build GRCh38
rs371114066 Chr.16: 70258183-70258183 on Build GRCh38
rs904743547 Chr.16: 70258087-70258087 on Build GRCh38
rs759414844 Chr.16: 70258113-70258113 on Build GRCh38
rs528932248 Chr.16: 70258221-70258221 on Build GRCh38
rs754266971 Chr.16: 70258140-70258140 on Build GRCh38
rs866956348 Chr.16: 70258163-70258163 on Build GRCh38
rs764070785 Chr.16: 70258054-70258054 on Build GRCh38
rs1015550480 Chr.16: 70258247-70258247 on Build GRCh38
rs367806417 Chr.16: 70258085-70258085 on Build GRCh38
rs772813834 Chr.16: 70258017-70258017 on Build GRCh38
rs1033993225 Chr.16: 70257907-70257907 on Build GRCh38
rs377378009 Chr.16: 70258126-70258126 on Build GRCh38
rs769098292 Chr.16: 70257999-70258000 on Build GRCh38
rs566411724 Chr.16: 70258268-70258268 on Build GRCh38
rs563959372 Chr.16: 70258141-70258141 on Build GRCh38
rs936240936 Chr.16: 70258098-70258098 on Build GRCh38
rs933860672 Chr.16: 70258273-70258273 on Build GRCh38
rs149777415 Chr.16: 70258200-70258200 on Build GRCh38
rs376280288 Chr.16: 70258182-70258182 on Build GRCh38
rs368044691 Chr.16: 70258216-70258216 on Build GRCh38
rs746654032 Chr.16: 70258177-70258177 on Build GRCh38
rs1037071978 Chr.16: 70258218-70258218 on Build GRCh38
rs769778018 Chr.16: 70258114-70258114 on Build GRCh38
rs374062105 Chr.16: 70258118-70258118 on Build GRCh38
rs373121705 Chr.16: 70258005-70258005 on Build GRCh38
rs763540477 Chr.16: 70258263-70258263 on Build GRCh38
rs984593263 Chr.16: 70258210-70258210 on Build GRCh38
rs758890265 Chr.16: 70258260-70258260 on Build GRCh38
rs970935447 Chr.16: 70257862-70257862 on Build GRCh38
rs147301964 Chr.16: 70257989-70257989 on Build GRCh38
rs570266481 Chr.16: 70258328-70258328 on Build GRCh38
rs574019434 Chr.16: 70257958-70257958 on Build GRCh38
rs766251338 Chr.16: 70258032-70258032 on Build GRCh38
rs753213408 Chr.16: 70258224-70258224 on Build GRCh38
rs867164461 Chr.16: 70258219-70258219 on Build GRCh38
rs1036936309 Chr.16: 70257933-70257933 on Build GRCh38
rs754212203 Chr.16: 70258215-70258215 on Build GRCh38
rs369616839 Chr.16: 70257995-70257995 on Build GRCh38

View Full Product Details