Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87360016-87360495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO31
Entrez Gene ID
79791
Gene Name
F-box protein 31
Gene Aliases
FBX14, FBXO14, Fbx31, MRT45, pp2386
UniGene
Hs.567582
Gene Chromosome Location
Chr.16: 87326987-87392107 on Build GRCh38
Transcripts
NM_001282683.1 NM_024735.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5066796 c.383G>T p.G128V Chr.16: 87360324-87360324 on Build GRCh38
4063424 c.400G>A p.V134I Chr.16: 87360307-87360307 on Build GRCh38
3691234 c.394C>T p.R132W Chr.16: 87360313-87360313 on Build GRCh38
3718505 c.392G>T p.C131F Chr.16: 87360315-87360315 on Build GRCh38
4600204 c.383G>A p.G128D Chr.16: 87360324-87360324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768118812 Chr.16: 87360425-87360425 on Build GRCh38
rs377320531 Chr.16: 87360439-87360439 on Build GRCh38
rs186994099 Chr.16: 87360436-87360436 on Build GRCh38
rs76089114 Chr.16: 87360403-87360403 on Build GRCh38
rs557650745 Chr.16: 87360151-87360151 on Build GRCh38
rs973155370 Chr.16: 87360093-87360093 on Build GRCh38
rs770449346 Chr.16: 87360302-87360302 on Build GRCh38
rs370916343 Chr.16: 87360308-87360308 on Build GRCh38
rs778618544 Chr.16: 87360307-87360307 on Build GRCh38
rs183734112 Chr.16: 87360167-87360167 on Build GRCh38
rs746480158 Chr.16: 87360300-87360300 on Build GRCh38
rs752289082 Chr.16: 87360282-87360282 on Build GRCh38
rs933984408 Chr.16: 87360123-87360123 on Build GRCh38
rs1051671456 Chr.16: 87360142-87360142 on Build GRCh38
rs958368181 Chr.16: 87360150-87360150 on Build GRCh38
rs772382132 Chr.16: 87360333-87360333 on Build GRCh38
rs189167855 Chr.16: 87360272-87360272 on Build GRCh38
rs958483283 Chr.16: 87360460-87360460 on Build GRCh38
rs753764661 Chr.16: 87360353-87360353 on Build GRCh38
rs767980650 Chr.16: 87360409-87360409 on Build GRCh38
rs753291406 Chr.16: 87360431-87360433 on Build GRCh38
rs774857564 Chr.16: 87360247-87360247 on Build GRCh38
rs753851122 Chr.16: 87360268-87360268 on Build GRCh38
rs1043251117 Chr.16: 87360216-87360216 on Build GRCh38
rs115568905 Chr.16: 87360074-87360074 on Build GRCh38
rs909996839 Chr.16: 87360070-87360070 on Build GRCh38
rs990998463 Chr.16: 87360175-87360175 on Build GRCh38
rs147853270 Chr.16: 87360064-87360064 on Build GRCh38
rs148914157 Chr.16: 87360422-87360422 on Build GRCh38
rs367869975 Chr.16: 87360413-87360413 on Build GRCh38
rs766382861 Chr.16: 87360267-87360267 on Build GRCh38
rs758118934 Chr.16: 87360284-87360284 on Build GRCh38
rs748507325 Chr.16: 87360424-87360424 on Build GRCh38
rs763721709 Chr.16: 87360230-87360230 on Build GRCh38
rs922708428 Chr.16: 87360408-87360408 on Build GRCh38
rs922556652 Chr.16: 87360082-87360082 on Build GRCh38
rs994507859 Chr.16: 87360448-87360448 on Build GRCh38
rs920238609 Chr.16: 87360115-87360115 on Build GRCh38
rs1007375228 Chr.16: 87360181-87360181 on Build GRCh38
rs567869152 Chr.16: 87360425-87360425 on Build GRCh38
rs576025966 Chr.16: 87360227-87360227 on Build GRCh38
rs764929099 Chr.16: 87360360-87360360 on Build GRCh38
rs192869714 Chr.16: 87360398-87360398 on Build GRCh38
rs374186770 Chr.16: 87360235-87360235 on Build GRCh38
rs867692259 Chr.16: 87360347-87360347 on Build GRCh38
rs774833866 Chr.16: 87360324-87360324 on Build GRCh38
rs1042895839 Chr.16: 87360449-87360449 on Build GRCh38
rs984302848 Chr.16: 87360312-87360312 on Build GRCh38
rs889012987 Chr.16: 87360156-87360156 on Build GRCh38
rs376226613 Chr.16: 87360343-87360343 on Build GRCh38
rs749777780 Chr.16: 87360416-87360416 on Build GRCh38
rs777115705 Chr.16: 87360299-87360299 on Build GRCh38
rs763419423 Chr.16: 87360044-87360044 on Build GRCh38
rs760660906 Chr.16: 87360260-87360260 on Build GRCh38
rs753553294 Chr.16: 87360238-87360238 on Build GRCh38
rs367705961 Chr.16: 87360415-87360415 on Build GRCh38
rs183025049 Chr.16: 87360414-87360414 on Build GRCh38
rs755705456 Chr.16: 87360127-87360127 on Build GRCh38
rs998969139 Chr.16: 87360242-87360242 on Build GRCh38
rs377700727 Chr.16: 87360355-87360355 on Build GRCh38
rs780118726 Chr.16: 87360472-87360472 on Build GRCh38
rs571824181 Chr.16: 87360450-87360450 on Build GRCh38
rs752206599 Chr.16: 87360081-87360081 on Build GRCh38
rs778774243 Chr.16: 87360278-87360278 on Build GRCh38
rs533399482 Chr.16: 87360180-87360180 on Build GRCh38
rs1053890244 Chr.16: 87360303-87360303 on Build GRCh38
rs768894531 Chr.16: 87360323-87360323 on Build GRCh38
rs773602385 Chr.16: 87360335-87360335 on Build GRCh38
rs752575998 Chr.16: 87360392-87360392 on Build GRCh38
rs773740194 Chr.16: 87360041-87360041 on Build GRCh38
rs760039419 Chr.16: 87360281-87360284 on Build GRCh38
rs916159406 Chr.16: 87360040-87360040 on Build GRCh38
rs973262203 Chr.16: 87360404-87360404 on Build GRCh38
rs761922165 Chr.16: 87360326-87360326 on Build GRCh38
rs750570438 Chr.16: 87360257-87360257 on Build GRCh38
rs755561559 Chr.16: 87360420-87360420 on Build GRCh38
rs116360938 Chr.16: 87360444-87360444 on Build GRCh38

View Full Product Details