Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9857301-9857797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLP2R
Entrez Gene ID
9340
Gene Name
glucagon like peptide 2 receptor
Gene Aliases
-
UniGene
Hs.248202
Gene Chromosome Location
Chr.17: 9825508-9894163 on Build GRCh38
Transcripts
NM_004246.2 XM_005256861.3 XM_011524077.2 XM_017025339.1 XM_017025340.1 XM_017025341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2883017 c.689T>C p.V230A Chr.17: 9857500-9857500 on Build GRCh38
5648901 c.655G>A p.A219T Chr.17: 9857466-9857466 on Build GRCh38
3524153 c.666C>T p.I222I Chr.17: 9857477-9857477 on Build GRCh38
7401135 c.645G>A p.M215I Chr.17: 9857456-9857456 on Build GRCh38
5055936 c.741T>C p.N247N Chr.17: 9857552-9857552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962118855 Chr.17: 9857659-9857659 on Build GRCh38
rs368424884 Chr.17: 9857672-9857672 on Build GRCh38
rs547844690 Chr.17: 9857671-9857671 on Build GRCh38
rs376976670 Chr.17: 9857425-9857425 on Build GRCh38
rs369180942 Chr.17: 9857379-9857379 on Build GRCh38
rs72533817 Chr.17: 9857771-9857772 on Build GRCh38
rs760776316 Chr.17: 9857555-9857555 on Build GRCh38
rs768284338 Chr.17: 9857615-9857615 on Build GRCh38
rs928072236 Chr.17: 9857714-9857714 on Build GRCh38
rs570393021 Chr.17: 9857752-9857752 on Build GRCh38
rs745379708 Chr.17: 9857395-9857395 on Build GRCh38
rs1028260027 Chr.17: 9857583-9857583 on Build GRCh38
rs773046774 Chr.17: 9857543-9857543 on Build GRCh38
rs768614899 Chr.17: 9857485-9857485 on Build GRCh38
rs374504643 Chr.17: 9857584-9857584 on Build GRCh38
rs564353436 Chr.17: 9857552-9857552 on Build GRCh38
rs575133662 Chr.17: 9857439-9857439 on Build GRCh38
rs11375352 Chr.17: 9857772-9857773 on Build GRCh38
rs147244387 Chr.17: 9857534-9857534 on Build GRCh38
rs1047791144 Chr.17: 9857447-9857447 on Build GRCh38
rs778276487 Chr.17: 9857475-9857475 on Build GRCh38
rs966385527 Chr.17: 9857571-9857571 on Build GRCh38
rs141478969 Chr.17: 9857500-9857500 on Build GRCh38
rs61730822 Chr.17: 9857511-9857511 on Build GRCh38
rs779871852 Chr.17: 9857614-9857614 on Build GRCh38
rs529830698 Chr.17: 9857510-9857510 on Build GRCh38
rs779215928 Chr.17: 9857597-9857597 on Build GRCh38
rs767705014 Chr.17: 9857553-9857553 on Build GRCh38
rs778151721 Chr.17: 9857388-9857388 on Build GRCh38
rs139807788 Chr.17: 9857538-9857538 on Build GRCh38
rs939403732 Chr.17: 9857717-9857717 on Build GRCh38
rs2293100 Chr.17: 9857399-9857399 on Build GRCh38
rs905339283 Chr.17: 9857440-9857440 on Build GRCh38
rs760776468 Chr.17: 9857573-9857573 on Build GRCh38
rs529844836 Chr.17: 9857629-9857629 on Build GRCh38
rs114514574 Chr.17: 9857437-9857437 on Build GRCh38
rs1019233499 Chr.17: 9857518-9857518 on Build GRCh38
rs374782907 Chr.17: 9857617-9857617 on Build GRCh38
rs558957337 Chr.17: 9857365-9857365 on Build GRCh38
rs546202420 Chr.17: 9857529-9857529 on Build GRCh38
rs755676522 Chr.17: 9857651-9857651 on Build GRCh38
rs767641586 Chr.17: 9857454-9857454 on Build GRCh38
rs376287843 Chr.17: 9857595-9857595 on Build GRCh38
rs983963904 Chr.17: 9857600-9857600 on Build GRCh38
rs559072312 Chr.17: 9857621-9857621 on Build GRCh38
rs943563460 Chr.17: 9857404-9857404 on Build GRCh38
rs199887462 Chr.17: 9857772-9857772 on Build GRCh38
rs540970913 Chr.17: 9857612-9857612 on Build GRCh38
rs768680298 Chr.17: 9857424-9857424 on Build GRCh38
rs139958183 Chr.17: 9857508-9857508 on Build GRCh38
rs981405492 Chr.17: 9857735-9857735 on Build GRCh38
rs761533585 Chr.17: 9857426-9857426 on Build GRCh38
rs530547212 Chr.17: 9857678-9857678 on Build GRCh38
rs756662981 Chr.17: 9857474-9857474 on Build GRCh38
rs752742094 Chr.17: 9857467-9857467 on Build GRCh38
rs562490708 Chr.17: 9857417-9857417 on Build GRCh38
rs773909315 Chr.17: 9857401-9857401 on Build GRCh38
rs745461178 Chr.17: 9857416-9857416 on Build GRCh38
rs774015683 Chr.17: 9857493-9857493 on Build GRCh38
rs964995797 Chr.17: 9857653-9857653 on Build GRCh38
rs140320330 Chr.17: 9857680-9857680 on Build GRCh38
rs535331789 Chr.17: 9857778-9857778 on Build GRCh38
rs143364780 Chr.17: 9857422-9857422 on Build GRCh38
rs946659916 Chr.17: 9857764-9857764 on Build GRCh38
rs150841329 Chr.17: 9857507-9857507 on Build GRCh38
rs763924736 Chr.17: 9857387-9857387 on Build GRCh38
rs61730821 Chr.17: 9857575-9857575 on Build GRCh38
rs370670488 Chr.17: 9857607-9857607 on Build GRCh38
rs776739218 Chr.17: 9857531-9857531 on Build GRCh38
rs796313189 Chr.17: 9857718-9857718 on Build GRCh38
rs199790348 Chr.17: 9857768-9857769 on Build GRCh38
rs973650507 Chr.17: 9857665-9857665 on Build GRCh38
rs61730823 Chr.17: 9857438-9857438 on Build GRCh38

View Full Product Details