Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44854526-44855103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFTUD2
Entrez Gene ID
9343
Gene Name
elongation factor Tu GTP binding domain containing 2
Gene Aliases
MFDGA, MFDM, SNRNP116, Snrp116, Snu114, U5-116KD
UniGene
Hs.151787
Gene Chromosome Location
Chr.17: 44850287-44899625 on Build GRCh38
Transcripts
NM_001142605.1 NM_001258353.1 NM_001258354.1 NM_004247.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5679530 c.2230A>G p.I744V Chr.17: 44854585-44854585 on Build GRCh38
3819754 c.2103G>C p.E701D Chr.17: 44854947-44854947 on Build GRCh38
1383725 c.2127G>A p.W709* Chr.17: 44854923-44854923 on Build GRCh38
3191574 c.2189G>A p.R730H Chr.17: 44854626-44854626 on Build GRCh38
160657 c.2188C>T p.R730C Chr.17: 44854627-44854627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371064748 Chr.17: 44854856-44854856 on Build GRCh38
rs749662563 Chr.17: 44854899-44854899 on Build GRCh38
rs776337931 Chr.17: 44854689-44854689 on Build GRCh38
rs547368508 Chr.17: 44854599-44854599 on Build GRCh38
rs772149927 Chr.17: 44854621-44854621 on Build GRCh38
rs766868272 Chr.17: 44855015-44855015 on Build GRCh38
rs1021039218 Chr.17: 44854843-44854843 on Build GRCh38
rs866491557 Chr.17: 44855078-44855078 on Build GRCh38
rs751168355 Chr.17: 44854995-44854995 on Build GRCh38
rs543603470 Chr.17: 44855058-44855058 on Build GRCh38
rs1044142510 Chr.17: 44854702-44854702 on Build GRCh38
rs2289672 Chr.17: 44854876-44854876 on Build GRCh38
rs372838025 Chr.17: 44854586-44854586 on Build GRCh38
rs747435119 Chr.17: 44855038-44855038 on Build GRCh38
rs745394259 Chr.17: 44854834-44854834 on Build GRCh38
rs936231771 Chr.17: 44854768-44854768 on Build GRCh38
rs779188601 Chr.17: 44854892-44854892 on Build GRCh38
rs766590111 Chr.17: 44854686-44854686 on Build GRCh38
rs368881340 Chr.17: 44854713-44854713 on Build GRCh38
rs769726529 Chr.17: 44854927-44854927 on Build GRCh38
rs754910520 Chr.17: 44854582-44854582 on Build GRCh38
rs373080968 Chr.17: 44854810-44854810 on Build GRCh38
rs761414449 Chr.17: 44855000-44855000 on Build GRCh38
rs189240355 Chr.17: 44854898-44854898 on Build GRCh38
rs755370465 Chr.17: 44854890-44854890 on Build GRCh38
rs112687260 Chr.17: 44854726-44854726 on Build GRCh38
rs371614000 Chr.17: 44854598-44854598 on Build GRCh38
rs758355093 Chr.17: 44854728-44854728 on Build GRCh38
rs201915172 Chr.17: 44854727-44854727 on Build GRCh38
rs753663094 Chr.17: 44854580-44854580 on Build GRCh38
rs371274310 Chr.17: 44855027-44855027 on Build GRCh38
rs750101452 Chr.17: 44855022-44855022 on Build GRCh38
rs190738272 Chr.17: 44854878-44854878 on Build GRCh38
rs142416068 Chr.17: 44854670-44854670 on Build GRCh38
rs755735051 Chr.17: 44854841-44854841 on Build GRCh38
rs554543064 Chr.17: 44855024-44855024 on Build GRCh38
rs116064629 Chr.17: 44854823-44854823 on Build GRCh38
rs558789762 Chr.17: 44855072-44855072 on Build GRCh38
rs777353434 Chr.17: 44854592-44854592 on Build GRCh38
rs917129609 Chr.17: 44855010-44855010 on Build GRCh38
rs763758806 Chr.17: 44854731-44854731 on Build GRCh38
rs529734245 Chr.17: 44854619-44854619 on Build GRCh38
rs759399590 Chr.17: 44854703-44854703 on Build GRCh38
rs770866863 Chr.17: 44854915-44854915 on Build GRCh38
rs572335936 Chr.17: 44854920-44854920 on Build GRCh38
rs183211282 Chr.17: 44854889-44854889 on Build GRCh38
rs759516915 Chr.17: 44855032-44855033 on Build GRCh38
rs549396378 Chr.17: 44854715-44854715 on Build GRCh38
rs552742192 Chr.17: 44854908-44854908 on Build GRCh38
rs746160734 Chr.17: 44855048-44855048 on Build GRCh38
rs899992378 Chr.17: 44854818-44854819 on Build GRCh38
rs759610669 Chr.17: 44854926-44854926 on Build GRCh38
rs1057518932 Chr.17: 44855003-44855003 on Build GRCh38
rs559081677 Chr.17: 44854607-44854607 on Build GRCh38
rs917013519 Chr.17: 44854672-44854672 on Build GRCh38
rs772561995 Chr.17: 44854897-44854897 on Build GRCh38
rs775415787 Chr.17: 44854935-44854935 on Build GRCh38
rs369821280 Chr.17: 44854591-44854591 on Build GRCh38
rs749275564 Chr.17: 44854863-44854864 on Build GRCh38
rs34474715 Chr.17: 44854560-44854561 on Build GRCh38
rs377195057 Chr.17: 44855034-44855034 on Build GRCh38
rs375605344 Chr.17: 44855035-44855035 on Build GRCh38
rs752451748 Chr.17: 44854863-44854863 on Build GRCh38
rs760825462 Chr.17: 44854547-44854547 on Build GRCh38
rs377040821 Chr.17: 44854587-44854587 on Build GRCh38
rs936049525 Chr.17: 44855041-44855041 on Build GRCh38
rs749635729 Chr.17: 44854601-44854601 on Build GRCh38
rs905552008 Chr.17: 44854819-44854819 on Build GRCh38
rs769205757 Chr.17: 44854913-44854913 on Build GRCh38
rs1014993537 Chr.17: 44854857-44854857 on Build GRCh38
rs764096756 Chr.17: 44854980-44854980 on Build GRCh38
rs935661136 Chr.17: 44854821-44854821 on Build GRCh38
rs756729771 Chr.17: 44854652-44854652 on Build GRCh38
rs376668474 Chr.17: 44854983-44854983 on Build GRCh38
rs780879118 Chr.17: 44854879-44854879 on Build GRCh38
rs192858231 Chr.17: 44854962-44854962 on Build GRCh38
rs866187948 Chr.17: 44854585-44854585 on Build GRCh38
rs766145232 Chr.17: 44854553-44854553 on Build GRCh38
rs370198047 Chr.17: 44855043-44855043 on Build GRCh38
rs773266149 Chr.17: 44854647-44854647 on Build GRCh38
rs8067689 Chr.17: 44854825-44854825 on Build GRCh38
rs774867992 Chr.17: 44854613-44854613 on Build GRCh38

View Full Product Details