Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42466259-42466815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0A1
Entrez Gene ID
535
Gene Name
ATPase H+ transporting V0 subunit a1
Gene Aliases
ATP6N1, ATP6N1A, Stv1, VPP1, Vph1, a1
UniGene
Hs.463074
Gene Chromosome Location
Chr.17: 42458844-42522579 on Build GRCh38
Transcripts
NM_001130020.1 NM_001130021.1 NM_005177.3 XM_005257459.1 XM_005257461.1 XM_005257463.2 XM_011524908.1 XM_017024766.1 XM_017024767.1 XM_017024768.1 XM_017024769.1 XM_017024770.1 XM_017024771.1 XM_017024772.1 XM_017024773.1 XM_017024774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4664450 c.145C>T p.R49W Chr.17: 42466456-42466456 on Build GRCh38
4833567 c.196+1G>C p.? Chr.17: 42466508-42466508 on Build GRCh38
3387901 c.184G>T p.D62Y Chr.17: 42466495-42466495 on Build GRCh38
1710308 c.196C>T p.R66* Chr.17: 42466507-42466507 on Build GRCh38
1302843 c.124C>G p.P42A Chr.17: 42466435-42466435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003905987 Chr.17: 42466680-42466680 on Build GRCh38
rs865900473 Chr.17: 42466741-42466741 on Build GRCh38
rs1037619831 Chr.17: 42466579-42466579 on Build GRCh38
rs1047862260 Chr.17: 42466626-42466626 on Build GRCh38
rs768424560 Chr.17: 42466531-42466531 on Build GRCh38
rs996778573 Chr.17: 42466639-42466639 on Build GRCh38
rs760907084 Chr.17: 42466386-42466386 on Build GRCh38
rs1027211635 Chr.17: 42466642-42466642 on Build GRCh38
rs908189868 Chr.17: 42466326-42466326 on Build GRCh38
rs776894379 Chr.17: 42466556-42466556 on Build GRCh38
rs759571746 Chr.17: 42466281-42466282 on Build GRCh38
rs557420611 Chr.17: 42466773-42466773 on Build GRCh38
rs769407335 Chr.17: 42466518-42466518 on Build GRCh38
rs568585054 Chr.17: 42466548-42466548 on Build GRCh38
rs988806906 Chr.17: 42466764-42466764 on Build GRCh38
rs1033256584 Chr.17: 42466323-42466323 on Build GRCh38
rs1033233195 Chr.17: 42466736-42466736 on Build GRCh38
rs778230728 Chr.17: 42466458-42466458 on Build GRCh38
rs765218025 Chr.17: 42466420-42466420 on Build GRCh38
rs956504729 Chr.17: 42466747-42466747 on Build GRCh38
rs780928938 Chr.17: 42466488-42466488 on Build GRCh38
rs756984374 Chr.17: 42466484-42466484 on Build GRCh38
rs766527684 Chr.17: 42466400-42466400 on Build GRCh38
rs1022696268 Chr.17: 42466705-42466705 on Build GRCh38
rs938187145 Chr.17: 42466447-42466447 on Build GRCh38
rs371946847 Chr.17: 42466496-42466496 on Build GRCh38
rs185561680 Chr.17: 42466720-42466720 on Build GRCh38
rs959849653 Chr.17: 42466718-42466718 on Build GRCh38
rs112416627 Chr.17: 42466532-42466532 on Build GRCh38
rs1042506897 Chr.17: 42466530-42466530 on Build GRCh38
rs749276092 Chr.17: 42466524-42466524 on Build GRCh38
rs1030059829 Chr.17: 42466792-42466792 on Build GRCh38
rs747331227 Chr.17: 42466476-42466476 on Build GRCh38
rs746682443 Chr.17: 42466411-42466412 on Build GRCh38
rs775674726 Chr.17: 42466457-42466457 on Build GRCh38
rs1033361453 Chr.17: 42466711-42466711 on Build GRCh38
rs1008925408 Chr.17: 42466601-42466601 on Build GRCh38
rs774844474 Chr.17: 42466521-42466521 on Build GRCh38
rs982515393 Chr.17: 42466322-42466322 on Build GRCh38
rs754431278 Chr.17: 42466414-42466415 on Build GRCh38
rs970396638 Chr.17: 42466337-42466337 on Build GRCh38
rs928951510 Chr.17: 42466614-42466614 on Build GRCh38
rs372654749 Chr.17: 42466534-42466534 on Build GRCh38
rs774643445 Chr.17: 42466706-42466706 on Build GRCh38
rs888299715 Chr.17: 42466658-42466658 on Build GRCh38
rs181312180 Chr.17: 42466604-42466604 on Build GRCh38
rs574868527 Chr.17: 42466632-42466632 on Build GRCh38
rs749509986 Chr.17: 42466699-42466699 on Build GRCh38
rs536115891 Chr.17: 42466577-42466577 on Build GRCh38
rs374011218 Chr.17: 42466407-42466407 on Build GRCh38
rs570998180 Chr.17: 42466409-42466409 on Build GRCh38
rs934088578 Chr.17: 42466526-42466526 on Build GRCh38
rs112804782 Chr.17: 42466455-42466455 on Build GRCh38
rs982313066 Chr.17: 42466368-42466368 on Build GRCh38

View Full Product Details