Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60709017-60709386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
UniGene
Hs.655028
Gene Chromosome Location
Chr.17: 60677811-61392838 on Build GRCh38
Transcripts
NM_001099432.1 NM_001320470.1 NM_017679.3 XM_005257472.1 XM_011524939.1 XM_011524940.1 XM_011524941.1 XM_011524942.2 XM_011524943.2 XM_011524944.1 XM_011524945.2 XM_011524946.2 XM_011524947.1 XM_011524948.2 XM_011524949.2 XM_011524950.1 XM_011524951.2 XM_017024783.1 XM_017024784.1 XM_017024785.1 XM_017024786.1 XM_017024787.1 XM_017024788.1 XM_017024789.1 XM_017024790.1 XM_017024791.1 XM_017024792.1 XM_017024793.1 XM_017024794.1 XM_017024795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3520361 c.319C>T p.P107S Chr.17: 60709323-60709323 on Build GRCh38
3520360 c.319C>T p.P107S Chr.17: 60709323-60709323 on Build GRCh38
6490128 c.276G>A p.L92L Chr.17: 60709280-60709280 on Build GRCh38
5556755 c.240T>C p.H80H Chr.17: 60709244-60709244 on Build GRCh38
6081901 c.294T>C p.S98S Chr.17: 60709298-60709298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754862650 Chr.17: 60709193-60709193 on Build GRCh38
rs902004593 Chr.17: 60709152-60709152 on Build GRCh38
rs748909128 Chr.17: 60709226-60709226 on Build GRCh38
rs556172730 Chr.17: 60709360-60709360 on Build GRCh38
rs1006809326 Chr.17: 60709079-60709079 on Build GRCh38
rs918749651 Chr.17: 60709043-60709043 on Build GRCh38
rs775826447 Chr.17: 60709258-60709258 on Build GRCh38
rs752528364 Chr.17: 60709326-60709326 on Build GRCh38
rs2643103 Chr.17: 60709264-60709264 on Build GRCh38
rs752722813 Chr.17: 60709202-60709202 on Build GRCh38
rs753412201 Chr.17: 60709357-60709357 on Build GRCh38
rs1041051457 Chr.17: 60709153-60709153 on Build GRCh38
rs768170416 Chr.17: 60709350-60709350 on Build GRCh38
rs375862347 Chr.17: 60709166-60709166 on Build GRCh38
rs775064542 Chr.17: 60709332-60709332 on Build GRCh38
rs746202861 Chr.17: 60709235-60709235 on Build GRCh38
rs767354992 Chr.17: 60709195-60709195 on Build GRCh38
rs931419440 Chr.17: 60709051-60709051 on Build GRCh38
rs779277894 Chr.17: 60709234-60709234 on Build GRCh38
rs889795338 Chr.17: 60709055-60709055 on Build GRCh38
rs765367069 Chr.17: 60709284-60709284 on Build GRCh38
rs375179010 Chr.17: 60709217-60709217 on Build GRCh38
rs1007748069 Chr.17: 60709070-60709070 on Build GRCh38
rs772551288 Chr.17: 60709252-60709252 on Build GRCh38
rs751329401 Chr.17: 60709192-60709192 on Build GRCh38
rs745724521 Chr.17: 60709106-60709106 on Build GRCh38
rs766099487 Chr.17: 60709191-60709191 on Build GRCh38
rs888006228 Chr.17: 60709044-60709044 on Build GRCh38
rs1048438534 Chr.17: 60709052-60709052 on Build GRCh38
rs34712615 Chr.17: 60709320-60709320 on Build GRCh38
rs767397116 Chr.17: 60709323-60709323 on Build GRCh38
rs761969945 Chr.17: 60709278-60709278 on Build GRCh38
rs753724841 Chr.17: 60709338-60709339 on Build GRCh38
rs368119215 Chr.17: 60709230-60709230 on Build GRCh38
rs773552597 Chr.17: 60709296-60709296 on Build GRCh38
rs1018220913 Chr.17: 60709136-60709136 on Build GRCh38
rs768670626 Chr.17: 60709271-60709271 on Build GRCh38
rs777449012 Chr.17: 60709216-60709216 on Build GRCh38
rs928492886 Chr.17: 60709352-60709352 on Build GRCh38
rs532663592 Chr.17: 60709336-60709336 on Build GRCh38
rs553923170 Chr.17: 60709102-60709102 on Build GRCh38
rs533891007 Chr.17: 60709270-60709270 on Build GRCh38
rs543256883 Chr.17: 60709259-60709259 on Build GRCh38

View Full Product Details