Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 54951605-54952164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX11
Entrez Gene ID
1353
Gene Name
COX11, cytochrome c oxidase copper chaperone
Gene Aliases
COX11P
UniGene
Hs.591171
Gene Chromosome Location
Chr.17: 54951898-54971024 on Build GRCh38
Transcripts
NR_027941.2 NR_027942.2 NR_135677.1 XM_017024194.1 XM_017024195.1 XM_017024196.1 NM_001321174.1 NM_001321175.1 NM_001321176.1 NM_005486.2 XM_017024002.1

Gene Symbol
TOM1L1
Entrez Gene ID
10040
Gene Name
target of myb1 like 1 membrane trafficking protein
Gene Aliases
SRCASM
UniGene
Hs.153504
Gene Chromosome Location
Chr.17: 54900691-54961967 on Build GRCh38
Transcripts
NR_027941.2 NR_027942.2 NR_135677.1 XM_017024194.1 XM_017024195.1 XM_017024196.1 NM_001321174.1 NM_001321175.1 NM_001321176.1 NM_005486.2 XM_017024002.1

SNP Detail

SNP ID Chromosome Location
rs1033122632 Chr.17: 54951953-54951953 on Build GRCh38
rs1023222791 Chr.17: 54951823-54951823 on Build GRCh38
rs375479079 Chr.17: 54951925-54951925 on Build GRCh38
rs1046275380 Chr.17: 54951783-54951783 on Build GRCh38
rs527499409 Chr.17: 54951973-54951973 on Build GRCh38
rs957822472 Chr.17: 54951966-54951966 on Build GRCh38
rs961400319 Chr.17: 54952133-54952133 on Build GRCh38
rs1010585388 Chr.17: 54951737-54951737 on Build GRCh38
rs555820651 Chr.17: 54951894-54951894 on Build GRCh38
rs1023613301 Chr.17: 54952052-54952052 on Build GRCh38
rs762416010 Chr.17: 54951929-54951929 on Build GRCh38
rs770631962 Chr.17: 54951661-54951661 on Build GRCh38
rs954701621 Chr.17: 54951892-54951892 on Build GRCh38
rs188962470 Chr.17: 54951681-54951681 on Build GRCh38
rs949040913 Chr.17: 54952018-54952018 on Build GRCh38
rs1002018453 Chr.17: 54951801-54951802 on Build GRCh38
rs975588386 Chr.17: 54951742-54951742 on Build GRCh38
rs960273794 Chr.17: 54951782-54951782 on Build GRCh38
rs976840673 Chr.17: 54951883-54951883 on Build GRCh38
rs961430603 Chr.17: 54951878-54951878 on Build GRCh38
rs1003322508 Chr.17: 54951755-54951755 on Build GRCh38
rs543460118 Chr.17: 54951839-54951839 on Build GRCh38
rs4468693 Chr.17: 54952015-54952015 on Build GRCh38
rs932554577 Chr.17: 54951674-54951674 on Build GRCh38
rs1037801035 Chr.17: 54951814-54951814 on Build GRCh38
rs888678059 Chr.17: 54952055-54952055 on Build GRCh38
rs780285763 Chr.17: 54952041-54952041 on Build GRCh38
rs899274207 Chr.17: 54951858-54951858 on Build GRCh38
rs993581317 Chr.17: 54951873-54951873 on Build GRCh38
rs138577382 Chr.17: 54952022-54952022 on Build GRCh38
rs984101864 Chr.17: 54951861-54951861 on Build GRCh38
rs3087650 Chr.17: 54951967-54951967 on Build GRCh38
rs58896794 Chr.17: 54951907-54951907 on Build GRCh38
rs568073092 Chr.17: 54951744-54951744 on Build GRCh38
rs937910566 Chr.17: 54952065-54952065 on Build GRCh38
rs370625860 Chr.17: 54951993-54951993 on Build GRCh38
rs776423815 Chr.17: 54951730-54951730 on Build GRCh38
rs987735341 Chr.17: 54951710-54951710 on Build GRCh38
rs549801962 Chr.17: 54951738-54951738 on Build GRCh38
rs905026866 Chr.17: 54951799-54951799 on Build GRCh38
rs902215004 Chr.17: 54951964-54951964 on Build GRCh38
rs566014536 Chr.17: 54951772-54951772 on Build GRCh38
rs1013545298 Chr.17: 54951821-54951821 on Build GRCh38
rs922427391 Chr.17: 54951897-54951897 on Build GRCh38
rs531296967 Chr.17: 54952079-54952079 on Build GRCh38
rs539707858 Chr.17: 54951773-54951773 on Build GRCh38
rs576336966 Chr.17: 54951838-54951838 on Build GRCh38
rs558185796 Chr.17: 54951830-54951830 on Build GRCh38
rs373314987 Chr.17: 54951682-54951682 on Build GRCh38
rs913566360 Chr.17: 54951777-54951777 on Build GRCh38
rs1005790232 Chr.17: 54952098-54952098 on Build GRCh38
rs1042062861 Chr.17: 54951932-54951932 on Build GRCh38
rs985747817 Chr.17: 54951923-54951923 on Build GRCh38
rs1015573760 Chr.17: 54951866-54951866 on Build GRCh38
rs1047805989 Chr.17: 54952132-54952132 on Build GRCh38
rs929386331 Chr.17: 54952123-54952123 on Build GRCh38
rs181364222 Chr.17: 54951754-54951754 on Build GRCh38
rs567321424 Chr.17: 54951752-54951752 on Build GRCh38
rs149333382 Chr.17: 54952111-54952111 on Build GRCh38
rs553993790 Chr.17: 54951767-54951767 on Build GRCh38

View Full Product Details