Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16659810-16660366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM7
Entrez Gene ID
79086
Gene Name
small integral membrane protein 7
Gene Aliases
C19orf42
UniGene
Hs.356467
Gene Chromosome Location
Chr.19: 16630741-16660177 on Build GRCh38
Transcripts
NM_001300925.1 NM_024104.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4521217 c.10G>A p.D4N Chr.19: 16660101-16660101 on Build GRCh38
7281481 c.68+4A>C p.? Chr.19: 16659955-16659955 on Build GRCh38
225401 c.8G>A p.G3E Chr.19: 16660103-16660103 on Build GRCh38
3822222 c.18G>A p.L6L Chr.19: 16660093-16660093 on Build GRCh38
5762991 c.64A>C p.K22Q Chr.19: 16659963-16659963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780143949 Chr.19: 16659970-16659970 on Build GRCh38
rs561514818 Chr.19: 16659884-16659884 on Build GRCh38
rs779886806 Chr.19: 16660036-16660036 on Build GRCh38
rs753667777 Chr.19: 16660096-16660096 on Build GRCh38
rs567819741 Chr.19: 16660345-16660345 on Build GRCh38
rs772687473 Chr.19: 16660149-16660149 on Build GRCh38
rs1017843596 Chr.19: 16660267-16660267 on Build GRCh38
rs1004194445 Chr.19: 16659880-16659880 on Build GRCh38
rs755164565 Chr.19: 16660040-16660040 on Build GRCh38
rs961403044 Chr.19: 16660265-16660265 on Build GRCh38
rs761065056 Chr.19: 16659942-16659942 on Build GRCh38
rs780833755 Chr.19: 16659917-16659917 on Build GRCh38
rs750308169 Chr.19: 16660027-16660027 on Build GRCh38
rs772037107 Chr.19: 16659921-16659921 on Build GRCh38
rs903062338 Chr.19: 16659945-16659945 on Build GRCh38
rs767377514 Chr.19: 16660024-16660024 on Build GRCh38
rs753715663 Chr.19: 16660280-16660280 on Build GRCh38
rs906859767 Chr.19: 16659941-16659941 on Build GRCh38
rs1022538148 Chr.19: 16660173-16660173 on Build GRCh38
rs766204300 Chr.19: 16660095-16660095 on Build GRCh38
rs774907116 Chr.19: 16659933-16659933 on Build GRCh38
rs113593313 Chr.19: 16659858-16659858 on Build GRCh38
rs996988870 Chr.19: 16660187-16660187 on Build GRCh38
rs770340486 Chr.19: 16660139-16660139 on Build GRCh38
rs748697754 Chr.19: 16659984-16659984 on Build GRCh38
rs770914845 Chr.19: 16659916-16659916 on Build GRCh38
rs11555682 Chr.19: 16660105-16660105 on Build GRCh38
rs753594591 Chr.19: 16660128-16660128 on Build GRCh38
rs750386355 Chr.19: 16660085-16660085 on Build GRCh38
rs971115750 Chr.19: 16659869-16659869 on Build GRCh38
rs974013292 Chr.19: 16660101-16660101 on Build GRCh38
rs751080840 Chr.19: 16660019-16660019 on Build GRCh38
rs963891693 Chr.19: 16660037-16660037 on Build GRCh38
rs929649140 Chr.19: 16659900-16659900 on Build GRCh38
rs201191626 Chr.19: 16660117-16660117 on Build GRCh38
rs771435869 Chr.19: 16660113-16660113 on Build GRCh38
rs781079606 Chr.19: 16659959-16659959 on Build GRCh38
rs768784994 Chr.19: 16659981-16659981 on Build GRCh38
rs375384050 Chr.19: 16660155-16660155 on Build GRCh38
rs754722201 Chr.19: 16660099-16660099 on Build GRCh38
rs781749876 Chr.19: 16660115-16660115 on Build GRCh38
rs777893054 Chr.19: 16660109-16660109 on Build GRCh38
rs201703542 Chr.19: 16660065-16660065 on Build GRCh38
rs891456688 Chr.19: 16660330-16660330 on Build GRCh38
rs908560583 Chr.19: 16660076-16660076 on Build GRCh38
rs374562152 Chr.19: 16659896-16659896 on Build GRCh38
rs773724100 Chr.19: 16660007-16660007 on Build GRCh38
rs147730191 Chr.19: 16659950-16659951 on Build GRCh38
rs919814794 Chr.19: 16660277-16660277 on Build GRCh38
rs757363553 Chr.19: 16660134-16660134 on Build GRCh38
rs761512672 Chr.19: 16660118-16660118 on Build GRCh38
rs751693301 Chr.19: 16660133-16660133 on Build GRCh38
rs376518073 Chr.19: 16660010-16660010 on Build GRCh38
rs545745635 Chr.19: 16660064-16660064 on Build GRCh38
rs752894428 Chr.19: 16660104-16660104 on Build GRCh38
rs375780814 Chr.19: 16659929-16659929 on Build GRCh38
rs763869905 Chr.19: 16660074-16660074 on Build GRCh38
rs958696579 Chr.19: 16659990-16659990 on Build GRCh38
rs763691363 Chr.19: 16659948-16659948 on Build GRCh38
rs767976557 Chr.19: 16659935-16659935 on Build GRCh38
rs780542810 Chr.19: 16660142-16660142 on Build GRCh38
rs1052831158 Chr.19: 16660329-16660329 on Build GRCh38
rs775327001 Chr.19: 16660154-16660154 on Build GRCh38
rs752332707 Chr.19: 16659946-16659946 on Build GRCh38
rs377411757 Chr.19: 16660122-16660122 on Build GRCh38
rs771529405 Chr.19: 16660120-16660120 on Build GRCh38
rs534734842 Chr.19: 16659930-16659930 on Build GRCh38
rs777757048 Chr.19: 16660132-16660132 on Build GRCh38
rs1023034353 Chr.19: 16659861-16659861 on Build GRCh38
rs62116931 Chr.19: 16660178-16660178 on Build GRCh38
rs746148694 Chr.19: 16659928-16659928 on Build GRCh38
rs573320920 Chr.19: 16659901-16659901 on Build GRCh38
rs34668869 Chr.19: 16659951-16659952 on Build GRCh38
rs894119967 Chr.19: 16659834-16659835 on Build GRCh38
rs751675385 Chr.19: 16660157-16660157 on Build GRCh38
rs375989684 Chr.19: 16659940-16659940 on Build GRCh38
rs554294243 Chr.19: 16660293-16660293 on Build GRCh38
rs749563129 Chr.19: 16660039-16660039 on Build GRCh38
rs765327170 Chr.19: 16660302-16660302 on Build GRCh38
rs772491030 Chr.19: 16660004-16660004 on Build GRCh38
rs372937095 Chr.19: 16660143-16660143 on Build GRCh38
rs372177899 Chr.19: 16659890-16659890 on Build GRCh38
rs748254028 Chr.19: 16660053-16660053 on Build GRCh38
rs766322755 Chr.19: 16660123-16660123 on Build GRCh38
rs757237224 Chr.19: 16659952-16659952 on Build GRCh38
rs975037940 Chr.19: 16660281-16660281 on Build GRCh38
rs757891500 Chr.19: 16660328-16660328 on Build GRCh38
rs760241495 Chr.19: 16660150-16660150 on Build GRCh38
rs756750611 Chr.19: 16660022-16660022 on Build GRCh38
rs958748990 Chr.19: 16659881-16659881 on Build GRCh38
rs888024546 Chr.19: 16659882-16659882 on Build GRCh38
rs779124915 Chr.19: 16660103-16660103 on Build GRCh38
rs370207299 Chr.19: 16660153-16660153 on Build GRCh38
rs774098375 Chr.19: 16660077-16660077 on Build GRCh38
rs567080472 Chr.19: 16660264-16660264 on Build GRCh38
rs1026515697 Chr.19: 16660249-16660249 on Build GRCh38
rs548735234 Chr.19: 16660112-16660112 on Build GRCh38
rs554219033 Chr.19: 16659982-16659982 on Build GRCh38
rs1015589584 Chr.19: 16660263-16660263 on Build GRCh38
rs777111974 Chr.19: 16660073-16660073 on Build GRCh38
rs751602585 Chr.19: 16659949-16659949 on Build GRCh38
rs771255419 Chr.19: 16660070-16660070 on Build GRCh38
rs746372765 Chr.19: 16660138-16660138 on Build GRCh38
rs530972947 Chr.19: 16660129-16660129 on Build GRCh38
rs939930111 Chr.19: 16659925-16659925 on Build GRCh38
rs758627120 Chr.19: 16660130-16660130 on Build GRCh38
rs764400065 Chr.19: 16660156-16660156 on Build GRCh38
rs772745495 Chr.19: 16660121-16660121 on Build GRCh38
rs551336747 Chr.19: 16659831-16659831 on Build GRCh38
rs62116932 Chr.19: 16660226-16660226 on Build GRCh38
rs772865061 Chr.19: 16659927-16659927 on Build GRCh38
rs759476944 Chr.19: 16660299-16660299 on Build GRCh38
rs535646468 Chr.19: 16660271-16660271 on Build GRCh38
rs376969628 Chr.19: 16660078-16660078 on Build GRCh38
rs967797252 Chr.19: 16659860-16659860 on Build GRCh38
rs1051096904 Chr.19: 16659913-16659913 on Build GRCh38
rs746258238 Chr.19: 16660116-16660116 on Build GRCh38
rs987743412 Chr.19: 16660266-16660266 on Build GRCh38
rs968515300 Chr.19: 16659899-16659899 on Build GRCh38
rs546947493 Chr.19: 16660254-16660254 on Build GRCh38
rs200423511 Chr.19: 16660009-16660009 on Build GRCh38
rs991526740 Chr.19: 16659883-16659883 on Build GRCh38
rs372157802 Chr.19: 16659961-16659961 on Build GRCh38
rs770357519 Chr.19: 16660151-16660151 on Build GRCh38
rs752250684 Chr.19: 16660315-16660315 on Build GRCh38
rs1049555262 Chr.19: 16659868-16659868 on Build GRCh38
rs368863801 Chr.19: 16660017-16660017 on Build GRCh38
rs921253098 Chr.19: 16659904-16659904 on Build GRCh38
rs28364716 Chr.19: 16660195-16660195 on Build GRCh38
rs770772975 Chr.19: 16660051-16660052 on Build GRCh38
rs986767493 Chr.19: 16660072-16660072 on Build GRCh38
rs956924929 Chr.19: 16659980-16659980 on Build GRCh38
rs1041519350 Chr.19: 16660144-16660144 on Build GRCh38
rs759335335 Chr.19: 16660126-16660126 on Build GRCh38
rs894508566 Chr.19: 16660171-16660171 on Build GRCh38
rs775194159 Chr.19: 16660312-16660312 on Build GRCh38
rs370384403 Chr.19: 16660125-16660125 on Build GRCh38
rs543013782 Chr.19: 16660011-16660011 on Build GRCh38
rs775346558 Chr.19: 16660015-16660015 on Build GRCh38
rs562898868 Chr.19: 16660016-16660016 on Build GRCh38
rs747542696 Chr.19: 16660066-16660066 on Build GRCh38
rs749508002 Chr.19: 16659977-16659977 on Build GRCh38
rs986904755 Chr.19: 16660240-16660240 on Build GRCh38
rs1056959900 Chr.19: 16660166-16660166 on Build GRCh38
rs60167396 Chr.19: 16659955-16659956 on Build GRCh38
rs368241585 Chr.19: 16659908-16659908 on Build GRCh38
rs746873520 Chr.19: 16659915-16659915 on Build GRCh38
rs1012840971 Chr.19: 16660174-16660174 on Build GRCh38
rs938505067 Chr.19: 16660339-16660339 on Build GRCh38
rs867053319 Chr.19: 16659847-16659847 on Build GRCh38
rs371996316 Chr.19: 16659914-16659914 on Build GRCh38
rs3833228 Chr.19: 16659954-16659955 on Build GRCh38
rs571119720 Chr.19: 16659857-16659857 on Build GRCh38
rs1043120839 Chr.19: 16660347-16660347 on Build GRCh38
rs369292799 Chr.19: 16659960-16659960 on Build GRCh38
rs531952683 Chr.19: 16660046-16660046 on Build GRCh38
rs776620101 Chr.19: 16660152-16660152 on Build GRCh38
rs781768063 Chr.19: 16660137-16660137 on Build GRCh38
rs938640380 Chr.19: 16660161-16660161 on Build GRCh38
rs375619230 Chr.19: 16660146-16660146 on Build GRCh38
rs988235758 Chr.19: 16659888-16659888 on Build GRCh38

View Full Product Details