Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55143563-55144161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM104B
Entrez Gene ID
90736
Gene Name
family with sequence similarity 104 member B
Gene Aliases
CXorf44
UniGene
Hs.415414
Gene Chromosome Location
Chr.X: 55143102-55161195 on Build GRCh38
Transcripts
NM_001166699.1 NM_138362.3 NR_030722.1 XM_017029951.1 XM_017029952.1 XM_017029953.1

SNP Detail

SNP ID Chromosome Location
rs935371274 Chr.X: 55144036-55144041 on Build GRCh38
rs1005474196 Chr.X: 55143624-55143624 on Build GRCh38
rs773501647 Chr.X: 55143748-55143748 on Build GRCh38
rs763176724 Chr.X: 55143764-55143764 on Build GRCh38
rs929946390 Chr.X: 55143864-55143864 on Build GRCh38
rs984577430 Chr.X: 55143674-55143674 on Build GRCh38
rs753635228 Chr.X: 55143865-55143865 on Build GRCh38
rs190743865 Chr.X: 55143708-55143708 on Build GRCh38
rs763345385 Chr.X: 55143957-55143960 on Build GRCh38
rs371701365 Chr.X: 55143765-55143765 on Build GRCh38
rs376275201 Chr.X: 55143769-55143769 on Build GRCh38
rs750202679 Chr.X: 55143672-55143672 on Build GRCh38
rs771601561 Chr.X: 55143904-55143904 on Build GRCh38
rs187545670 Chr.X: 55143872-55143872 on Build GRCh38
rs1011074227 Chr.X: 55144015-55144015 on Build GRCh38
rs748661269 Chr.X: 55143919-55143919 on Build GRCh38
rs778592536 Chr.X: 55143888-55143888 on Build GRCh38
rs746685216 Chr.X: 55143779-55143779 on Build GRCh38
rs773925321 Chr.X: 55143930-55143930 on Build GRCh38
rs770230698 Chr.X: 55143921-55143921 on Build GRCh38
rs142344764 Chr.X: 55143826-55143826 on Build GRCh38
rs753735140 Chr.X: 55143632-55143632 on Build GRCh38
rs759708772 Chr.X: 55144035-55144035 on Build GRCh38
rs894932645 Chr.X: 55144005-55144005 on Build GRCh38
rs762886877 Chr.X: 55144083-55144083 on Build GRCh38
rs745414700 Chr.X: 55143897-55143897 on Build GRCh38
rs140046402 Chr.X: 55143822-55143822 on Build GRCh38
rs182883175 Chr.X: 55143813-55143813 on Build GRCh38
rs985609353 Chr.X: 55143627-55143627 on Build GRCh38
rs550563666 Chr.X: 55143600-55143600 on Build GRCh38
rs201973835 Chr.X: 55144115-55144115 on Build GRCh38
rs757189490 Chr.X: 55143639-55143639 on Build GRCh38
rs773286840 Chr.X: 55143907-55143907 on Build GRCh38
rs758192030 Chr.X: 55143697-55143697 on Build GRCh38
rs770142946 Chr.X: 55143740-55143740 on Build GRCh38
rs150734068 Chr.X: 55143805-55143805 on Build GRCh38
rs1015994104 Chr.X: 55143713-55143713 on Build GRCh38
rs778619997 Chr.X: 55143643-55143643 on Build GRCh38
rs772073411 Chr.X: 55144114-55144114 on Build GRCh38
rs34236371 Chr.X: 55143797-55143798 on Build GRCh38
rs903905742 Chr.X: 55144036-55144038 on Build GRCh38
rs1052381157 Chr.X: 55144087-55144087 on Build GRCh38
rs777722893 Chr.X: 55143853-55143853 on Build GRCh38
rs771251110 Chr.X: 55143971-55143971 on Build GRCh38
rs190968793 Chr.X: 55143989-55143989 on Build GRCh38

View Full Product Details