Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70269544-70270114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARR3
Entrez Gene ID
407
Gene Name
arrestin 3
Gene Aliases
ARRX, cArr
UniGene
Hs.308
Gene Chromosome Location
Chr.X: 70268335-70281840 on Build GRCh38
Transcripts
NM_004312.2 XM_017029518.1

SNP Detail

SNP ID Chromosome Location
rs375956429 Chr.X: 70269738-70269738 on Build GRCh38
rs3818862 Chr.X: 70269680-70269680 on Build GRCh38
rs1035976491 Chr.X: 70269592-70269592 on Build GRCh38
rs374227491 Chr.X: 70269910-70269910 on Build GRCh38
rs373855053 Chr.X: 70269633-70269633 on Build GRCh38
rs753952600 Chr.X: 70269942-70269942 on Build GRCh38
rs755323057 Chr.X: 70269832-70269832 on Build GRCh38
rs991733816 Chr.X: 70269760-70269760 on Build GRCh38
rs35785088 Chr.X: 70269825-70269826 on Build GRCh38
rs774044464 Chr.X: 70270049-70270049 on Build GRCh38
rs754246901 Chr.X: 70269804-70269804 on Build GRCh38
rs113212197 Chr.X: 70269840-70269840 on Build GRCh38
rs772162596 Chr.X: 70269865-70269865 on Build GRCh38
rs370520229 Chr.X: 70269740-70269740 on Build GRCh38
rs748602483 Chr.X: 70269638-70269638 on Build GRCh38
rs774286761 Chr.X: 70269711-70269711 on Build GRCh38
rs748953247 Chr.X: 70269926-70269926 on Build GRCh38
rs867246791 Chr.X: 70270020-70270020 on Build GRCh38
rs749094995 Chr.X: 70269664-70269664 on Build GRCh38
rs768340764 Chr.X: 70269705-70269705 on Build GRCh38
rs755148106 Chr.X: 70270077-70270078 on Build GRCh38
rs959448581 Chr.X: 70269758-70269758 on Build GRCh38
rs769488154 Chr.X: 70269663-70269663 on Build GRCh38
rs960180114 Chr.X: 70269605-70269605 on Build GRCh38
rs899668208 Chr.X: 70269977-70269977 on Build GRCh38
rs779615525 Chr.X: 70269924-70269924 on Build GRCh38
rs748277235 Chr.X: 70269822-70269822 on Build GRCh38
rs745491385 Chr.X: 70269651-70269651 on Build GRCh38
rs868487934 Chr.X: 70269744-70269744 on Build GRCh38
rs780733870 Chr.X: 70269889-70269889 on Build GRCh38
rs999101203 Chr.X: 70269572-70269572 on Build GRCh38
rs1033715441 Chr.X: 70269702-70269702 on Build GRCh38
rs950020736 Chr.X: 70269872-70269872 on Build GRCh38
rs370184846 Chr.X: 70269611-70269611 on Build GRCh38
rs947457111 Chr.X: 70270031-70270031 on Build GRCh38
rs201503536 Chr.X: 70270067-70270067 on Build GRCh38
rs756083016 Chr.X: 70269579-70269579 on Build GRCh38
rs776645594 Chr.X: 70269610-70269610 on Build GRCh38
rs750506444 Chr.X: 70269726-70269726 on Build GRCh38
rs1009799533 Chr.X: 70269591-70269591 on Build GRCh38
rs773226154 Chr.X: 70269873-70269873 on Build GRCh38
rs973850774 Chr.X: 70269686-70269686 on Build GRCh38
rs1802952 Chr.X: 70269859-70269859 on Build GRCh38
rs745442722 Chr.X: 70269890-70269890 on Build GRCh38
rs770430092 Chr.X: 70269639-70269639 on Build GRCh38
rs200409414 Chr.X: 70269703-70269703 on Build GRCh38
rs919581681 Chr.X: 70269779-70269779 on Build GRCh38
rs766381934 Chr.X: 70269916-70269916 on Build GRCh38
rs767593405 Chr.X: 70269714-70269714 on Build GRCh38
rs148653835 Chr.X: 70269878-70269878 on Build GRCh38
rs917065967 Chr.X: 70269788-70269788 on Build GRCh38
rs761590846 Chr.X: 70269625-70269625 on Build GRCh38
rs368519383 Chr.X: 70270055-70270055 on Build GRCh38
rs780602455 Chr.X: 70269641-70269641 on Build GRCh38
rs766751922 Chr.X: 70269955-70269955 on Build GRCh38

View Full Product Details