Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 77899166-77899727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7B
Entrez Gene ID
1349
Gene Name
cytochrome c oxidase subunit 7B
Gene Aliases
APLCC, LSDMCA2
UniGene
Hs.522699
Gene Chromosome Location
Chr.X: 77899464-77905384 on Build GRCh38
Transcripts
NM_001866.2

SNP Detail

SNP ID Chromosome Location
rs782218608 Chr.X: 77899574-77899574 on Build GRCh38
rs782330612 Chr.X: 77899599-77899599 on Build GRCh38
rs963709282 Chr.X: 77899494-77899494 on Build GRCh38
rs782488046 Chr.X: 77899547-77899547 on Build GRCh38
rs782647960 Chr.X: 77899492-77899492 on Build GRCh38
rs782303725 Chr.X: 77899382-77899382 on Build GRCh38
rs995862116 Chr.X: 77899667-77899667 on Build GRCh38
rs948051496 Chr.X: 77899638-77899638 on Build GRCh38
rs782801997 Chr.X: 77899242-77899242 on Build GRCh38
rs782807803 Chr.X: 77899653-77899653 on Build GRCh38
rs782165808 Chr.X: 77899426-77899426 on Build GRCh38
rs1025863236 Chr.X: 77899442-77899442 on Build GRCh38
rs782574819 Chr.X: 77899613-77899613 on Build GRCh38
rs139781829 Chr.X: 77899430-77899430 on Build GRCh38
rs374380697 Chr.X: 77899626-77899626 on Build GRCh38
rs781994164 Chr.X: 77899455-77899455 on Build GRCh38
rs782507227 Chr.X: 77899359-77899359 on Build GRCh38
rs372770371 Chr.X: 77899483-77899484 on Build GRCh38
rs1012238495 Chr.X: 77899322-77899322 on Build GRCh38
rs782332231 Chr.X: 77899447-77899447 on Build GRCh38
rs1023588098 Chr.X: 77899328-77899328 on Build GRCh38
rs370681452 Chr.X: 77899560-77899560 on Build GRCh38
rs782269461 Chr.X: 77899597-77899597 on Build GRCh38
rs368265984 Chr.X: 77899519-77899519 on Build GRCh38
rs56278774 Chr.X: 77899243-77899243 on Build GRCh38
rs142805660 Chr.X: 77899247-77899247 on Build GRCh38
rs896561987 Chr.X: 77899615-77899615 on Build GRCh38
rs782000863 Chr.X: 77899641-77899641 on Build GRCh38
rs781870001 Chr.X: 77899603-77899604 on Build GRCh38
rs782112788 Chr.X: 77899491-77899491 on Build GRCh38
rs782351434 Chr.X: 77899628-77899628 on Build GRCh38
rs11552077 Chr.X: 77899533-77899533 on Build GRCh38
rs1045046198 Chr.X: 77899649-77899649 on Build GRCh38
rs946919889 Chr.X: 77899522-77899522 on Build GRCh38
rs924139812 Chr.X: 77899530-77899530 on Build GRCh38
rs781888900 Chr.X: 77899703-77899703 on Build GRCh38
rs782793919 Chr.X: 77899512-77899512 on Build GRCh38
rs782237291 Chr.X: 77899624-77899624 on Build GRCh38
rs374052875 Chr.X: 77899507-77899507 on Build GRCh38
rs782600544 Chr.X: 77899548-77899548 on Build GRCh38
rs782568175 Chr.X: 77899525-77899525 on Build GRCh38
rs781942592 Chr.X: 77899402-77899402 on Build GRCh38
rs782424056 Chr.X: 77899392-77899392 on Build GRCh38
rs782703625 Chr.X: 77899528-77899528 on Build GRCh38
rs781788728 Chr.X: 77899537-77899537 on Build GRCh38

View Full Product Details