Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136209123-136209579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHL1
Entrez Gene ID
2273
Gene Name
four and a half LIM domains 1
Gene Aliases
FHL-1, FHL1A, FHL1B, FLH1A, KYOT, RBMX1A, RBMX1B, SLIM, SLIM-1, SLIM1, SLIMMER, XMPMA
UniGene
Hs.435369
Gene Chromosome Location
Chr.X: 136146702-136211359 on Build GRCh38
Transcripts
NM_001159699.1 NM_001159700.1 NM_001159701.1 NM_001159702.2 NM_001159703.1 NM_001159704.1 NM_001167819.1 NM_001449.4 NR_027621.1 XM_006724743.2 XM_006724744.2 XM_006724745.3 XM_006724746.2 XM_006724747.2 XM_011531316.1 XM_017029357.1

SNP Detail

SNP ID Chromosome Location
rs372165290 Chr.X: 136209277-136209277 on Build GRCh38
rs756718769 Chr.X: 136209213-136209213 on Build GRCh38
rs779090125 Chr.X: 136209305-136209305 on Build GRCh38
rs1022356084 Chr.X: 136209153-136209153 on Build GRCh38
rs367592190 Chr.X: 136209314-136209314 on Build GRCh38
rs764170405 Chr.X: 136209559-136209559 on Build GRCh38
rs1039061577 Chr.X: 136209181-136209181 on Build GRCh38
rs965891572 Chr.X: 136209514-136209514 on Build GRCh38
rs762830333 Chr.X: 136209432-136209432 on Build GRCh38
rs769812399 Chr.X: 136209409-136209409 on Build GRCh38
rs991073749 Chr.X: 136209548-136209548 on Build GRCh38
rs780478822 Chr.X: 136209460-136209460 on Build GRCh38
rs751141361 Chr.X: 136209440-136209440 on Build GRCh38
rs1007536702 Chr.X: 136209175-136209175 on Build GRCh38
rs780608136 Chr.X: 136209487-136209487 on Build GRCh38
rs893395681 Chr.X: 136209143-136209143 on Build GRCh38
rs775440569 Chr.X: 136209415-136209415 on Build GRCh38
rs886044371 Chr.X: 136209251-136209251 on Build GRCh38
rs575040348 Chr.X: 136209527-136209527 on Build GRCh38
rs749911623 Chr.X: 136209459-136209459 on Build GRCh38
rs997598973 Chr.X: 136209202-136209202 on Build GRCh38
rs751174469 Chr.X: 136209208-136209208 on Build GRCh38
rs766962380 Chr.X: 136209454-136209454 on Build GRCh38
rs886042893 Chr.X: 136209321-136209321 on Build GRCh38
rs746341848 Chr.X: 136209237-136209237 on Build GRCh38
rs772013758 Chr.X: 136209338-136209338 on Build GRCh38
rs962965208 Chr.X: 136209543-136209543 on Build GRCh38
rs757498351 Chr.X: 136209550-136209550 on Build GRCh38
rs760504186 Chr.X: 136209549-136209549 on Build GRCh38
rs199818971 Chr.X: 136209351-136209351 on Build GRCh38
rs781745050 Chr.X: 136209228-136209228 on Build GRCh38
rs754255484 Chr.X: 136209468-136209468 on Build GRCh38
rs752489174 Chr.X: 136209196-136209196 on Build GRCh38
rs267606811 Chr.X: 136209392-136209392 on Build GRCh38
rs780429969 Chr.X: 136209268-136209268 on Build GRCh38
rs1004479734 Chr.X: 136209341-136209341 on Build GRCh38
rs901889534 Chr.X: 136209193-136209193 on Build GRCh38
rs756598405 Chr.X: 136209265-136209265 on Build GRCh38
rs758814784 Chr.X: 136209435-136209435 on Build GRCh38
rs373785682 Chr.X: 136209299-136209299 on Build GRCh38
rs758130406 Chr.X: 136209197-136209197 on Build GRCh38
rs886043137 Chr.X: 136209321-136209322 on Build GRCh38
rs371767332 Chr.X: 136209388-136209388 on Build GRCh38
rs761439164 Chr.X: 136209442-136209442 on Build GRCh38
rs886935189 Chr.X: 136209289-136209289 on Build GRCh38
rs749585308 Chr.X: 136209281-136209281 on Build GRCh38
rs763783551 Chr.X: 136209200-136209200 on Build GRCh38
rs952030324 Chr.X: 136209551-136209551 on Build GRCh38

View Full Product Details